Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 134283546-134283819 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLB1L3
Entrez Gene ID
112937
Gene Name
galactosidase beta 1 like 3
Gene Aliases
-
UniGene
Hs.721382
Gene Chromosome Location
Chr.11: 134275447-134321364 on Build GRCh38
Transcripts
NM_001080407.2 XM_011542566.2 XM_011542567.2 XM_011542568.2 XM_011542569.2 XM_011542570.2 XM_011542571.2 XM_017017155.1 XM_017017156.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6284267 c.528-6C>T p.? Chr.11: 134283731-134283731 on Build GRCh38
5696392 c.531G>T p.W177C Chr.11: 134283740-134283740 on Build GRCh38
6537566 c.576C>G p.S192R Chr.11: 134283785-134283785 on Build GRCh38
4145554 c.585A>T p.E195D Chr.11: 134283794-134283794 on Build GRCh38
5510557 c.592G>A p.E198K Chr.11: 134283801-134283801 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764308327 Chr.11: 134283584-134283584 on Build GRCh38
rs1034549083 Chr.11: 134283651-134283651 on Build GRCh38
rs772838127 Chr.11: 134283714-134283714 on Build GRCh38
rs560037047 Chr.11: 134283701-134283701 on Build GRCh38
rs1011498916 Chr.11: 134283781-134283781 on Build GRCh38
rs887486660 Chr.11: 134283586-134283586 on Build GRCh38
rs1012677964 Chr.11: 134283587-134283587 on Build GRCh38
rs955863996 Chr.11: 134283617-134283617 on Build GRCh38
rs776187224 Chr.11: 134283700-134283700 on Build GRCh38
rs535443828 Chr.11: 134283570-134283570 on Build GRCh38
rs748520706 Chr.11: 134283778-134283778 on Build GRCh38
rs755965170 Chr.11: 134283737-134283737 on Build GRCh38
rs746007894 Chr.11: 134283690-134283690 on Build GRCh38
rs776804316 Chr.11: 134283706-134283706 on Build GRCh38
rs746064989 Chr.11: 134283745-134283745 on Build GRCh38
rs539928115 Chr.11: 134283609-134283609 on Build GRCh38
rs758589618 Chr.11: 134283753-134283753 on Build GRCh38
rs376729461 Chr.11: 134283782-134283782 on Build GRCh38
rs372101608 Chr.11: 134283730-134283730 on Build GRCh38
rs942566934 Chr.11: 134283618-134283618 on Build GRCh38
rs923998224 Chr.11: 134283581-134283581 on Build GRCh38
rs919548765 Chr.11: 134283661-134283661 on Build GRCh38
rs970986507 Chr.11: 134283674-134283674 on Build GRCh38
rs573381094 Chr.11: 134283620-134283620 on Build GRCh38
rs752364995 Chr.11: 134283734-134283734 on Build GRCh38
rs532846433 Chr.11: 134283682-134283682 on Build GRCh38
rs768978439 Chr.11: 134283771-134283771 on Build GRCh38
rs754962406 Chr.11: 134283596-134283596 on Build GRCh38
rs201858924 Chr.11: 134283696-134283696 on Build GRCh38
rs531525435 Chr.11: 134283756-134283756 on Build GRCh38
rs374231483 Chr.11: 134283721-134283721 on Build GRCh38
rs562507189 Chr.11: 134283677-134283677 on Build GRCh38
rs910531549 Chr.11: 134283652-134283652 on Build GRCh38
rs888138557 Chr.11: 134283632-134283632 on Build GRCh38
rs1049662935 Chr.11: 134283678-134283678 on Build GRCh38
rs374634023 Chr.11: 134283702-134283702 on Build GRCh38
rs1002724182 Chr.11: 134283608-134283608 on Build GRCh38
rs1044172750 Chr.11: 134283713-134283713 on Build GRCh38
rs558008869 Chr.11: 134283618-134283618 on Build GRCh38
rs1040641631 Chr.11: 134283656-134283656 on Build GRCh38
rs993700872 Chr.11: 134283582-134283582 on Build GRCh38
rs540854989 Chr.11: 134283623-134283623 on Build GRCh38
rs201115970 Chr.11: 134283719-134283719 on Build GRCh38
rs779332653 Chr.11: 134283743-134283743 on Build GRCh38
rs550517910 Chr.11: 134283757-134283757 on Build GRCh38
rs138981320 Chr.11: 134283585-134283585 on Build GRCh38
rs562179330 Chr.11: 134283653-134283653 on Build GRCh38
rs556954327 Chr.11: 134283572-134283572 on Build GRCh38
rs492383 Chr.11: 134283624-134283624 on Build GRCh38
rs764936397 Chr.11: 134283712-134283712 on Build GRCh38
rs544198470 Chr.11: 134283663-134283663 on Build GRCh38
rs368809721 Chr.11: 134283710-134283710 on Build GRCh38
rs200592634 Chr.11: 134283785-134283785 on Build GRCh38
rs202008670 Chr.11: 134283790-134283790 on Build GRCh38
rs1023026220 Chr.11: 134283588-134283588 on Build GRCh38
rs559291086 Chr.11: 134283629-134283629 on Build GRCh38
rs765847708 Chr.11: 134283720-134283720 on Build GRCh38
rs1025284987 Chr.11: 134283628-134283628 on Build GRCh38
rs148979023 Chr.11: 134283775-134283775 on Build GRCh38
rs986245322 Chr.11: 134283634-134283634 on Build GRCh38
rs367609057 Chr.11: 134283724-134283724 on Build GRCh38

View Full Product Details