Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 168216848-168217111 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDX60
Entrez Gene ID
55601
Gene Name
DEXD/H-box helicase 60
Gene Aliases
-
UniGene
Hs.591710
Gene Chromosome Location
Chr.4: 168216291-168318807 on Build GRCh38
Transcripts
NM_017631.5 XM_011532103.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5983893 c.5082T>A p.V1694V Chr.4: 168216990-168216990 on Build GRCh38
1053010 c.5080G>T p.V1694F Chr.4: 168216992-168216992 on Build GRCh38
5060225 c.5082T>C p.V1694V Chr.4: 168216990-168216990 on Build GRCh38
7179101 c.5075A>T p.D1692V Chr.4: 168216997-168216997 on Build GRCh38
6430575 c.5069A>G p.E1690G Chr.4: 168217003-168217003 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751028354 Chr.4: 168217023-168217023 on Build GRCh38
rs763663491 Chr.4: 168216922-168216922 on Build GRCh38
rs773918250 Chr.4: 168216878-168216878 on Build GRCh38
rs537859190 Chr.4: 168216992-168216992 on Build GRCh38
rs767131414 Chr.4: 168217032-168217032 on Build GRCh38
rs375786387 Chr.4: 168217047-168217047 on Build GRCh38
rs375688526 Chr.4: 168216894-168216894 on Build GRCh38
rs747879641 Chr.4: 168216937-168216937 on Build GRCh38
rs754458944 Chr.4: 168217024-168217024 on Build GRCh38
rs747056499 Chr.4: 168217081-168217081 on Build GRCh38
rs560349056 Chr.4: 168217084-168217084 on Build GRCh38
rs755820279 Chr.4: 168217057-168217057 on Build GRCh38
rs764712951 Chr.4: 168216881-168216881 on Build GRCh38
rs747089851 Chr.4: 168216994-168216994 on Build GRCh38
rs756782109 Chr.4: 168216989-168216990 on Build GRCh38
rs1030834745 Chr.4: 168217013-168217013 on Build GRCh38
rs1005760088 Chr.4: 168216928-168216928 on Build GRCh38
rs765887495 Chr.4: 168216921-168216921 on Build GRCh38
rs755906708 Chr.4: 168216948-168216948 on Build GRCh38
rs1034736510 Chr.4: 168216922-168216922 on Build GRCh38
rs752339441 Chr.4: 168217054-168217054 on Build GRCh38
rs574017606 Chr.4: 168216926-168216926 on Build GRCh38
rs775611019 Chr.4: 168216993-168216993 on Build GRCh38
rs867643981 Chr.4: 168216985-168216985 on Build GRCh38
rs749123911 Chr.4: 168217070-168217070 on Build GRCh38
rs754615127 Chr.4: 168216930-168216930 on Build GRCh38
rs766821714 Chr.4: 168217022-168217022 on Build GRCh38
rs753371155 Chr.4: 168216939-168216942 on Build GRCh38
rs765694779 Chr.4: 168217005-168217005 on Build GRCh38
rs956486163 Chr.4: 168217045-168217045 on Build GRCh38
rs759044495 Chr.4: 168216918-168216918 on Build GRCh38
rs376016470 Chr.4: 168217074-168217074 on Build GRCh38
rs766959438 Chr.4: 168217045-168217045 on Build GRCh38
rs749208035 Chr.4: 168216988-168216988 on Build GRCh38
rs763520296 Chr.4: 168217021-168217021 on Build GRCh38
rs765786477 Chr.4: 168216885-168216885 on Build GRCh38
rs768875546 Chr.4: 168216998-168216998 on Build GRCh38
rs1011891389 Chr.4: 168216982-168216982 on Build GRCh38
rs372799055 Chr.4: 168216999-168216999 on Build GRCh38
rs373415986 Chr.4: 168217083-168217083 on Build GRCh38
rs777523331 Chr.4: 168217065-168217065 on Build GRCh38
rs752267884 Chr.4: 168216882-168216882 on Build GRCh38
rs773774011 Chr.4: 168217014-168217014 on Build GRCh38
rs1047207982 Chr.4: 168216876-168216876 on Build GRCh38
rs1023941932 Chr.4: 168217050-168217050 on Build GRCh38
rs987295471 Chr.4: 168217049-168217049 on Build GRCh38
rs369028315 Chr.4: 168216913-168216913 on Build GRCh38
rs80220511 Chr.4: 168217078-168217078 on Build GRCh38
rs780727215 Chr.4: 168216931-168216931 on Build GRCh38

View Full Product Details