Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 1632620-1632891 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DLGAP2
Entrez Gene ID
9228
Gene Name
DLG associated protein 2
Gene Aliases
DAP2, SAPAP2
UniGene
Hs.113287
Gene Chromosome Location
Chr.8: 1501366-1708476 on Build GRCh38
Transcripts
NM_001277161.1 NM_004745.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1177364 c.1446C>T p.F482F Chr.8: 1632856-1632856 on Build GRCh38
1177363 c.1512C>T p.F504F Chr.8: 1632856-1632856 on Build GRCh38
216811 c.1428G>A p.A476A Chr.8: 1632838-1632838 on Build GRCh38
4465488 c.1518C>T p.S506S Chr.8: 1632862-1632862 on Build GRCh38
3646748 c.1529C>T p.S510F Chr.8: 1632873-1632873 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764037054 Chr.8: 1632869-1632870 on Build GRCh38
rs755405597 Chr.8: 1632846-1632846 on Build GRCh38
rs527705861 Chr.8: 1632838-1632838 on Build GRCh38
rs758278309 Chr.8: 1632832-1632832 on Build GRCh38
rs759117146 Chr.8: 1632725-1632725 on Build GRCh38
rs773179434 Chr.8: 1632805-1632805 on Build GRCh38
rs779256141 Chr.8: 1632856-1632856 on Build GRCh38
rs935299984 Chr.8: 1632688-1632688 on Build GRCh38
rs997917912 Chr.8: 1632751-1632751 on Build GRCh38
rs868249344 Chr.8: 1632655-1632655 on Build GRCh38
rs772480572 Chr.8: 1632857-1632857 on Build GRCh38
rs963555125 Chr.8: 1632810-1632810 on Build GRCh38
rs545731381 Chr.8: 1632782-1632782 on Build GRCh38
rs371385968 Chr.8: 1632784-1632784 on Build GRCh38
rs377087907 Chr.8: 1632803-1632803 on Build GRCh38
rs777058938 Chr.8: 1632869-1632869 on Build GRCh38
rs1002310017 Chr.8: 1632762-1632762 on Build GRCh38
rs1052390312 Chr.8: 1632698-1632698 on Build GRCh38
rs1033753377 Chr.8: 1632765-1632765 on Build GRCh38
rs765259860 Chr.8: 1632818-1632818 on Build GRCh38
rs563340476 Chr.8: 1632774-1632774 on Build GRCh38
rs771170255 Chr.8: 1632868-1632868 on Build GRCh38
rs760904358 Chr.8: 1632806-1632806 on Build GRCh38
rs1021382957 Chr.8: 1632777-1632777 on Build GRCh38
rs886064317 Chr.8: 1632676-1632676 on Build GRCh38
rs562893257 Chr.8: 1632713-1632713 on Build GRCh38
rs774899060 Chr.8: 1632797-1632797 on Build GRCh38
rs749697759 Chr.8: 1632795-1632795 on Build GRCh38
rs893720109 Chr.8: 1632708-1632708 on Build GRCh38
rs780760175 Chr.8: 1632844-1632844 on Build GRCh38
rs1038613181 Chr.8: 1632850-1632850 on Build GRCh38
rs758050692 Chr.8: 1632783-1632783 on Build GRCh38
rs373697861 Chr.8: 1632785-1632785 on Build GRCh38
rs769132819 Chr.8: 1632796-1632796 on Build GRCh38
rs186669393 Chr.8: 1632680-1632680 on Build GRCh38
rs1046843949 Chr.8: 1632681-1632681 on Build GRCh38
rs7013788 Chr.8: 1632773-1632773 on Build GRCh38
rs370260485 Chr.8: 1632837-1632837 on Build GRCh38
rs979798568 Chr.8: 1632787-1632787 on Build GRCh38
rs923073882 Chr.8: 1632816-1632816 on Build GRCh38
rs982240696 Chr.8: 1632849-1632849 on Build GRCh38
rs371127232 Chr.8: 1632843-1632843 on Build GRCh38
rs576962906 Chr.8: 1632778-1632778 on Build GRCh38
rs898737929 Chr.8: 1632853-1632853 on Build GRCh38
rs534578969 Chr.8: 1632642-1632642 on Build GRCh38
rs761700440 Chr.8: 1632662-1632662 on Build GRCh38
rs906563069 Chr.8: 1632683-1632683 on Build GRCh38
rs899600266 Chr.8: 1632750-1632750 on Build GRCh38
rs933066417 Chr.8: 1632817-1632817 on Build GRCh38
rs375368240 Chr.8: 1632863-1632863 on Build GRCh38
rs761879955 Chr.8: 1632800-1632800 on Build GRCh38
rs1029814272 Chr.8: 1632761-1632761 on Build GRCh38
rs759460676 Chr.8: 1632813-1632813 on Build GRCh38
rs200142637 Chr.8: 1632862-1632862 on Build GRCh38
rs115628832 Chr.8: 1632748-1632748 on Build GRCh38
rs749642714 Chr.8: 1632845-1632845 on Build GRCh38
rs752613425 Chr.8: 1632829-1632829 on Build GRCh38
rs753662881 Chr.8: 1632811-1632811 on Build GRCh38
rs574463499 Chr.8: 1632719-1632719 on Build GRCh38
rs780431217 Chr.8: 1632793-1632793 on Build GRCh38
rs533480207 Chr.8: 1632749-1632749 on Build GRCh38
rs1010752422 Chr.8: 1632740-1632740 on Build GRCh38

View Full Product Details