Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 83063017-83063242 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COPS4
Entrez Gene ID
51138
Gene Name
COP9 signalosome subunit 4
Gene Aliases
CSN4
UniGene
Hs.190384
Gene Chromosome Location
Chr.4: 83035086-83075818 on Build GRCh38
Transcripts
NM_001258006.1 NM_016129.2 XM_011532019.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6043783 c.718C>G p.Q240E Chr.4: 83063078-83063078 on Build GRCh38
6866684 c.835C>T p.Q279* Chr.4: 83063195-83063195 on Build GRCh38
6306707 c.808G>T p.D270Y Chr.4: 83063168-83063168 on Build GRCh38
7305303 c.731G>A p.R244Q Chr.4: 83063091-83063091 on Build GRCh38
6634088 c.852G>A p.M284I Chr.4: 83063212-83063212 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761816763 Chr.4: 83063098-83063098 on Build GRCh38
rs750559338 Chr.4: 83063101-83063101 on Build GRCh38
rs767664893 Chr.4: 83063100-83063100 on Build GRCh38
rs1042526172 Chr.4: 83063140-83063140 on Build GRCh38
rs763446404 Chr.4: 83063060-83063060 on Build GRCh38
rs752715718 Chr.4: 83063176-83063176 on Build GRCh38
rs757799679 Chr.4: 83063191-83063191 on Build GRCh38
rs377239931 Chr.4: 83063146-83063146 on Build GRCh38
rs529877293 Chr.4: 83063071-83063071 on Build GRCh38
rs200927595 Chr.4: 83063170-83063170 on Build GRCh38
rs753752591 Chr.4: 83063158-83063158 on Build GRCh38
rs775876615 Chr.4: 83063058-83063058 on Build GRCh38
rs906707638 Chr.4: 83063143-83063143 on Build GRCh38
rs766053246 Chr.4: 83063152-83063152 on Build GRCh38
rs373901019 Chr.4: 83063194-83063194 on Build GRCh38
rs140152209 Chr.4: 83063216-83063216 on Build GRCh38
rs754951130 Chr.4: 83063165-83063165 on Build GRCh38
rs945621781 Chr.4: 83063085-83063085 on Build GRCh38
rs74891864 Chr.4: 83063073-83063073 on Build GRCh38
rs552229546 Chr.4: 83063116-83063116 on Build GRCh38
rs746611483 Chr.4: 83063206-83063206 on Build GRCh38
rs780387126 Chr.4: 83063217-83063217 on Build GRCh38

View Full Product Details