Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 221739646-221739871 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DUSP10
Entrez Gene ID
11221
Gene Name
dual specificity phosphatase 10
Gene Aliases
MKP-5, MKP5
UniGene
Hs.497822
Gene Chromosome Location
Chr.1: 221701420-221742176 on Build GRCh38
Transcripts
NM_007207.5 NR_111940.1 XM_017000147.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
137803 c.55C>T p.R19* Chr.1: 221739690-221739690 on Build GRCh38
1339036 c.21C>T p.D7D Chr.1: 221739724-221739724 on Build GRCh38
7112974 c.67C>G p.L23V Chr.1: 221739678-221739678 on Build GRCh38
7237863 c.22G>C p.D8H Chr.1: 221739723-221739723 on Build GRCh38
3803978 c.20A>T p.D7V Chr.1: 221739725-221739725 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769341595 Chr.1: 221739759-221739759 on Build GRCh38
rs766836021 Chr.1: 221739785-221739785 on Build GRCh38
rs779074182 Chr.1: 221739693-221739693 on Build GRCh38
rs748390873 Chr.1: 221739694-221739694 on Build GRCh38
rs776606563 Chr.1: 221739736-221739736 on Build GRCh38
rs757684298 Chr.1: 221739674-221739674 on Build GRCh38
rs201096194 Chr.1: 221739760-221739760 on Build GRCh38
rs1047917515 Chr.1: 221739689-221739689 on Build GRCh38
rs147859958 Chr.1: 221739745-221739745 on Build GRCh38
rs986357618 Chr.1: 221739836-221739836 on Build GRCh38
rs750728887 Chr.1: 221739681-221739681 on Build GRCh38
rs373505357 Chr.1: 221739717-221739717 on Build GRCh38
rs752361079 Chr.1: 221739673-221739673 on Build GRCh38
rs780562110 Chr.1: 221739686-221739686 on Build GRCh38
rs954564673 Chr.1: 221739804-221739804 on Build GRCh38
rs761005519 Chr.1: 221739784-221739784 on Build GRCh38
rs771011005 Chr.1: 221739723-221739723 on Build GRCh38
rs370026750 Chr.1: 221739690-221739690 on Build GRCh38
rs546322180 Chr.1: 221739724-221739724 on Build GRCh38
rs749378098 Chr.1: 221739687-221739687 on Build GRCh38
rs767852820 Chr.1: 221739678-221739678 on Build GRCh38
rs144387908 Chr.1: 221739706-221739706 on Build GRCh38
rs763668838 Chr.1: 221739762-221739762 on Build GRCh38
rs1021654993 Chr.1: 221739828-221739828 on Build GRCh38
rs772366448 Chr.1: 221739703-221739703 on Build GRCh38
rs767207636 Chr.1: 221739827-221739827 on Build GRCh38
rs959312582 Chr.1: 221739748-221739748 on Build GRCh38
rs750808897 Chr.1: 221739777-221739777 on Build GRCh38
rs760865445 Chr.1: 221739769-221739771 on Build GRCh38
rs768946454 Chr.1: 221739789-221739790 on Build GRCh38

View Full Product Details