Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 110225976-110226235 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNC4
Entrez Gene ID
3749
Gene Name
potassium voltage-gated channel subfamily C member 4
Gene Aliases
C1orf30, HKSHIIIC, KSHIIIC, KV3.4
UniGene
Hs.153521
Gene Chromosome Location
Chr.1: 110210714-110282649 on Build GRCh38
Transcripts
NM_001039574.2 NM_004978.4 NR_036437.1 XM_006710625.3 XM_006710627.3 XM_011541401.2 XM_011541402.1 XM_011541403.1 XM_011541404.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4621958 c.1741G>C p.D581H Chr.1: 110226100-110226100 on Build GRCh38
5989066 c.1690T>A p.S564T Chr.1: 110226049-110226049 on Build GRCh38
4621957 c.1741G>C p.D581H Chr.1: 110226100-110226100 on Build GRCh38
5993269 c.1723C>T p.R575* Chr.1: 110226082-110226082 on Build GRCh38
6453340 c.1633G>A p.D545N Chr.1: 110225992-110225992 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200087078 Chr.1: 110226077-110226077 on Build GRCh38
rs773560393 Chr.1: 110226145-110226145 on Build GRCh38
rs566567590 Chr.1: 110226173-110226173 on Build GRCh38
rs1039184220 Chr.1: 110226211-110226211 on Build GRCh38
rs780133268 Chr.1: 110226019-110226019 on Build GRCh38
rs989784731 Chr.1: 110226006-110226006 on Build GRCh38
rs945183018 Chr.1: 110226039-110226039 on Build GRCh38
rs752419978 Chr.1: 110226124-110226124 on Build GRCh38
rs994543122 Chr.1: 110226072-110226072 on Build GRCh38
rs779480669 Chr.1: 110226204-110226204 on Build GRCh38
rs761152942 Chr.1: 110226213-110226213 on Build GRCh38
rs137885705 Chr.1: 110226073-110226073 on Build GRCh38
rs57855778 Chr.1: 110226088-110226088 on Build GRCh38
rs150591419 Chr.1: 110226126-110226126 on Build GRCh38
rs200184574 Chr.1: 110226172-110226172 on Build GRCh38
rs200865430 Chr.1: 110226063-110226063 on Build GRCh38
rs377454400 Chr.1: 110226000-110226000 on Build GRCh38
rs777571978 Chr.1: 110226157-110226157 on Build GRCh38
rs761187683 Chr.1: 110226083-110226083 on Build GRCh38
rs967405599 Chr.1: 110226203-110226203 on Build GRCh38
rs371724703 Chr.1: 110226057-110226057 on Build GRCh38
rs777563000 Chr.1: 110226096-110226096 on Build GRCh38
rs768423888 Chr.1: 110226210-110226210 on Build GRCh38
rs775101038 Chr.1: 110226070-110226070 on Build GRCh38
rs139597550 Chr.1: 110226156-110226156 on Build GRCh38
rs943242157 Chr.1: 110226175-110226175 on Build GRCh38
rs987169758 Chr.1: 110226208-110226208 on Build GRCh38
rs957586508 Chr.1: 110226111-110226111 on Build GRCh38
rs769314818 Chr.1: 110226201-110226201 on Build GRCh38
rs781228862 Chr.1: 110226182-110226182 on Build GRCh38
rs375796717 Chr.1: 110226047-110226047 on Build GRCh38
rs898810500 Chr.1: 110226071-110226071 on Build GRCh38
rs779275404 Chr.1: 110226043-110226043 on Build GRCh38
rs767282952 Chr.1: 110226097-110226097 on Build GRCh38
rs745763348 Chr.1: 110226188-110226188 on Build GRCh38
rs748944906 Chr.1: 110226209-110226209 on Build GRCh38
rs34480599 Chr.1: 110226093-110226093 on Build GRCh38
rs149082882 Chr.1: 110226001-110226001 on Build GRCh38
rs374706301 Chr.1: 110226149-110226149 on Build GRCh38
rs766376210 Chr.1: 110226084-110226084 on Build GRCh38
rs201758498 Chr.1: 110226117-110226117 on Build GRCh38
rs751659808 Chr.1: 110226037-110226038 on Build GRCh38
rs780976462 Chr.1: 110226103-110226103 on Build GRCh38
rs201472923 Chr.1: 110226162-110226162 on Build GRCh38
rs750916005 Chr.1: 110226082-110226082 on Build GRCh38
rs371019224 Chr.1: 110226029-110226029 on Build GRCh38
rs745430301 Chr.1: 110226107-110226107 on Build GRCh38
rs762543109 Chr.1: 110226127-110226127 on Build GRCh38
rs771973719 Chr.1: 110226052-110226052 on Build GRCh38
rs754018819 Chr.1: 110226085-110226085 on Build GRCh38
rs1033288250 Chr.1: 110226179-110226179 on Build GRCh38
rs770965544 Chr.1: 110226098-110226098 on Build GRCh38
rs771171754 Chr.1: 110226214-110226214 on Build GRCh38
rs772336869 Chr.1: 110226139-110226139 on Build GRCh38
rs956244922 Chr.1: 110225997-110225997 on Build GRCh38
rs867005209 Chr.1: 110226189-110226189 on Build GRCh38
rs757615011 Chr.1: 110226094-110226094 on Build GRCh38
rs755140783 Chr.1: 110226109-110226111 on Build GRCh38
rs755186151 Chr.1: 110226036-110226036 on Build GRCh38
rs758622508 Chr.1: 110226160-110226160 on Build GRCh38
rs147798518 Chr.1: 110226059-110226059 on Build GRCh38
rs142302926 Chr.1: 110226064-110226064 on Build GRCh38
rs773067084 Chr.1: 110226056-110226056 on Build GRCh38
rs1033122106 Chr.1: 110226104-110226104 on Build GRCh38
rs754280867 Chr.1: 110226153-110226153 on Build GRCh38
rs760944501 Chr.1: 110226148-110226148 on Build GRCh38
rs373438977 Chr.1: 110225996-110225996 on Build GRCh38
rs146318129 Chr.1: 110226159-110226159 on Build GRCh38
rs781746509 Chr.1: 110226179-110226180 on Build GRCh38
rs548644103 Chr.1: 110226101-110226101 on Build GRCh38
rs150481020 Chr.1: 110226086-110226086 on Build GRCh38

View Full Product Details