Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 113869432-113869695 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GRAMD1C
Entrez Gene ID
54762
Gene Name
GRAM domain containing 1C
Gene Aliases
-
UniGene
Hs.24583
Gene Chromosome Location
Chr.3: 113828182-113947174 on Build GRCh38
Transcripts
NM_017577.4 XM_005247546.2 XM_005247547.1 XM_011512930.1 XM_017006646.1 XM_017006647.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
325432 c.181A>G p.S61G Chr.3: 113869513-113869513 on Build GRCh38
4430988 c.185C>A p.S62Y Chr.3: 113869517-113869517 on Build GRCh38
7370152 c.211T>C p.Y71H Chr.3: 113869543-113869543 on Build GRCh38
3944701 c.259+3A>G p.? Chr.3: 113869594-113869594 on Build GRCh38
6377087 c.178T>G p.S60A Chr.3: 113869510-113869510 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754410078 Chr.3: 113869627-113869627 on Build GRCh38
rs755324754 Chr.3: 113869630-113869630 on Build GRCh38
rs887783687 Chr.3: 113869632-113869632 on Build GRCh38
rs766941187 Chr.3: 113869610-113869610 on Build GRCh38
rs754231761 Chr.3: 113869642-113869643 on Build GRCh38
rs760278065 Chr.3: 113869500-113869500 on Build GRCh38
rs771822001 Chr.3: 113869472-113869472 on Build GRCh38
rs760893840 Chr.3: 113869609-113869609 on Build GRCh38
rs772579112 Chr.3: 113869485-113869485 on Build GRCh38
rs993558779 Chr.3: 113869562-113869562 on Build GRCh38
rs770819436 Chr.3: 113869502-113869502 on Build GRCh38
rs779412493 Chr.3: 113869635-113869635 on Build GRCh38
rs967937847 Chr.3: 113869483-113869483 on Build GRCh38
rs901998959 Chr.3: 113869546-113869546 on Build GRCh38
rs991832440 Chr.3: 113869646-113869646 on Build GRCh38
rs1469559 Chr.3: 113869598-113869598 on Build GRCh38
rs538758082 Chr.3: 113869476-113869479 on Build GRCh38
rs752556125 Chr.3: 113869569-113869569 on Build GRCh38
rs750439517 Chr.3: 113869602-113869602 on Build GRCh38
rs761930126 Chr.3: 113869584-113869584 on Build GRCh38
rs541486779 Chr.3: 113869658-113869658 on Build GRCh38
rs149464871 Chr.3: 113869641-113869641 on Build GRCh38
rs756247000 Chr.3: 113869604-113869604 on Build GRCh38
rs765023465 Chr.3: 113869523-113869523 on Build GRCh38
rs759271869 Chr.3: 113869517-113869517 on Build GRCh38
rs764693418 Chr.3: 113869641-113869642 on Build GRCh38
rs201700492 Chr.3: 113869510-113869510 on Build GRCh38

View Full Product Details