Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 77268702-77268971 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GALR1
Entrez Gene ID
2587
Gene Name
galanin receptor 1
Gene Aliases
GALNR, GALNR1
UniGene
Hs.272191
Gene Chromosome Location
Chr.18: 77250052-77270142 on Build GRCh38
Transcripts
NM_001480.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
398063 c.865C>T p.H289Y Chr.18: 77268717-77268717 on Build GRCh38
7369186 c.1025C>T p.P342L Chr.18: 77268877-77268877 on Build GRCh38
295543 c.972A>G p.K324K Chr.18: 77268824-77268824 on Build GRCh38
275258 c.862G>A p.A288T Chr.18: 77268714-77268714 on Build GRCh38
261066 c.997G>T p.E333* Chr.18: 77268849-77268849 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755651870 Chr.18: 77268946-77268946 on Build GRCh38
rs1013468870 Chr.18: 77268912-77268912 on Build GRCh38
rs761257157 Chr.18: 77268910-77268910 on Build GRCh38
rs1039817747 Chr.18: 77268811-77268811 on Build GRCh38
rs374047751 Chr.18: 77268861-77268861 on Build GRCh38
rs745369709 Chr.18: 77268886-77268886 on Build GRCh38
rs373846302 Chr.18: 77268906-77268906 on Build GRCh38
rs377700030 Chr.18: 77268921-77268921 on Build GRCh38
rs774918810 Chr.18: 77268769-77268769 on Build GRCh38
rs5376 Chr.18: 77268853-77268853 on Build GRCh38
rs779826907 Chr.18: 77268896-77268896 on Build GRCh38
rs771160406 Chr.18: 77268783-77268783 on Build GRCh38
rs770839108 Chr.18: 77268830-77268831 on Build GRCh38
rs757290716 Chr.18: 77268936-77268936 on Build GRCh38
rs762251608 Chr.18: 77268784-77268784 on Build GRCh38
rs1002259772 Chr.18: 77268929-77268929 on Build GRCh38
rs766501938 Chr.18: 77268815-77268815 on Build GRCh38
rs757088730 Chr.18: 77268865-77268865 on Build GRCh38
rs369126559 Chr.18: 77268889-77268889 on Build GRCh38
rs745551553 Chr.18: 77268744-77268744 on Build GRCh38
rs372077517 Chr.18: 77268945-77268945 on Build GRCh38
rs553756626 Chr.18: 77268813-77268813 on Build GRCh38
rs778691702 Chr.18: 77268723-77268723 on Build GRCh38
rs904886819 Chr.18: 77268924-77268924 on Build GRCh38
rs201229811 Chr.18: 77268736-77268736 on Build GRCh38
rs531351765 Chr.18: 77268934-77268934 on Build GRCh38
rs773398427 Chr.18: 77268908-77268908 on Build GRCh38
rs776837217 Chr.18: 77268914-77268914 on Build GRCh38
rs748559973 Chr.18: 77268899-77268899 on Build GRCh38
rs772751317 Chr.18: 77268788-77268788 on Build GRCh38
rs201172922 Chr.18: 77268926-77268926 on Build GRCh38
rs199730182 Chr.18: 77268728-77268728 on Build GRCh38
rs758387946 Chr.18: 77268731-77268731 on Build GRCh38
rs928120017 Chr.18: 77268862-77268862 on Build GRCh38
rs777506870 Chr.18: 77268859-77268859 on Build GRCh38
rs145820336 Chr.18: 77268789-77268789 on Build GRCh38
rs765432250 Chr.18: 77268820-77268820 on Build GRCh38
rs759512675 Chr.18: 77268915-77268915 on Build GRCh38
rs866904324 Chr.18: 77268849-77268849 on Build GRCh38
rs753801878 Chr.18: 77268819-77268819 on Build GRCh38
rs913882960 Chr.18: 77268839-77268839 on Build GRCh38
rs780521070 Chr.18: 77268743-77268743 on Build GRCh38
rs777666336 Chr.18: 77268732-77268732 on Build GRCh38
rs775722901 Chr.18: 77268923-77268923 on Build GRCh38
rs771275729 Chr.18: 77268911-77268911 on Build GRCh38
rs746799506 Chr.18: 77268812-77268816 on Build GRCh38
rs936533743 Chr.18: 77268876-77268876 on Build GRCh38
rs376957505 Chr.18: 77268740-77268740 on Build GRCh38
rs767338851 Chr.18: 77268938-77268938 on Build GRCh38
rs1055291363 Chr.18: 77268894-77268894 on Build GRCh38
rs773821311 Chr.18: 77268802-77268802 on Build GRCh38
rs989912271 Chr.18: 77268766-77268766 on Build GRCh38
rs113010483 Chr.18: 77268873-77268873 on Build GRCh38
rs5378 Chr.18: 77268943-77268943 on Build GRCh38
rs371953398 Chr.18: 77268751-77268751 on Build GRCh38
rs5377 Chr.18: 77268877-77268877 on Build GRCh38
rs774695515 Chr.18: 77268844-77268844 on Build GRCh38
rs769268550 Chr.18: 77268764-77268764 on Build GRCh38

View Full Product Details