Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 4408999-4409272 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHAF1A
Entrez Gene ID
10036
Gene Name
chromatin assembly factor 1 subunit A
Gene Aliases
CAF-1, CAF1, CAF1B, CAF1P150, P150
UniGene
Hs.79018
Gene Chromosome Location
Chr.19: 4402596-4448322 on Build GRCh38
Transcripts
NM_005483.2 XM_011527605.2 XM_011527606.2 XM_011527607.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
997642 c.237T>C p.C79C Chr.19: 4409036-4409036 on Build GRCh38
4429913 c.314G>A p.R105K Chr.19: 4409113-4409113 on Build GRCh38
5629097 c.266C>G p.P89R Chr.19: 4409065-4409065 on Build GRCh38
5856043 c.424_425CC>TT p.P142F Chr.19: 4409223-4409224 on Build GRCh38
6454041 c.280G>A p.G94R Chr.19: 4409079-4409079 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374609997 Chr.19: 4409039-4409039 on Build GRCh38
rs752386071 Chr.19: 4409032-4409032 on Build GRCh38
rs145582898 Chr.19: 4409168-4409168 on Build GRCh38
rs1021336376 Chr.19: 4409042-4409042 on Build GRCh38
rs141725942 Chr.19: 4409187-4409187 on Build GRCh38
rs148067278 Chr.19: 4409176-4409176 on Build GRCh38
rs780395542 Chr.19: 4409071-4409071 on Build GRCh38
rs145585672 Chr.19: 4409108-4409108 on Build GRCh38
rs754186708 Chr.19: 4409142-4409142 on Build GRCh38
rs546954156 Chr.19: 4409182-4409182 on Build GRCh38
rs765750348 Chr.19: 4409159-4409159 on Build GRCh38
rs1022490964 Chr.19: 4409251-4409251 on Build GRCh38
rs760263178 Chr.19: 4409126-4409126 on Build GRCh38
rs770861602 Chr.19: 4409111-4409111 on Build GRCh38
rs200402574 Chr.19: 4409057-4409057 on Build GRCh38
rs755091463 Chr.19: 4409214-4409214 on Build GRCh38
rs769702995 Chr.19: 4409102-4409102 on Build GRCh38
rs202229686 Chr.19: 4409078-4409078 on Build GRCh38
rs144604203 Chr.19: 4409148-4409148 on Build GRCh38
rs144939143 Chr.19: 4409066-4409066 on Build GRCh38
rs764500811 Chr.19: 4409044-4409044 on Build GRCh38
rs149070934 Chr.19: 4409144-4409144 on Build GRCh38
rs150150583 Chr.19: 4409224-4409224 on Build GRCh38
rs112641643 Chr.19: 4409156-4409156 on Build GRCh38
rs367610752 Chr.19: 4409053-4409053 on Build GRCh38
rs754166460 Chr.19: 4409052-4409052 on Build GRCh38
rs755188149 Chr.19: 4409175-4409175 on Build GRCh38
rs745861368 Chr.19: 4409200-4409200 on Build GRCh38
rs778880559 Chr.19: 4409189-4409189 on Build GRCh38
rs774179482 Chr.19: 4409115-4409115 on Build GRCh38
rs138530415 Chr.19: 4409163-4409163 on Build GRCh38
rs775109223 Chr.19: 4409118-4409118 on Build GRCh38
rs759026569 Chr.19: 4409117-4409117 on Build GRCh38
rs150043386 Chr.19: 4409065-4409065 on Build GRCh38
rs781495112 Chr.19: 4409148-4409150 on Build GRCh38
rs367896225 Chr.19: 4409234-4409234 on Build GRCh38
rs193058821 Chr.19: 4409104-4409104 on Build GRCh38
rs140374238 Chr.19: 4409167-4409167 on Build GRCh38
rs373856871 Chr.19: 4409249-4409249 on Build GRCh38

View Full Product Details