Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 6225013-6225258 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ICMT
Entrez Gene ID
23463
Gene Name
isoprenylcysteine carboxyl methyltransferase
Gene Aliases
HSTE14, MST098, MSTP098, PCCMT, PCMT, PPMT
UniGene
Hs.515688
Gene Chromosome Location
Chr.1: 6221193-6235984 on Build GRCh38
Transcripts
NM_012405.3 XM_011541140.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1715930 c.831A>G p.I277M Chr.1: 6225104-6225104 on Build GRCh38
2238266 c.697G>A p.G233S Chr.1: 6225238-6225238 on Build GRCh38
2238264 c.787T>C p.Y263H Chr.1: 6225148-6225148 on Build GRCh38
4185545 c.766A>G p.I256V Chr.1: 6225169-6225169 on Build GRCh38
4878346 c.740G>A p.R247Q Chr.1: 6225195-6225195 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs556416674 Chr.1: 6225051-6225051 on Build GRCh38
rs753689810 Chr.1: 6225236-6225236 on Build GRCh38
rs545382166 Chr.1: 6225075-6225075 on Build GRCh38
rs370297787 Chr.1: 6225068-6225068 on Build GRCh38
rs756719639 Chr.1: 6225213-6225214 on Build GRCh38
rs766290610 Chr.1: 6225161-6225161 on Build GRCh38
rs768201294 Chr.1: 6225093-6225093 on Build GRCh38
rs763778174 Chr.1: 6225106-6225106 on Build GRCh38
rs757486258 Chr.1: 6225076-6225076 on Build GRCh38
rs772928711 Chr.1: 6225052-6225052 on Build GRCh38
rs764272807 Chr.1: 6225201-6225201 on Build GRCh38
rs762149296 Chr.1: 6225216-6225216 on Build GRCh38
rs202193560 Chr.1: 6225224-6225224 on Build GRCh38
rs773035350 Chr.1: 6225134-6225134 on Build GRCh38
rs181532322 Chr.1: 6225238-6225238 on Build GRCh38
rs564071041 Chr.1: 6225119-6225119 on Build GRCh38
rs35408357 Chr.1: 6225158-6225158 on Build GRCh38
rs148244027 Chr.1: 6225092-6225092 on Build GRCh38
rs756498433 Chr.1: 6225233-6225233 on Build GRCh38
rs375617854 Chr.1: 6225065-6225065 on Build GRCh38
rs1048591952 Chr.1: 6225086-6225086 on Build GRCh38
rs773992104 Chr.1: 6225048-6225048 on Build GRCh38
rs141302565 Chr.1: 6225199-6225199 on Build GRCh38
rs773796051 Chr.1: 6225112-6225112 on Build GRCh38
rs756629497 Chr.1: 6225087-6225087 on Build GRCh38
rs370724223 Chr.1: 6225046-6225046 on Build GRCh38
rs969059488 Chr.1: 6225140-6225140 on Build GRCh38
rs760484405 Chr.1: 6225139-6225139 on Build GRCh38
rs1001914999 Chr.1: 6225164-6225164 on Build GRCh38
rs201101726 Chr.1: 6225235-6225235 on Build GRCh38
rs751043505 Chr.1: 6225179-6225181 on Build GRCh38
rs746186766 Chr.1: 6225079-6225079 on Build GRCh38
rs79611532 Chr.1: 6225066-6225066 on Build GRCh38
rs764279726 Chr.1: 6225074-6225074 on Build GRCh38
rs776614885 Chr.1: 6225187-6225187 on Build GRCh38
rs763749930 Chr.1: 6225149-6225151 on Build GRCh38
rs141918869 Chr.1: 6225210-6225210 on Build GRCh38
rs750859428 Chr.1: 6225226-6225226 on Build GRCh38
rs149108074 Chr.1: 6225089-6225089 on Build GRCh38
rs774885426 Chr.1: 6225044-6225044 on Build GRCh38
rs746380911 Chr.1: 6225034-6225034 on Build GRCh38
rs778429985 Chr.1: 6225239-6225239 on Build GRCh38
rs760548833 Chr.1: 6225059-6225059 on Build GRCh38
rs770295482 Chr.1: 6225036-6225036 on Build GRCh38
rs537892201 Chr.1: 6225174-6225174 on Build GRCh38
rs781714309 Chr.1: 6225078-6225078 on Build GRCh38
rs759332536 Chr.1: 6225200-6225200 on Build GRCh38
rs768234392 Chr.1: 6225047-6225047 on Build GRCh38

View Full Product Details