Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 141477181-141477438 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PCDHGA1
Entrez Gene ID
56114
Gene Name
protocadherin gamma subfamily A, 1
Gene Aliases
PCDH-GAMMA-A1
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141330685-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032092.1 NM_003735.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3 NM_002588.3 NM_032402.1 NM_032403.2

Gene Symbol
PCDHGA10
Entrez Gene ID
56106
Gene Name
protocadherin gamma subfamily A, 10
Gene Aliases
PCDH-GAMMA-A10
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141413176-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032092.1 NM_003735.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3 NM_002588.3 NM_032402.1 NM_032403.2

Gene Symbol
PCDHGA11
Entrez Gene ID
56105
Gene Name
protocadherin gamma subfamily A, 11
Gene Aliases
PCDH-GAMMA-A11
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141420970-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032092.1 NM_003735.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3 NM_002588.3 NM_032402.1 NM_032403.2

Gene Symbol
PCDHGA12
Entrez Gene ID
26025
Gene Name
protocadherin gamma subfamily A, 12
Gene Aliases
CDH21, FIB3, PCDH-GAMMA-A12
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141430591-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032092.1 NM_003735.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3 NM_002588.3 NM_032402.1 NM_032403.2

Gene Symbol
PCDHGA2
Entrez Gene ID
56113
Gene Name
protocadherin gamma subfamily A, 2
Gene Aliases
PCDH-GAMMA-A2
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141338760-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032092.1 NM_003735.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3 NM_002588.3 NM_032402.1 NM_032403.2

Gene Symbol
PCDHGA3
Entrez Gene ID
56112
Gene Name
protocadherin gamma subfamily A, 3
Gene Aliases
PCDH-GAMMA-A3
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141344034-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032092.1 NM_003735.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3 NM_002588.3 NM_032402.1 NM_032403.2

Gene Symbol
PCDHGA4
Entrez Gene ID
56111
Gene Name
protocadherin gamma subfamily A, 4
Gene Aliases
PCDH-GAMMA-A4
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141355025-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032092.1 NM_003735.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3 NM_002588.3 NM_032402.1 NM_032403.2

Gene Symbol
PCDHGA5
Entrez Gene ID
56110
Gene Name
protocadherin gamma subfamily A, 5
Gene Aliases
CDH-GAMMA-A5, ME3, PCDH-GAMMA-A5
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141364331-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032092.1 NM_003735.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3 NM_002588.3 NM_032402.1 NM_032403.2

Gene Symbol
PCDHGA6
Entrez Gene ID
56109
Gene Name
protocadherin gamma subfamily A, 6
Gene Aliases
PCDH-GAMMA-A6
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141374084-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032092.1 NM_003735.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3 NM_002588.3 NM_032402.1 NM_032403.2

Gene Symbol
PCDHGA7
Entrez Gene ID
56108
Gene Name
protocadherin gamma subfamily A, 7
Gene Aliases
PCDH-GAMMA-A7
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141382739-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032092.1 NM_003735.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3 NM_002588.3 NM_032402.1 NM_032403.2

Gene Symbol
PCDHGA8
Entrez Gene ID
9708
Gene Name
protocadherin gamma subfamily A, 8
Gene Aliases
PCDH-GAMMA-A8
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141391916-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032092.1 NM_003735.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3 NM_002588.3 NM_032402.1 NM_032403.2

Gene Symbol
PCDHGA9
Entrez Gene ID
56107
Gene Name
protocadherin gamma subfamily A, 9
Gene Aliases
PCDH-GAMMA-A9
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141402953-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032092.1 NM_003735.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3 NM_002588.3 NM_032402.1 NM_032403.2

Gene Symbol
PCDHGB1
Entrez Gene ID
56104
Gene Name
protocadherin gamma subfamily B, 1
Gene Aliases
PCDH-GAMMA-B1
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141350261-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032092.1 NM_003735.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3 NM_002588.3 NM_032402.1 NM_032403.2

Gene Symbol
PCDHGB2
Entrez Gene ID
56103
Gene Name
protocadherin gamma subfamily B, 2
Gene Aliases
PCDH-GAMMA-B2
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141360136-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032092.1 NM_003735.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3 NM_002588.3 NM_032402.1 NM_032403.2

Gene Symbol
PCDHGB3
Entrez Gene ID
56102
Gene Name
protocadherin gamma subfamily B, 3
Gene Aliases
PCDH-GAMMA-B3
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141370264-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032092.1 NM_003735.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3 NM_002588.3 NM_032402.1 NM_032403.2

Gene Symbol
PCDHGB4
Entrez Gene ID
8641
Gene Name
protocadherin gamma subfamily B, 4
Gene Aliases
CDH20, FIB2, PCDH-GAMMA-B4
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141387885-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032092.1 NM_003735.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3 NM_002588.3 NM_032402.1 NM_032403.2

Gene Symbol
PCDHGB5
Entrez Gene ID
56101
Gene Name
protocadherin gamma subfamily B, 5
Gene Aliases
PCDH-GAMMA-B5
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141398128-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032092.1 NM_003735.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3 NM_002588.3 NM_032402.1 NM_032403.2

Gene Symbol
PCDHGB6
Entrez Gene ID
56100
Gene Name
protocadherin gamma subfamily B, 6
Gene Aliases
PCDH-GAMMA-B6
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141408203-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032092.1 NM_003735.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3 NM_002588.3 NM_032402.1 NM_032403.2

Gene Symbol
PCDHGB7
Entrez Gene ID
56099
Gene Name
protocadherin gamma subfamily B, 7
Gene Aliases
ME6, PCDH-GAMMA-B7
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141417647-141512981 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032092.1 NM_003735.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3 NM_002588.3 NM_032402.1 NM_032403.2

Gene Symbol
PCDHGC3
Entrez Gene ID
5098
Gene Name
protocadherin gamma subfamily C, 3
Gene Aliases
PC43, PCDH-GAMMA-C3, PCDH2
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141476002-141512977 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032092.1 NM_003735.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3 NM_002588.3 NM_032402.1 NM_032403.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7222117 c.1254G>C p.E418D Chr.5: 141477370-141477370 on Build GRCh38
3827213 c.1141G>A p.A381T Chr.5: 141477257-141477257 on Build GRCh38
6328422 c.1296C>T p.A432A Chr.5: 141477412-141477412 on Build GRCh38
224290 c.1177C>T p.P393S Chr.5: 141477293-141477293 on Build GRCh38
4712647 c.1297G>A p.G433R Chr.5: 141477413-141477413 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771608717 Chr.5: 141477221-141477221 on Build GRCh38
rs920445601 Chr.5: 141477362-141477362 on Build GRCh38
rs778684539 Chr.5: 141477205-141477205 on Build GRCh38
rs768074414 Chr.5: 141477293-141477293 on Build GRCh38
rs753131175 Chr.5: 141477294-141477294 on Build GRCh38
rs932467433 Chr.5: 141477349-141477349 on Build GRCh38
rs771293212 Chr.5: 141477415-141477415 on Build GRCh38
rs145288114 Chr.5: 141477334-141477334 on Build GRCh38
rs779570150 Chr.5: 141477226-141477226 on Build GRCh38
rs781253466 Chr.5: 141477401-141477401 on Build GRCh38
rs769467027 Chr.5: 141477255-141477255 on Build GRCh38
rs1050849561 Chr.5: 141477357-141477357 on Build GRCh38
rs201153430 Chr.5: 141477300-141477306 on Build GRCh38
rs1004061582 Chr.5: 141477406-141477406 on Build GRCh38
rs774649069 Chr.5: 141477417-141477417 on Build GRCh38
rs761731200 Chr.5: 141477252-141477252 on Build GRCh38
rs756549446 Chr.5: 141477301-141477301 on Build GRCh38
rs147674746 Chr.5: 141477348-141477348 on Build GRCh38
rs748017565 Chr.5: 141477404-141477404 on Build GRCh38
rs60063068 Chr.5: 141477262-141477262 on Build GRCh38
rs201599682 Chr.5: 141477206-141477206 on Build GRCh38
rs759477848 Chr.5: 141477289-141477289 on Build GRCh38
rs759728291 Chr.5: 141477418-141477418 on Build GRCh38
rs754438240 Chr.5: 141477365-141477365 on Build GRCh38
rs758066525 Chr.5: 141477331-141477331 on Build GRCh38
rs543767777 Chr.5: 141477263-141477263 on Build GRCh38
rs773174763 Chr.5: 141477409-141477409 on Build GRCh38
rs140933475 Chr.5: 141477405-141477405 on Build GRCh38
rs773034406 Chr.5: 141477313-141477318 on Build GRCh38
rs751714522 Chr.5: 141477284-141477284 on Build GRCh38
rs749044339 Chr.5: 141477410-141477410 on Build GRCh38
rs199952854 Chr.5: 141477297-141477297 on Build GRCh38
rs766672047 Chr.5: 141477283-141477283 on Build GRCh38
rs367944211 Chr.5: 141477244-141477244 on Build GRCh38
rs763239421 Chr.5: 141477265-141477265 on Build GRCh38
rs372168887 Chr.5: 141477322-141477322 on Build GRCh38
rs992168113 Chr.5: 141477253-141477253 on Build GRCh38
rs768648952 Chr.5: 141477230-141477230 on Build GRCh38
rs780852225 Chr.5: 141477210-141477210 on Build GRCh38
rs745497348 Chr.5: 141477217-141477217 on Build GRCh38

View Full Product Details