Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 99105744-99105986 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SYNM
Entrez Gene ID
23336
Gene Name
synemin
Gene Aliases
DMN, SYN
UniGene
Hs.207106
Gene Chromosome Location
Chr.15: 99105069-99136586 on Build GRCh38
Transcripts
NM_015286.5 NM_145728.2 XM_017022035.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6585230 c.715G>A p.E239K Chr.15: 99105917-99105917 on Build GRCh38
4654751 c.633G>C p.E211D Chr.15: 99105832-99105832 on Build GRCh38
4880526 c.605A>C p.Y202S Chr.15: 99105804-99105804 on Build GRCh38
4654753 c.641G>T p.R214L Chr.15: 99105840-99105840 on Build GRCh38
6574320 c.761T>G p.L254R Chr.15: 99105963-99105963 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs150690767 Chr.15: 99105914-99105914 on Build GRCh38
rs371419668 Chr.15: 99105808-99105808 on Build GRCh38
rs781969816 Chr.15: 99105901-99105901 on Build GRCh38
rs200549249 Chr.15: 99105903-99105903 on Build GRCh38
rs782227705 Chr.15: 99105863-99105863 on Build GRCh38
rs549266157 Chr.15: 99105860-99105860 on Build GRCh38
rs764724634 Chr.15: 99105866-99105866 on Build GRCh38
rs782484171 Chr.15: 99105892-99105892 on Build GRCh38
rs781864601 Chr.15: 99105915-99105915 on Build GRCh38
rs979388201 Chr.15: 99105935-99105935 on Build GRCh38
rs60530833 Chr.15: 99105901-99105902 on Build GRCh38
rs782167493 Chr.15: 99105912-99105912 on Build GRCh38
rs782259134 Chr.15: 99105898-99105898 on Build GRCh38
rs531355545 Chr.15: 99105891-99105891 on Build GRCh38
rs925256483 Chr.15: 99105944-99105944 on Build GRCh38
rs782546315 Chr.15: 99105923-99105923 on Build GRCh38
rs767930656 Chr.15: 99105941-99105941 on Build GRCh38
rs775197709 Chr.15: 99105889-99105889 on Build GRCh38
rs782380449 Chr.15: 99105899-99105899 on Build GRCh38
rs782049192 Chr.15: 99105878-99105878 on Build GRCh38
rs781831400 Chr.15: 99105884-99105884 on Build GRCh38
rs112146171 Chr.15: 99105770-99105770 on Build GRCh38
rs782067959 Chr.15: 99105900-99105901 on Build GRCh38
rs782342328 Chr.15: 99105926-99105926 on Build GRCh38
rs782197393 Chr.15: 99105893-99105893 on Build GRCh38
rs571341865 Chr.15: 99105939-99105939 on Build GRCh38
rs567113863 Chr.15: 99105868-99105868 on Build GRCh38
rs762712860 Chr.15: 99105928-99105928 on Build GRCh38
rs5030689 Chr.15: 99105797-99105797 on Build GRCh38
rs782626403 Chr.15: 99105925-99105925 on Build GRCh38
rs368062336 Chr.15: 99105775-99105775 on Build GRCh38
rs782453410 Chr.15: 99105921-99105921 on Build GRCh38
rs782304866 Chr.15: 99105894-99105894 on Build GRCh38
rs782536747 Chr.15: 99105763-99105763 on Build GRCh38
rs782120693 Chr.15: 99105882-99105882 on Build GRCh38
rs781819805 Chr.15: 99105920-99105920 on Build GRCh38
rs782567415 Chr.15: 99105922-99105922 on Build GRCh38
rs782188060 Chr.15: 99105955-99105955 on Build GRCh38
rs3184104 Chr.15: 99105969-99105969 on Build GRCh38
rs781931807 Chr.15: 99105869-99105869 on Build GRCh38
rs782032860 Chr.15: 99105896-99105896 on Build GRCh38
rs377708395 Chr.15: 99105800-99105800 on Build GRCh38
rs782458458 Chr.15: 99105886-99105886 on Build GRCh38
rs782088317 Chr.15: 99105907-99105907 on Build GRCh38
rs782652806 Chr.15: 99105768-99105768 on Build GRCh38
rs782538223 Chr.15: 99105890-99105890 on Build GRCh38
rs373403078 Chr.15: 99105786-99105786 on Build GRCh38
rs781983716 Chr.15: 99105821-99105821 on Build GRCh38

View Full Product Details