Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 141431976-141432217 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PCDHGA1
Entrez Gene ID
56114
Gene Name
protocadherin gamma subfamily A, 1
Gene Aliases
PCDH-GAMMA-A1
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141330685-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032092.1 NM_003735.2 NM_032094.1 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3

Gene Symbol
PCDHGA10
Entrez Gene ID
56106
Gene Name
protocadherin gamma subfamily A, 10
Gene Aliases
PCDH-GAMMA-A10
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141413176-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032092.1 NM_003735.2 NM_032094.1 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3

Gene Symbol
PCDHGA11
Entrez Gene ID
56105
Gene Name
protocadherin gamma subfamily A, 11
Gene Aliases
PCDH-GAMMA-A11
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141420970-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032092.1 NM_003735.2 NM_032094.1 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3

Gene Symbol
PCDHGA12
Entrez Gene ID
26025
Gene Name
protocadherin gamma subfamily A, 12
Gene Aliases
CDH21, FIB3, PCDH-GAMMA-A12
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141430591-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032092.1 NM_003735.2 NM_032094.1 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3

Gene Symbol
PCDHGA2
Entrez Gene ID
56113
Gene Name
protocadherin gamma subfamily A, 2
Gene Aliases
PCDH-GAMMA-A2
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141338760-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032092.1 NM_003735.2 NM_032094.1 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3

Gene Symbol
PCDHGA3
Entrez Gene ID
56112
Gene Name
protocadherin gamma subfamily A, 3
Gene Aliases
PCDH-GAMMA-A3
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141344034-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032092.1 NM_003735.2 NM_032094.1 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3

Gene Symbol
PCDHGA4
Entrez Gene ID
56111
Gene Name
protocadherin gamma subfamily A, 4
Gene Aliases
PCDH-GAMMA-A4
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141355025-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032092.1 NM_003735.2 NM_032094.1 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3

Gene Symbol
PCDHGA5
Entrez Gene ID
56110
Gene Name
protocadherin gamma subfamily A, 5
Gene Aliases
CDH-GAMMA-A5, ME3, PCDH-GAMMA-A5
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141364331-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032092.1 NM_003735.2 NM_032094.1 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3

Gene Symbol
PCDHGA6
Entrez Gene ID
56109
Gene Name
protocadherin gamma subfamily A, 6
Gene Aliases
PCDH-GAMMA-A6
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141374084-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032092.1 NM_003735.2 NM_032094.1 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3

Gene Symbol
PCDHGA7
Entrez Gene ID
56108
Gene Name
protocadherin gamma subfamily A, 7
Gene Aliases
PCDH-GAMMA-A7
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141382739-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032092.1 NM_003735.2 NM_032094.1 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3

Gene Symbol
PCDHGA8
Entrez Gene ID
9708
Gene Name
protocadherin gamma subfamily A, 8
Gene Aliases
PCDH-GAMMA-A8
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141391916-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032092.1 NM_003735.2 NM_032094.1 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3

Gene Symbol
PCDHGA9
Entrez Gene ID
56107
Gene Name
protocadherin gamma subfamily A, 9
Gene Aliases
PCDH-GAMMA-A9
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141402953-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032092.1 NM_003735.2 NM_032094.1 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3

Gene Symbol
PCDHGB1
Entrez Gene ID
56104
Gene Name
protocadherin gamma subfamily B, 1
Gene Aliases
PCDH-GAMMA-B1
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141350261-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032092.1 NM_003735.2 NM_032094.1 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3

Gene Symbol
PCDHGB2
Entrez Gene ID
56103
Gene Name
protocadherin gamma subfamily B, 2
Gene Aliases
PCDH-GAMMA-B2
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141360136-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032092.1 NM_003735.2 NM_032094.1 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3

Gene Symbol
PCDHGB3
Entrez Gene ID
56102
Gene Name
protocadherin gamma subfamily B, 3
Gene Aliases
PCDH-GAMMA-B3
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141370264-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032092.1 NM_003735.2 NM_032094.1 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3

Gene Symbol
PCDHGB4
Entrez Gene ID
8641
Gene Name
protocadherin gamma subfamily B, 4
Gene Aliases
CDH20, FIB2, PCDH-GAMMA-B4
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141387885-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032092.1 NM_003735.2 NM_032094.1 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3

Gene Symbol
PCDHGB5
Entrez Gene ID
56101
Gene Name
protocadherin gamma subfamily B, 5
Gene Aliases
PCDH-GAMMA-B5
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141398128-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032092.1 NM_003735.2 NM_032094.1 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3

Gene Symbol
PCDHGB6
Entrez Gene ID
56100
Gene Name
protocadherin gamma subfamily B, 6
Gene Aliases
PCDH-GAMMA-B6
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141408203-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032092.1 NM_003735.2 NM_032094.1 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3

Gene Symbol
PCDHGB7
Entrez Gene ID
56099
Gene Name
protocadherin gamma subfamily B, 7
Gene Aliases
ME6, PCDH-GAMMA-B7
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141417647-141512981 on Build GRCh38
Transcripts
NM_018912.2 NM_018913.2 NM_018914.2 NM_032092.1 NM_003735.2 NM_032094.1 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018921.2 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_018926.2 NM_018927.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4916613 c.1415T>A p.V472D Chr.5: 141432174-141432174 on Build GRCh38
736621 c.1302G>A p.T434T Chr.5: 141432061-141432061 on Build GRCh38
4982881 c.1413C>T p.L471L Chr.5: 141432172-141432172 on Build GRCh38
6740767 c.1269C>T p.T423T Chr.5: 141432028-141432028 on Build GRCh38
345712 c.1235_1236GG>TT p.R412>? Chr.5: 141431994-141431995 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368480052 Chr.5: 141432155-141432155 on Build GRCh38
rs764482722 Chr.5: 141432066-141432066 on Build GRCh38
rs763154183 Chr.5: 141432096-141432096 on Build GRCh38
rs202246871 Chr.5: 141432038-141432038 on Build GRCh38
rs778393699 Chr.5: 141432014-141432014 on Build GRCh38
rs147057088 Chr.5: 141432065-141432065 on Build GRCh38
rs766026174 Chr.5: 141432136-141432136 on Build GRCh38
rs1012790217 Chr.5: 141432087-141432087 on Build GRCh38
rs773484841 Chr.5: 141432092-141432092 on Build GRCh38
rs192631651 Chr.5: 141432052-141432052 on Build GRCh38
rs746940912 Chr.5: 141432021-141432021 on Build GRCh38
rs763104309 Chr.5: 141432042-141432042 on Build GRCh38
rs950514553 Chr.5: 141432139-141432139 on Build GRCh38
rs768627576 Chr.5: 141432025-141432025 on Build GRCh38
rs774386438 Chr.5: 141432102-141432102 on Build GRCh38
rs753088299 Chr.5: 141432115-141432115 on Build GRCh38
rs148798222 Chr.5: 141432106-141432106 on Build GRCh38
rs776543767 Chr.5: 141432028-141432028 on Build GRCh38
rs759576434 Chr.5: 141432109-141432109 on Build GRCh38
rs114326665 Chr.5: 141432171-141432171 on Build GRCh38
rs572724741 Chr.5: 141432069-141432069 on Build GRCh38
rs950537869 Chr.5: 141432150-141432150 on Build GRCh38
rs376415955 Chr.5: 141432082-141432082 on Build GRCh38
rs768038692 Chr.5: 141432110-141432110 on Build GRCh38
rs774512372 Chr.5: 141432184-141432184 on Build GRCh38
rs781198519 Chr.5: 141432162-141432162 on Build GRCh38
rs199724525 Chr.5: 141432104-141432104 on Build GRCh38
rs768682551 Chr.5: 141432086-141432086 on Build GRCh38
rs764363553 Chr.5: 141432120-141432120 on Build GRCh38
rs760017836 Chr.5: 141432060-141432060 on Build GRCh38
rs752124614 Chr.5: 141432059-141432059 on Build GRCh38
rs771650925 Chr.5: 141432016-141432016 on Build GRCh38
rs373867677 Chr.5: 141432049-141432049 on Build GRCh38
rs376661062 Chr.5: 141432185-141432185 on Build GRCh38
rs200790843 Chr.5: 141432196-141432196 on Build GRCh38
rs368786187 Chr.5: 141432163-141432163 on Build GRCh38
rs374200575 Chr.5: 141432105-141432105 on Build GRCh38
rs559707772 Chr.5: 141432167-141432167 on Build GRCh38
rs144065251 Chr.5: 141432085-141432085 on Build GRCh38
rs745405194 Chr.5: 141432015-141432015 on Build GRCh38
rs372826902 Chr.5: 141432017-141432017 on Build GRCh38
rs755338230 Chr.5: 141432001-141432001 on Build GRCh38
rs749977977 Chr.5: 141432078-141432078 on Build GRCh38
rs775299266 Chr.5: 141432193-141432193 on Build GRCh38
rs773254891 Chr.5: 141432041-141432041 on Build GRCh38
rs771177256 Chr.5: 141432173-141432173 on Build GRCh38
rs745506362 Chr.5: 141432140-141432152 on Build GRCh38
rs780799160 Chr.5: 141432093-141432093 on Build GRCh38
rs943491593 Chr.5: 141432180-141432180 on Build GRCh38
rs144317211 Chr.5: 141432088-141432088 on Build GRCh38
rs562175068 Chr.5: 141432172-141432172 on Build GRCh38
rs756906117 Chr.5: 141432012-141432012 on Build GRCh38
rs761106133 Chr.5: 141432117-141432117 on Build GRCh38
rs750150518 Chr.5: 141432126-141432126 on Build GRCh38
rs138689793 Chr.5: 141432006-141432006 on Build GRCh38
rs747966355 Chr.5: 141432168-141432168 on Build GRCh38
rs878896624 Chr.5: 141432045-141432045 on Build GRCh38
rs776132438 Chr.5: 141432197-141432197 on Build GRCh38
rs138510025 Chr.5: 141432079-141432079 on Build GRCh38
rs758099753 Chr.5: 141432129-141432129 on Build GRCh38
rs751145850 Chr.5: 141432148-141432148 on Build GRCh38
rs771050429 Chr.5: 141432098-141432098 on Build GRCh38
rs769631339 Chr.5: 141432178-141432179 on Build GRCh38
rs370491149 Chr.5: 141432091-141432091 on Build GRCh38
rs781525225 Chr.5: 141432002-141432002 on Build GRCh38

View Full Product Details