Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 145846807-145846546 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NUDT17
Entrez Gene ID
200035
Gene Name
nudix hydrolase 17
Gene Aliases
-
UniGene
Hs.585066
Gene Chromosome Location
Chr.1: 145844985-145848954 on Build GRCh38
Transcripts
NM_001012758.2 XM_011509260.2 XM_011509262.1 XM_017000560.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5507995 c.493G>C p.E165Q Chr.1: 145846688-145846688 on Build GRCh38
1472536 c.438G>A p.E146E Chr.1: 145846633-145846633 on Build GRCh38
895230 c.421C>T p.R141* Chr.1: 145846616-145846616 on Build GRCh38

View Full Product Details