Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 35434579-35434840 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PAMR1
Entrez Gene ID
25891
Gene Name
peptidase domain containing associated with muscle regeneration 1
Gene Aliases
DKFZP586H2123, FP938, RAMP
UniGene
Hs.55044
Gene Chromosome Location
Chr.11: 35431827-35530300 on Build GRCh38
Transcripts
NM_001001991.2 NM_001282675.1 NM_001282676.1 NM_015430.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4032573 c.1433G>A p.R478H Chr.11: 35434756-35434756 on Build GRCh38
4032571 c.1571C>T p.T524I Chr.11: 35434618-35434618 on Build GRCh38
6223179 c.1483G>T p.G495C Chr.11: 35434706-35434706 on Build GRCh38
4032572 c.1556C>T p.A519V Chr.11: 35434633-35434633 on Build GRCh38
4955163 c.1401G>A p.E467E Chr.11: 35434788-35434788 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs111324432 Chr.11: 35434719-35434719 on Build GRCh38
rs993743389 Chr.11: 35434609-35434609 on Build GRCh38
rs372153293 Chr.11: 35434704-35434704 on Build GRCh38
rs866190960 Chr.11: 35434691-35434691 on Build GRCh38
rs142129952 Chr.11: 35434601-35434601 on Build GRCh38
rs760816665 Chr.11: 35434714-35434714 on Build GRCh38
rs190221893 Chr.11: 35434646-35434646 on Build GRCh38
rs757673735 Chr.11: 35434667-35434667 on Build GRCh38
rs201251563 Chr.11: 35434810-35434810 on Build GRCh38
rs1021309725 Chr.11: 35434645-35434645 on Build GRCh38
rs766883087 Chr.11: 35434742-35434742 on Build GRCh38
rs769785970 Chr.11: 35434629-35434629 on Build GRCh38
rs200076924 Chr.11: 35434668-35434668 on Build GRCh38
rs760864929 Chr.11: 35434654-35434654 on Build GRCh38
rs977188614 Chr.11: 35434650-35434650 on Build GRCh38
rs372294319 Chr.11: 35434628-35434628 on Build GRCh38
rs1021862850 Chr.11: 35434808-35434808 on Build GRCh38
rs199522886 Chr.11: 35434648-35434648 on Build GRCh38
rs758959191 Chr.11: 35434722-35434722 on Build GRCh38
rs114824436 Chr.11: 35434757-35434757 on Build GRCh38
rs769734442 Chr.11: 35434703-35434703 on Build GRCh38
rs776344981 Chr.11: 35434627-35434627 on Build GRCh38
rs774954835 Chr.11: 35434712-35434712 on Build GRCh38
rs747014256 Chr.11: 35434790-35434790 on Build GRCh38
rs984913116 Chr.11: 35434633-35434633 on Build GRCh38
rs150412568 Chr.11: 35434800-35434800 on Build GRCh38
rs749110579 Chr.11: 35434770-35434770 on Build GRCh38
rs750965616 Chr.11: 35434747-35434747 on Build GRCh38
rs749806723 Chr.11: 35434696-35434702 on Build GRCh38
rs750922979 Chr.11: 35434720-35434720 on Build GRCh38
rs372889720 Chr.11: 35434756-35434756 on Build GRCh38
rs754545805 Chr.11: 35434749-35434749 on Build GRCh38
rs546750875 Chr.11: 35434636-35434636 on Build GRCh38
rs201440508 Chr.11: 35434687-35434687 on Build GRCh38
rs762695518 Chr.11: 35434728-35434741 on Build GRCh38
rs148382684 Chr.11: 35434707-35434707 on Build GRCh38
rs775376621 Chr.11: 35434710-35434710 on Build GRCh38
rs901473900 Chr.11: 35434792-35434792 on Build GRCh38
rs754086758 Chr.11: 35434659-35434659 on Build GRCh38
rs768832915 Chr.11: 35434799-35434799 on Build GRCh38
rs559149381 Chr.11: 35434777-35434777 on Build GRCh38
rs35216474 Chr.11: 35434605-35434605 on Build GRCh38
rs952154039 Chr.11: 35434630-35434630 on Build GRCh38
rs759640195 Chr.11: 35434642-35434642 on Build GRCh38
rs765501262 Chr.11: 35434641-35434641 on Build GRCh38
rs924381135 Chr.11: 35434652-35434652 on Build GRCh38
rs866374715 Chr.11: 35434738-35434738 on Build GRCh38
rs143377049 Chr.11: 35434700-35434700 on Build GRCh38
rs771944611 Chr.11: 35434616-35434616 on Build GRCh38
rs764391713 Chr.11: 35434658-35434658 on Build GRCh38
rs375546092 Chr.11: 35434637-35434637 on Build GRCh38
rs577038460 Chr.11: 35434718-35434718 on Build GRCh38
rs753313677 Chr.11: 35434706-35434706 on Build GRCh38
rs146399533 Chr.11: 35434679-35434679 on Build GRCh38
rs768491202 Chr.11: 35434625-35434625 on Build GRCh38
rs770927342 Chr.11: 35434776-35434776 on Build GRCh38
rs368838060 Chr.11: 35434750-35434750 on Build GRCh38
rs537281348 Chr.11: 35434686-35434686 on Build GRCh38
rs141351261 Chr.11: 35434651-35434651 on Build GRCh38
rs777770027 Chr.11: 35434764-35434764 on Build GRCh38
rs768134226 Chr.11: 35434692-35434692 on Build GRCh38

View Full Product Details