Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 1669969-1670228 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC35E2B
Entrez Gene ID
728661
Gene Name
solute carrier family 35 member E2B
Gene Aliases
SLC35E2
UniGene
Hs.655255
Gene Chromosome Location
Chr.1: 1661478-1692804 on Build GRCh38
Transcripts
NM_001110781.2 NM_001290264.1 XM_011542072.1

SNP Detail

SNP ID Chromosome Location
rs759793398 Chr.1: 1670198-1670198 on Build GRCh38
rs753946957 Chr.1: 1670085-1670086 on Build GRCh38
rs776957676 Chr.1: 1670197-1670197 on Build GRCh38
rs373497506 Chr.1: 1670098-1670098 on Build GRCh38
rs377734355 Chr.1: 1670118-1670118 on Build GRCh38
rs770709500 Chr.1: 1670210-1670210 on Build GRCh38
rs1020369972 Chr.1: 1670057-1670057 on Build GRCh38
rs572880205 Chr.1: 1670097-1670097 on Build GRCh38
rs754649609 Chr.1: 1670051-1670051 on Build GRCh38
rs758316480 Chr.1: 1670109-1670109 on Build GRCh38
rs779583866 Chr.1: 1670091-1670094 on Build GRCh38
rs575817010 Chr.1: 1670016-1670016 on Build GRCh38
rs746999072 Chr.1: 1670193-1670193 on Build GRCh38
rs139260872 Chr.1: 1670192-1670192 on Build GRCh38
rs773469875 Chr.1: 1670013-1670013 on Build GRCh38
rs978974337 Chr.1: 1670127-1670127 on Build GRCh38
rs555458885 Chr.1: 1670207-1670207 on Build GRCh38
rs747544634 Chr.1: 1670190-1670190 on Build GRCh38
rs771550055 Chr.1: 1670108-1670108 on Build GRCh38
rs752936487 Chr.1: 1670036-1670036 on Build GRCh38
rs372106488 Chr.1: 1670033-1670033 on Build GRCh38
rs370941743 Chr.1: 1670086-1670086 on Build GRCh38

View Full Product Details