Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 40190563-40190828 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RLF
Entrez Gene ID
6018
Gene Name
rearranged L-myc fusion
Gene Aliases
ZN-15L, ZNF292L
UniGene
Hs.205627
Gene Chromosome Location
Chr.1: 40161369-40240921 on Build GRCh38
Transcripts
NM_012421.3 XM_017002016.1 XM_017002017.1 XM_017002018.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6881927 c.442G>A p.E148K Chr.1: 40190821-40190821 on Build GRCh38
6460462 c.414T>C p.L138L Chr.1: 40190793-40190793 on Build GRCh38
187830 c.403G>A p.E135K Chr.1: 40190782-40190782 on Build GRCh38
909388 c.441T>C p.C147C Chr.1: 40190820-40190820 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1003806854 Chr.1: 40190739-40190739 on Build GRCh38
rs543466563 Chr.1: 40190752-40190752 on Build GRCh38
rs970634322 Chr.1: 40190735-40190735 on Build GRCh38
rs374264863 Chr.1: 40190730-40190730 on Build GRCh38
rs775967063 Chr.1: 40190732-40190732 on Build GRCh38
rs377669104 Chr.1: 40190781-40190781 on Build GRCh38
rs1033547199 Chr.1: 40190720-40190720 on Build GRCh38
rs771350379 Chr.1: 40190627-40190627 on Build GRCh38
rs574809556 Chr.1: 40190714-40190714 on Build GRCh38
rs898814809 Chr.1: 40190691-40190691 on Build GRCh38
rs777137641 Chr.1: 40190747-40190747 on Build GRCh38
rs113860168 Chr.1: 40190624-40190624 on Build GRCh38
rs767198286 Chr.1: 40190629-40190629 on Build GRCh38
rs769106056 Chr.1: 40190743-40190743 on Build GRCh38
rs541173702 Chr.1: 40190607-40190607 on Build GRCh38
rs765368714 Chr.1: 40190765-40190765 on Build GRCh38
rs1012156062 Chr.1: 40190709-40190709 on Build GRCh38
rs563655330 Chr.1: 40190766-40190766 on Build GRCh38
rs766055366 Chr.1: 40190798-40190798 on Build GRCh38
rs952061149 Chr.1: 40190659-40190659 on Build GRCh38
rs760979646 Chr.1: 40190734-40190734 on Build GRCh38
rs140609951 Chr.1: 40190796-40190796 on Build GRCh38
rs754839476 Chr.1: 40190804-40190804 on Build GRCh38

View Full Product Details