Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 2771401-2771670 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SRRM2
Entrez Gene ID
23524
Gene Name
serine/arginine repetitive matrix 2
Gene Aliases
300-KD, CWF21, Cwc21, HSPC075, SRL300, SRm300
UniGene
Hs.433343
Gene Chromosome Location
Chr.16: 2752329-2771412 on Build GRCh38
Transcripts
NM_016333.3 XM_005255226.1 NM_007108.3 NM_207013.2

Gene Symbol
TCEB2
Entrez Gene ID
6923
Gene Name
transcription elongation factor B subunit 2
Gene Aliases
ELOB, SIII
UniGene
Hs.172772
Gene Chromosome Location
Chr.16: 2771414-2777296 on Build GRCh38
Transcripts
NM_016333.3 XM_005255226.1 NM_007108.3 NM_207013.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7113990 c.479G>T p.G160V Chr.16: 2771479-2771479 on Build GRCh38
3109588 c.442C>T p.P148S Chr.16: 2771516-2771516 on Build GRCh38
5621978 c.424G>A p.A142T Chr.16: 2771534-2771534 on Build GRCh38
3109589 c.435G>A p.T145T Chr.16: 2771523-2771523 on Build GRCh38
3109591 c.362T>C p.V121A Chr.16: 2771596-2771596 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760693721 Chr.16: 2771451-2771451 on Build GRCh38
rs747934062 Chr.16: 2771608-2771609 on Build GRCh38
rs762011880 Chr.16: 2771505-2771505 on Build GRCh38
rs761234252 Chr.16: 2771432-2771435 on Build GRCh38
rs200603469 Chr.16: 2771443-2771443 on Build GRCh38
rs768424617 Chr.16: 2771605-2771605 on Build GRCh38
rs564715488 Chr.16: 2771543-2771543 on Build GRCh38
rs747670781 Chr.16: 2771429-2771429 on Build GRCh38
rs773553329 Chr.16: 2771615-2771615 on Build GRCh38
rs770679025 Chr.16: 2771444-2771444 on Build GRCh38
rs779016860 Chr.16: 2771615-2771615 on Build GRCh38
rs764833034 Chr.16: 2771461-2771464 on Build GRCh38
rs756846212 Chr.16: 2771531-2771531 on Build GRCh38
rs370330180 Chr.16: 2771557-2771557 on Build GRCh38
rs79638577 Chr.16: 2771597-2771597 on Build GRCh38
rs1025623238 Chr.16: 2771510-2771510 on Build GRCh38
rs769825458 Chr.16: 2771639-2771639 on Build GRCh38
rs759557375 Chr.16: 2771427-2771430 on Build GRCh38
rs372745911 Chr.16: 2771435-2771435 on Build GRCh38
rs541931588 Chr.16: 2771428-2771429 on Build GRCh38
rs753289154 Chr.16: 2771630-2771630 on Build GRCh38
rs747651035 Chr.16: 2771468-2771468 on Build GRCh38
rs193122310 Chr.16: 2771485-2771485 on Build GRCh38
rs373129178 Chr.16: 2771458-2771458 on Build GRCh38
rs1047224490 Chr.16: 2771558-2771558 on Build GRCh38
rs773606482 Chr.16: 2771581-2771581 on Build GRCh38
rs763384913 Chr.16: 2771582-2771582 on Build GRCh38
rs750965170 Chr.16: 2771463-2771463 on Build GRCh38
rs374236114 Chr.16: 2771607-2771607 on Build GRCh38
rs779783218 Chr.16: 2771599-2771599 on Build GRCh38
rs769437553 Chr.16: 2771431-2771431 on Build GRCh38
rs778507762 Chr.16: 2771593-2771593 on Build GRCh38
rs756899786 Chr.16: 2771452-2771453 on Build GRCh38
rs553919891 Chr.16: 2771562-2771563 on Build GRCh38
rs762777288 Chr.16: 2771438-2771438 on Build GRCh38
rs748204375 Chr.16: 2771610-2771610 on Build GRCh38
rs759967585 Chr.16: 2771627-2771627 on Build GRCh38
rs767447633 Chr.16: 2771447-2771447 on Build GRCh38
rs200168704 Chr.16: 2771480-2771480 on Build GRCh38
rs780778101 Chr.16: 2771427-2771427 on Build GRCh38
rs368610933 Chr.16: 2771621-2771621 on Build GRCh38
rs775387787 Chr.16: 2771563-2771563 on Build GRCh38
rs781113021 Chr.16: 2771586-2771587 on Build GRCh38
rs1038674922 Chr.16: 2771540-2771540 on Build GRCh38
rs967206788 Chr.16: 2771491-2771491 on Build GRCh38
rs771282800 Chr.16: 2771620-2771620 on Build GRCh38
rs765511882 Chr.16: 2771580-2771580 on Build GRCh38
rs754199376 Chr.16: 2771444-2771444 on Build GRCh38
rs772133791 Chr.16: 2771626-2771631 on Build GRCh38
rs745571275 Chr.16: 2771594-2771594 on Build GRCh38
rs761291670 Chr.16: 2771633-2771633 on Build GRCh38
rs898700503 Chr.16: 2771441-2771441 on Build GRCh38
rs780074064 Chr.16: 2771541-2771542 on Build GRCh38
rs752101364 Chr.16: 2771584-2771584 on Build GRCh38
rs780255123 Chr.16: 2771626-2771628 on Build GRCh38
rs8017 Chr.16: 2771572-2771572 on Build GRCh38
rs372065842 Chr.16: 2771636-2771636 on Build GRCh38
rs746829193 Chr.16: 2771601-2771601 on Build GRCh38
rs963998540 Chr.16: 2771484-2771484 on Build GRCh38
rs920189162 Chr.16: 2771646-2771646 on Build GRCh38
rs1038555293 Chr.16: 2771426-2771426 on Build GRCh38
rs748949711 Chr.16: 2771471-2771471 on Build GRCh38
rs775602329 Chr.16: 2771626-2771629 on Build GRCh38
rs751268116 Chr.16: 2771648-2771648 on Build GRCh38
rs764787422 Chr.16: 2771638-2771638 on Build GRCh38
rs573242037 Chr.16: 2771495-2771495 on Build GRCh38
rs778558745 Chr.16: 2771541-2771541 on Build GRCh38
rs770755187 Chr.16: 2771472-2771472 on Build GRCh38
rs145844597 Chr.16: 2771585-2771585 on Build GRCh38
rs770041121 Chr.16: 2771612-2771612 on Build GRCh38
rs758913464 Chr.16: 2771464-2771464 on Build GRCh38
rs201628798 Chr.16: 2771523-2771523 on Build GRCh38
rs923580836 Chr.16: 2771455-2771455 on Build GRCh38
rs752715553 Chr.16: 2771448-2771448 on Build GRCh38
rs1031463071 Chr.16: 2771617-2771617 on Build GRCh38
rs992685694 Chr.16: 2771619-2771619 on Build GRCh38
rs367582616 Chr.16: 2771642-2771642 on Build GRCh38
rs960966924 Chr.16: 2771516-2771516 on Build GRCh38
rs554461759 Chr.16: 2771519-2771519 on Build GRCh38
rs779482898 Chr.16: 2771462-2771462 on Build GRCh38
rs138405434 Chr.16: 2771588-2771588 on Build GRCh38
rs367976124 Chr.16: 2771578-2771578 on Build GRCh38
rs763332020 Chr.16: 2771618-2771618 on Build GRCh38
rs1005626834 Chr.16: 2771533-2771533 on Build GRCh38
rs563206993 Chr.16: 2771568-2771568 on Build GRCh38
rs149027706 Chr.16: 2771508-2771508 on Build GRCh38
rs200070374 Chr.16: 2771583-2771583 on Build GRCh38
rs1007909655 Chr.16: 2771475-2771475 on Build GRCh38
rs764338557 Chr.16: 2771452-2771452 on Build GRCh38
rs767872586 Chr.16: 2771628-2771628 on Build GRCh38
rs768781603 Chr.16: 2771574-2771574 on Build GRCh38
rs774259338 Chr.16: 2771445-2771445 on Build GRCh38
rs766959241 Chr.16: 2771512-2771512 on Build GRCh38
rs774448231 Chr.16: 2771426-2771427 on Build GRCh38
rs942436127 Chr.16: 2771625-2771625 on Build GRCh38
rs756792853 Chr.16: 2771589-2771589 on Build GRCh38
rs185064686 Chr.16: 2771486-2771486 on Build GRCh38
rs4036 Chr.16: 2771524-2771524 on Build GRCh38
rs376053410 Chr.16: 2771502-2771502 on Build GRCh38
rs974310000 Chr.16: 2771643-2771643 on Build GRCh38
rs764132351 Chr.16: 2771504-2771504 on Build GRCh38
rs752054059 Chr.16: 2771515-2771515 on Build GRCh38
rs772677442 Chr.16: 2771467-2771467 on Build GRCh38
rs771135263 Chr.16: 2771614-2771615 on Build GRCh38
rs1040849647 Chr.16: 2771482-2771482 on Build GRCh38
rs776511641 Chr.16: 2771577-2771577 on Build GRCh38
rs369392329 Chr.16: 2771454-2771454 on Build GRCh38
rs749903482 Chr.16: 2771470-2771474 on Build GRCh38
rs764425637 Chr.16: 2771436-2771437 on Build GRCh38
rs746940870 Chr.16: 2771554-2771558 on Build GRCh38
rs757862999 Chr.16: 2771505-2771506 on Build GRCh38
rs930158417 Chr.16: 2771545-2771545 on Build GRCh38
rs768272140 Chr.16: 2771424-2771424 on Build GRCh38
rs79886307 Chr.16: 2771507-2771507 on Build GRCh38
rs372215429 Chr.16: 2771640-2771640 on Build GRCh38

View Full Product Details