Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 130910171-130910424 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SNX19
Entrez Gene ID
399979
Gene Name
sorting nexin 19
Gene Aliases
CHET8
UniGene
Hs.444024
Gene Chromosome Location
Chr.11: 130866254-130916488 on Build GRCh38
Transcripts
NM_001301089.1 NM_014758.2 XM_005271546.4 XM_011542819.2 XM_011542820.2 XM_011542821.2 XM_011542822.2 XM_011542824.2 XM_011542825.2 XM_011542826.2 XM_011542827.2 XM_017017725.1 XM_017017726.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7341732 c.1895T>C p.L632P Chr.11: 130910289-130910289 on Build GRCh38
6879195 c.1850C>T p.P617L Chr.11: 130910334-130910334 on Build GRCh38
3808810 c.1884C>T p.A628A Chr.11: 130910300-130910300 on Build GRCh38
6859135 c.1814-5_1814-4insT p.? Chr.11: 130910374-130910375 on Build GRCh38
6879196 c.1849C>T p.P617S Chr.11: 130910335-130910335 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745382645 Chr.11: 130910362-130910362 on Build GRCh38
rs768747763 Chr.11: 130910371-130910371 on Build GRCh38
rs1047814480 Chr.11: 130910260-130910260 on Build GRCh38
rs751599115 Chr.11: 130910306-130910306 on Build GRCh38
rs775546862 Chr.11: 130910367-130910367 on Build GRCh38
rs752878460 Chr.11: 130910375-130910380 on Build GRCh38
rs575157043 Chr.11: 130910211-130910211 on Build GRCh38
rs681988 Chr.11: 130910336-130910336 on Build GRCh38
rs529587776 Chr.11: 130910276-130910276 on Build GRCh38
rs1237478 Chr.11: 130910375-130910375 on Build GRCh38
rs371167065 Chr.11: 130910343-130910343 on Build GRCh38
rs554200347 Chr.11: 130910374-130910375 on Build GRCh38
rs772935787 Chr.11: 130910352-130910352 on Build GRCh38
rs752919945 Chr.11: 130910374-130910378 on Build GRCh38
rs750642764 Chr.11: 130910298-130910298 on Build GRCh38
rs659160 Chr.11: 130910345-130910345 on Build GRCh38
rs775640664 Chr.11: 130910247-130910247 on Build GRCh38
rs760536406 Chr.11: 130910270-130910270 on Build GRCh38
rs761220739 Chr.11: 130910308-130910309 on Build GRCh38
rs773869028 Chr.11: 130910240-130910240 on Build GRCh38
rs751379124 Chr.11: 130910374-130910379 on Build GRCh38
rs577532999 Chr.11: 130910198-130910198 on Build GRCh38
rs796584708 Chr.11: 130910374-130910375 on Build GRCh38
rs758984901 Chr.11: 130910241-130910241 on Build GRCh38
rs766534162 Chr.11: 130910304-130910304 on Build GRCh38
rs769503392 Chr.11: 130910311-130910311 on Build GRCh38
rs778147893 Chr.11: 130910335-130910335 on Build GRCh38
rs681982 Chr.11: 130910330-130910330 on Build GRCh38
rs681536 Chr.11: 130910251-130910251 on Build GRCh38
rs770210348 Chr.11: 130910221-130910221 on Build GRCh38
rs113123833 Chr.11: 130910375-130910375 on Build GRCh38
rs543842226 Chr.11: 130910227-130910227 on Build GRCh38
rs771543583 Chr.11: 130910363-130910363 on Build GRCh38
rs796595425 Chr.11: 130910375-130910376 on Build GRCh38
rs767425731 Chr.11: 130910245-130910245 on Build GRCh38
rs149459173 Chr.11: 130910354-130910354 on Build GRCh38
rs756444075 Chr.11: 130910333-130910333 on Build GRCh38
rs556602428 Chr.11: 130910375-130910376 on Build GRCh38
rs372369099 Chr.11: 130910374-130910374 on Build GRCh38
rs755015810 Chr.11: 130910308-130910308 on Build GRCh38
rs764031207 Chr.11: 130910284-130910284 on Build GRCh38
rs755229207 Chr.11: 130910217-130910217 on Build GRCh38
rs201127404 Chr.11: 130910237-130910237 on Build GRCh38
rs781720446 Chr.11: 130910318-130910318 on Build GRCh38
rs760734580 Chr.11: 130910368-130910368 on Build GRCh38
rs202209451 Chr.11: 130910375-130910379 on Build GRCh38
rs777973614 Chr.11: 130910375-130910378 on Build GRCh38
rs1014897221 Chr.11: 130910288-130910288 on Build GRCh38
rs758655328 Chr.11: 130910299-130910299 on Build GRCh38

View Full Product Details