Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 62994311-62994572 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC22A8
Entrez Gene ID
9376
Gene Name
solute carrier family 22 member 8
Gene Aliases
OAT3
UniGene
Hs.266223
Gene Chromosome Location
Chr.11: 62992824-63015845 on Build GRCh38
Transcripts
NM_001184732.1 NM_001184733.1 NM_001184736.1 NM_004254.3 XM_011545364.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4797892 c.1202C>T p.T401I Chr.11: 62994556-62994556 on Build GRCh38
6552209 c.1207G>A p.V403M Chr.11: 62994551-62994551 on Build GRCh38
6306674 c.1216+9C>A p.? Chr.11: 62994533-62994533 on Build GRCh38
4983902 c.1200C>T p.L400L Chr.11: 62994558-62994558 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs546697598 Chr.11: 62994464-62994464 on Build GRCh38
rs773430800 Chr.11: 62994541-62994541 on Build GRCh38
rs961404766 Chr.11: 62994425-62994425 on Build GRCh38
rs575926672 Chr.11: 62994390-62994390 on Build GRCh38
rs1030254454 Chr.11: 62994433-62994433 on Build GRCh38
rs79895775 Chr.11: 62994410-62994410 on Build GRCh38
rs540265235 Chr.11: 62994487-62994487 on Build GRCh38
rs954504591 Chr.11: 62994454-62994454 on Build GRCh38
rs1002396656 Chr.11: 62994374-62994374 on Build GRCh38
rs1036202109 Chr.11: 62994353-62994353 on Build GRCh38
rs774206147 Chr.11: 62994491-62994491 on Build GRCh38
rs777778792 Chr.11: 62994389-62994389 on Build GRCh38
rs766729668 Chr.11: 62994550-62994550 on Build GRCh38
rs957790590 Chr.11: 62994488-62994488 on Build GRCh38
rs772720461 Chr.11: 62994388-62994388 on Build GRCh38
rs560117156 Chr.11: 62994543-62994543 on Build GRCh38
rs77253319 Chr.11: 62994399-62994399 on Build GRCh38
rs555957457 Chr.11: 62994364-62994364 on Build GRCh38
rs758932398 Chr.11: 62994352-62994352 on Build GRCh38
rs993379924 Chr.11: 62994431-62994431 on Build GRCh38
rs770537147 Chr.11: 62994409-62994409 on Build GRCh38
rs375750340 Chr.11: 62994524-62994524 on Build GRCh38
rs761830630 Chr.11: 62994522-62994522 on Build GRCh38
rs989672548 Chr.11: 62994490-62994490 on Build GRCh38

View Full Product Details