Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 49489470-49489715 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPATS2
Entrez Gene ID
65244
Gene Name
spermatogenesis associated serine rich 2
Gene Aliases
Nbla00526, P59SCR, SCR59, SPATA10
UniGene
Hs.654826
Gene Chromosome Location
Chr.12: 49366905-49527441 on Build GRCh38
Transcripts
NM_001293285.1 NM_001293286.1 NM_023071.3 NR_120663.1 XM_011538690.2 XM_011538691.2 XM_017019840.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7295869 c.113C>T p.A38V Chr.12: 49489472-49489472 on Build GRCh38
1705563 c.161T>C p.V54A Chr.12: 49489520-49489520 on Build GRCh38
3792652 c.211G>A p.E71K Chr.12: 49489570-49489570 on Build GRCh38
190118 c.119G>A p.R40H Chr.12: 49489478-49489478 on Build GRCh38
940141 c.148G>T p.E50* Chr.12: 49489507-49489507 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs2272478 Chr.12: 49489588-49489588 on Build GRCh38
rs779099983 Chr.12: 49489562-49489562 on Build GRCh38
rs941751748 Chr.12: 49489660-49489660 on Build GRCh38
rs767550899 Chr.12: 49489526-49489526 on Build GRCh38
rs750273703 Chr.12: 49489532-49489532 on Build GRCh38
rs1037442460 Chr.12: 49489663-49489663 on Build GRCh38
rs778728933 Chr.12: 49489616-49489616 on Build GRCh38
rs915349706 Chr.12: 49489549-49489549 on Build GRCh38
rs111615076 Chr.12: 49489518-49489518 on Build GRCh38
rs781728684 Chr.12: 49489618-49489618 on Build GRCh38
rs185361091 Chr.12: 49489563-49489563 on Build GRCh38
rs748051226 Chr.12: 49489634-49489634 on Build GRCh38
rs752991508 Chr.12: 49489579-49489579 on Build GRCh38
rs934574050 Chr.12: 49489654-49489654 on Build GRCh38
rs746306528 Chr.12: 49489623-49489623 on Build GRCh38
rs755116677 Chr.12: 49489557-49489557 on Build GRCh38
rs983082033 Chr.12: 49489614-49489614 on Build GRCh38
rs747475241 Chr.12: 49489605-49489605 on Build GRCh38
rs924547825 Chr.12: 49489648-49489648 on Build GRCh38
rs747208087 Chr.12: 49489555-49489555 on Build GRCh38
rs777828466 Chr.12: 49489589-49489589 on Build GRCh38
rs771184115 Chr.12: 49489609-49489609 on Build GRCh38
rs914364348 Chr.12: 49489505-49489505 on Build GRCh38
rs907541262 Chr.12: 49489657-49489657 on Build GRCh38

View Full Product Details