Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 191033311-191033542 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
STAT4
Entrez Gene ID
6775
Gene Name
signal transducer and activator of transcription 4
Gene Aliases
SLEB11
UniGene
Hs.735572
Gene Chromosome Location
Chr.2: 191029576-191172684 on Build GRCh38
Transcripts
NM_001243835.1 NM_003151.3 XM_006712719.3 XM_011511705.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1288240 c.1849A>T p.S617C Chr.2: 191033493-191033493 on Build GRCh38
4898440 c.1825T>C p.F609L Chr.2: 191033517-191033517 on Build GRCh38
6587844 c.1814G>T p.G605V Chr.2: 191033528-191033528 on Build GRCh38
95285 c.1844C>G p.S615C Chr.2: 191033498-191033498 on Build GRCh38
4920711 c.1837G>A p.D613N Chr.2: 191033505-191033505 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs980147049 Chr.2: 191033395-191033395 on Build GRCh38
rs748265285 Chr.2: 191033477-191033477 on Build GRCh38
rs753717175 Chr.2: 191033445-191033445 on Build GRCh38
rs866114669 Chr.2: 191033359-191033359 on Build GRCh38
rs201561308 Chr.2: 191033484-191033484 on Build GRCh38
rs1050592569 Chr.2: 191033464-191033464 on Build GRCh38
rs927495263 Chr.2: 191033428-191033428 on Build GRCh38
rs779081265 Chr.2: 191033465-191033465 on Build GRCh38
rs933016975 Chr.2: 191033460-191033460 on Build GRCh38
rs777832373 Chr.2: 191033492-191033492 on Build GRCh38
rs554498147 Chr.2: 191033444-191033444 on Build GRCh38
rs536471585 Chr.2: 191033407-191033407 on Build GRCh38
rs754973281 Chr.2: 191033461-191033461 on Build GRCh38
rs867832285 Chr.2: 191033426-191033426 on Build GRCh38
rs968790243 Chr.2: 191033365-191033365 on Build GRCh38
rs901590089 Chr.2: 191033353-191033353 on Build GRCh38

View Full Product Details