Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 108008454-108008719 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC5A7
Entrez Gene ID
60482
Gene Name
solute carrier family 5 member 7
Gene Aliases
CHT, CHT1, HMN7A
UniGene
Hs.287758
Gene Chromosome Location
Chr.2: 107986514-108014497 on Build GRCh38
Transcripts
NM_001305005.2 NM_001305006.2 NM_001305007.2 NM_021815.4 XM_011511580.2 XM_017004628.1 XM_017004629.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4563037 c.954G>A p.M318I Chr.2: 108008523-108008523 on Build GRCh38
4512595 c.908C>T p.T303I Chr.2: 108008477-108008477 on Build GRCh38
3723603 c.1012G>C p.G338R Chr.2: 108008581-108008581 on Build GRCh38
6783133 c.1059G>T p.L353F Chr.2: 108008628-108008628 on Build GRCh38
7286831 c.981C>T p.L327L Chr.2: 108008550-108008550 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762453964 Chr.2: 108008641-108008641 on Build GRCh38
rs770501137 Chr.2: 108008559-108008559 on Build GRCh38
rs147656110 Chr.2: 108008651-108008651 on Build GRCh38
rs1011037891 Chr.2: 108008550-108008550 on Build GRCh38
rs746802512 Chr.2: 108008557-108008557 on Build GRCh38
rs775122957 Chr.2: 108008623-108008623 on Build GRCh38
rs745483747 Chr.2: 108008605-108008606 on Build GRCh38
rs778842636 Chr.2: 108008508-108008508 on Build GRCh38
rs766515861 Chr.2: 108008688-108008688 on Build GRCh38
rs761044523 Chr.2: 108008655-108008655 on Build GRCh38
rs192964655 Chr.2: 108008691-108008691 on Build GRCh38
rs746914288 Chr.2: 108008658-108008658 on Build GRCh38
rs769383404 Chr.2: 108008602-108008602 on Build GRCh38
rs758084765 Chr.2: 108008608-108008611 on Build GRCh38
rs1044463744 Chr.2: 108008543-108008543 on Build GRCh38
rs376624433 Chr.2: 108008650-108008650 on Build GRCh38
rs905933912 Chr.2: 108008592-108008592 on Build GRCh38
rs745405514 Chr.2: 108008595-108008595 on Build GRCh38
rs542281272 Chr.2: 108008547-108008547 on Build GRCh38
rs776353506 Chr.2: 108008562-108008562 on Build GRCh38
rs140481686 Chr.2: 108008596-108008596 on Build GRCh38
rs758323741 Chr.2: 108008532-108008532 on Build GRCh38
rs868566231 Chr.2: 108008639-108008639 on Build GRCh38
rs748131882 Chr.2: 108008514-108008514 on Build GRCh38

View Full Product Details