Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 21140414-21140675 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SAP18
Entrez Gene ID
10284
Gene Name
Sin3A associated protein 18
Gene Aliases
2HOR0202, SAP18P
UniGene
Hs.524899
Gene Chromosome Location
Chr.13: 21140514-21149085 on Build GRCh38
Transcripts
NM_005870.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5980459 c.14C>T p.S5L Chr.13: 21140623-21140623 on Build GRCh38
3981956 c.18C>T p.R6R Chr.13: 21140627-21140627 on Build GRCh38
7254615 c.64C>A p.R22S Chr.13: 21140673-21140673 on Build GRCh38
7283823 c.37A>T p.K13* Chr.13: 21140646-21140646 on Build GRCh38
4957815 c.37A>G p.K13E Chr.13: 21140646-21140646 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760246435 Chr.13: 21140538-21140538 on Build GRCh38
rs998156028 Chr.13: 21140598-21140598 on Build GRCh38
rs139949359 Chr.13: 21140610-21140610 on Build GRCh38
rs778665161 Chr.13: 21140558-21140558 on Build GRCh38
rs370333573 Chr.13: 21140567-21140567 on Build GRCh38
rs768613316 Chr.13: 21140527-21140527 on Build GRCh38
rs975446690 Chr.13: 21140445-21140445 on Build GRCh38
rs772931538 Chr.13: 21140541-21140541 on Build GRCh38
rs775068346 Chr.13: 21140564-21140564 on Build GRCh38
rs1042805999 Chr.13: 21140484-21140484 on Build GRCh38
rs373936573 Chr.13: 21140591-21140591 on Build GRCh38
rs552329707 Chr.13: 21140583-21140583 on Build GRCh38
rs759431578 Chr.13: 21140529-21140531 on Build GRCh38
rs375651525 Chr.13: 21140582-21140582 on Build GRCh38
rs774972864 Chr.13: 21140592-21140592 on Build GRCh38
rs373660324 Chr.13: 21140456-21140456 on Build GRCh38
rs772015507 Chr.13: 21140531-21140535 on Build GRCh38
rs774921169 Chr.13: 21140537-21140538 on Build GRCh38
rs777722330 Chr.13: 21140543-21140543 on Build GRCh38
rs908359977 Chr.13: 21140461-21140461 on Build GRCh38
rs749806992 Chr.13: 21140517-21140517 on Build GRCh38
rs112687601 Chr.13: 21140576-21140576 on Build GRCh38
rs750433806 Chr.13: 21140604-21140604 on Build GRCh38
rs199958048 Chr.13: 21140542-21140542 on Build GRCh38
rs777404407 Chr.13: 21140625-21140625 on Build GRCh38
rs758460442 Chr.13: 21140507-21140507 on Build GRCh38
rs934333275 Chr.13: 21140492-21140492 on Build GRCh38
rs747038252 Chr.13: 21140639-21140639 on Build GRCh38
rs548145674 Chr.13: 21140575-21140575 on Build GRCh38
rs752464689 Chr.13: 21140620-21140620 on Build GRCh38
rs200936744 Chr.13: 21140539-21140539 on Build GRCh38
rs752852568 Chr.13: 21140506-21140506 on Build GRCh38
rs781351427 Chr.13: 21140549-21140549 on Build GRCh38
rs1003849675 Chr.13: 21140654-21140654 on Build GRCh38
rs772607672 Chr.13: 21140568-21140568 on Build GRCh38
rs762835068 Chr.13: 21140577-21140577 on Build GRCh38
rs761716479 Chr.13: 21140532-21140532 on Build GRCh38
rs563914498 Chr.13: 21140444-21140444 on Build GRCh38
rs941311914 Chr.13: 21140464-21140464 on Build GRCh38
rs758288241 Chr.13: 21140579-21140579 on Build GRCh38
rs763338973 Chr.13: 21140589-21140589 on Build GRCh38
rs764614475 Chr.13: 21140573-21140573 on Build GRCh38
rs374776695 Chr.13: 21140569-21140569 on Build GRCh38
rs540223029 Chr.13: 21140519-21140519 on Build GRCh38
rs769356286 Chr.13: 21140438-21140439 on Build GRCh38
rs774312518 Chr.13: 21140530-21140530 on Build GRCh38
rs751601385 Chr.13: 21140544-21140544 on Build GRCh38
rs908412190 Chr.13: 21140566-21140566 on Build GRCh38
rs200122454 Chr.13: 21140515-21140515 on Build GRCh38
rs758249818 Chr.13: 21140624-21140624 on Build GRCh38
rs754273316 Chr.13: 21140540-21140540 on Build GRCh38
rs530592109 Chr.13: 21140578-21140578 on Build GRCh38
rs1017859131 Chr.13: 21140534-21140534 on Build GRCh38
rs564217124 Chr.13: 21140500-21140500 on Build GRCh38
rs978956587 Chr.13: 21140554-21140554 on Build GRCh38
rs922366222 Chr.13: 21140446-21140446 on Build GRCh38
rs769582269 Chr.13: 21140518-21140518 on Build GRCh38
rs777899487 Chr.13: 21140511-21140511 on Build GRCh38
rs964979702 Chr.13: 21140627-21140627 on Build GRCh38
rs1029317634 Chr.13: 21140548-21140548 on Build GRCh38
rs893298516 Chr.13: 21140520-21140520 on Build GRCh38
rs1038281216 Chr.13: 21140497-21140497 on Build GRCh38
rs761615849 Chr.13: 21140502-21140502 on Build GRCh38
rs746060471 Chr.13: 21140553-21140553 on Build GRCh38
rs775933057 Chr.13: 21140572-21140572 on Build GRCh38
rs201454186 Chr.13: 21140531-21140531 on Build GRCh38
rs763649081 Chr.13: 21140599-21140599 on Build GRCh38
rs1053219185 Chr.13: 21140494-21140494 on Build GRCh38
rs904288486 Chr.13: 21140484-21140485 on Build GRCh38
rs745523301 Chr.13: 21140584-21140584 on Build GRCh38
rs757445435 Chr.13: 21140545-21140545 on Build GRCh38
rs575712951 Chr.13: 21140452-21140452 on Build GRCh38
rs771292591 Chr.13: 21140537-21140537 on Build GRCh38
rs769516459 Chr.13: 21140586-21140586 on Build GRCh38
rs751622316 Chr.13: 21140574-21140574 on Build GRCh38
rs570548873 Chr.13: 21140599-21140599 on Build GRCh38
rs955130207 Chr.13: 21140551-21140551 on Build GRCh38
rs368287644 Chr.13: 21140570-21140570 on Build GRCh38
rs982643722 Chr.13: 21140460-21140460 on Build GRCh38
rs190705962 Chr.13: 21140442-21140442 on Build GRCh38
rs1029371490 Chr.13: 21140602-21140602 on Build GRCh38
rs765768187 Chr.13: 21140571-21140571 on Build GRCh38
rs767650009 Chr.13: 21140601-21140601 on Build GRCh38

View Full Product Details