Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 112154874-112155147 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC9C1
Entrez Gene ID
285335
Gene Name
solute carrier family 9 member C1
Gene Aliases
NHE, NHE-10, SLC9A10, sperm-NHE
UniGene
Hs.680112
Gene Chromosome Location
Chr.3: 112140898-112294258 on Build GRCh38
Transcripts
NM_001320531.1 NM_183061.2 NR_135297.1 XM_006713603.2 XM_011512718.1 XM_011512719.1 XM_011512720.1 XM_011512721.1 XM_011512725.2 XM_017006246.1 XM_017006247.1 XM_017006248.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1036345 c.3400G>T p.E1134* Chr.3: 112155014-112155014 on Build GRCh38
7306343 c.3365-7_3365-6insTT p.? Chr.3: 112155055-112155056 on Build GRCh38
5041218 c.3380T>C p.L1127S Chr.3: 112155034-112155034 on Build GRCh38
1632714 c.3382G>A p.E1128K Chr.3: 112155032-112155032 on Build GRCh38
3585358 c.3372T>C p.V1124V Chr.3: 112155042-112155042 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs11442415 Chr.3: 112155063-112155064 on Build GRCh38
rs564252554 Chr.3: 112154946-112154946 on Build GRCh38
rs776747275 Chr.3: 112155060-112155061 on Build GRCh38
rs769473421 Chr.3: 112155053-112155054 on Build GRCh38
rs765348453 Chr.3: 112155066-112155067 on Build GRCh38
rs745503181 Chr.3: 112155026-112155026 on Build GRCh38
rs775401526 Chr.3: 112155065-112155065 on Build GRCh38
rs772880247 Chr.3: 112154993-112154993 on Build GRCh38
rs777369299 Chr.3: 112155011-112155011 on Build GRCh38
rs74425027 Chr.3: 112154955-112154955 on Build GRCh38
rs763897150 Chr.3: 112155072-112155072 on Build GRCh38
rs991920895 Chr.3: 112155068-112155068 on Build GRCh38
rs780869052 Chr.3: 112155070-112155070 on Build GRCh38
rs763385348 Chr.3: 112155067-112155068 on Build GRCh38
rs928137520 Chr.3: 112154910-112154910 on Build GRCh38
rs11369523 Chr.3: 112155055-112155056 on Build GRCh38
rs796441141 Chr.3: 112155056-112155057 on Build GRCh38
rs939429365 Chr.3: 112154926-112154926 on Build GRCh38
rs972563141 Chr.3: 112154930-112154930 on Build GRCh38
rs112312670 Chr.3: 112155070-112155071 on Build GRCh38
rs760610151 Chr.3: 112155088-112155088 on Build GRCh38
rs763007113 Chr.3: 112155070-112155070 on Build GRCh38
rs776366120 Chr.3: 112155025-112155025 on Build GRCh38
rs888349728 Chr.3: 112154970-112154971 on Build GRCh38
rs575908855 Chr.3: 112155056-112155056 on Build GRCh38
rs769750802 Chr.3: 112155035-112155035 on Build GRCh38
rs201930245 Chr.3: 112155069-112155070 on Build GRCh38
rs751905678 Chr.3: 112155068-112155069 on Build GRCh38
rs779248025 Chr.3: 112154999-112154999 on Build GRCh38
rs754212558 Chr.3: 112154969-112154969 on Build GRCh38
rs748696000 Chr.3: 112155000-112155000 on Build GRCh38
rs762950840 Chr.3: 112154950-112154950 on Build GRCh38
rs773343674 Chr.3: 112155084-112155084 on Build GRCh38
rs761849563 Chr.3: 112155065-112155066 on Build GRCh38
rs199505555 Chr.3: 112154948-112154948 on Build GRCh38
rs770622085 Chr.3: 112155024-112155024 on Build GRCh38
rs752450134 Chr.3: 112155097-112155098 on Build GRCh38
rs750925375 Chr.3: 112155067-112155070 on Build GRCh38
rs368331012 Chr.3: 112154965-112154965 on Build GRCh38
rs750420762 Chr.3: 112154960-112154960 on Build GRCh38
rs1033516333 Chr.3: 112154986-112154986 on Build GRCh38
rs560064004 Chr.3: 112154966-112154966 on Build GRCh38
rs758813597 Chr.3: 112155008-112155008 on Build GRCh38
rs139017505 Chr.3: 112155022-112155022 on Build GRCh38
rs796567815 Chr.3: 112155069-112155070 on Build GRCh38

View Full Product Details