Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 14133594-14133829 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD83
Entrez Gene ID
9308
Gene Name
CD83 molecule
Gene Aliases
BL11, HB15
UniGene
Hs.595133
Gene Chromosome Location
Chr.6: 14117256-14136918 on Build GRCh38
Transcripts
NM_001040280.1 NM_001251901.1 NM_004233.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5482021 c.489+6T>C p.? Chr.6: 14133761-14133761 on Build GRCh38
287958 c.429G>A p.A143A Chr.6: 14133695-14133695 on Build GRCh38
5554548 c.383G>T p.G128V Chr.6: 14133649-14133649 on Build GRCh38
6228170 c.428C>T p.A143V Chr.6: 14133694-14133694 on Build GRCh38
1568045 c.405A>T p.E135D Chr.6: 14133671-14133671 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs916007578 Chr.6: 14133741-14133741 on Build GRCh38
rs749112220 Chr.6: 14133800-14133801 on Build GRCh38
rs753945558 Chr.6: 14133668-14133668 on Build GRCh38
rs768336281 Chr.6: 14133630-14133630 on Build GRCh38
rs761664037 Chr.6: 14133633-14133633 on Build GRCh38
rs371626841 Chr.6: 14133788-14133788 on Build GRCh38
rs776776232 Chr.6: 14133791-14133791 on Build GRCh38
rs781703320 Chr.6: 14133751-14133751 on Build GRCh38
rs747700115 Chr.6: 14133780-14133780 on Build GRCh38
rs767692885 Chr.6: 14133803-14133804 on Build GRCh38
rs757715128 Chr.6: 14133737-14133737 on Build GRCh38
rs375070046 Chr.6: 14133804-14133804 on Build GRCh38
rs771857367 Chr.6: 14133782-14133782 on Build GRCh38
rs778041658 Chr.6: 14133723-14133723 on Build GRCh38
rs759573925 Chr.6: 14133692-14133692 on Build GRCh38
rs778614974 Chr.6: 14133777-14133777 on Build GRCh38
rs770478942 Chr.6: 14133638-14133638 on Build GRCh38
rs771821727 Chr.6: 14133644-14133644 on Build GRCh38
rs775589485 Chr.6: 14133632-14133632 on Build GRCh38
rs751954501 Chr.6: 14133732-14133732 on Build GRCh38
rs745629653 Chr.6: 14133770-14133777 on Build GRCh38
rs868083425 Chr.6: 14133712-14133712 on Build GRCh38
rs770960046 Chr.6: 14133789-14133789 on Build GRCh38
rs1051599205 Chr.6: 14133745-14133745 on Build GRCh38
rs112090479 Chr.6: 14133702-14133702 on Build GRCh38
rs758657694 Chr.6: 14133708-14133708 on Build GRCh38
rs768794015 Chr.6: 14133802-14133803 on Build GRCh38
rs775524882 Chr.6: 14133695-14133695 on Build GRCh38
rs569054787 Chr.6: 14133622-14133622 on Build GRCh38
rs367551619 Chr.6: 14133768-14133768 on Build GRCh38
rs770243436 Chr.6: 14133676-14133676 on Build GRCh38
rs973241483 Chr.6: 14133648-14133648 on Build GRCh38
rs201764790 Chr.6: 14133663-14133663 on Build GRCh38
rs781201443 Chr.6: 14133770-14133782 on Build GRCh38
rs773090892 Chr.6: 14133785-14133785 on Build GRCh38
rs773156134 Chr.6: 14133650-14133650 on Build GRCh38
rs769902095 Chr.6: 14133776-14133778 on Build GRCh38
rs199946337 Chr.6: 14133754-14133754 on Build GRCh38
rs180756952 Chr.6: 14133664-14133664 on Build GRCh38
rs775591705 Chr.6: 14133782-14133783 on Build GRCh38
rs200108463 Chr.6: 14133694-14133694 on Build GRCh38
rs1057108136 Chr.6: 14133696-14133696 on Build GRCh38
rs768117958 Chr.6: 14133774-14133774 on Build GRCh38

View Full Product Details