Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 68879020-68879247 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACTN1
Entrez Gene ID
87
Gene Name
actinin alpha 1
Gene Aliases
BDPLT15
UniGene
Hs.235750
Gene Chromosome Location
Chr.14: 68874123-68979366 on Build GRCh38
Transcripts
NM_001102.3 NM_001130004.1 NM_001130005.1 XM_011537265.2 XM_011537266.2 XM_011537267.2 XM_011537268.2 XM_017021720.1 XM_017021721.1 XM_017021722.1 XM_017021723.1 XM_017021725.1 XM_017021726.1 XM_017021727.1 XM_017021728.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6592962 c.2289C>T p.S763S Chr.14: 68879061-68879061 on Build GRCh38
6141046 c.2323C>T p.L775F Chr.14: 68879027-68879027 on Build GRCh38
6882609 c.2281-1G>A p.? Chr.14: 68879070-68879070 on Build GRCh38
5740143 c.2327T>A p.I776N Chr.14: 68879023-68879023 on Build GRCh38
5740144 c.2327T>A p.I776N Chr.14: 68879023-68879023 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1010584851 Chr.14: 68879144-68879144 on Build GRCh38
rs529401816 Chr.14: 68879162-68879162 on Build GRCh38
rs747842736 Chr.14: 68879078-68879078 on Build GRCh38
rs1140976 Chr.14: 68879047-68879047 on Build GRCh38
rs779488360 Chr.14: 68879142-68879142 on Build GRCh38
rs1030918931 Chr.14: 68879227-68879227 on Build GRCh38
rs1133206 Chr.14: 68879061-68879061 on Build GRCh38
rs768216429 Chr.14: 68879046-68879046 on Build GRCh38
rs111972819 Chr.14: 68879080-68879080 on Build GRCh38
rs762353765 Chr.14: 68879095-68879095 on Build GRCh38
rs765447500 Chr.14: 68879119-68879119 on Build GRCh38
rs932786559 Chr.14: 68879217-68879217 on Build GRCh38
rs921429610 Chr.14: 68879203-68879203 on Build GRCh38
rs532055520 Chr.14: 68879154-68879154 on Build GRCh38
rs202127204 Chr.14: 68879084-68879084 on Build GRCh38
rs111358019 Chr.14: 68879124-68879124 on Build GRCh38
rs181459 Chr.14: 68879161-68879161 on Build GRCh38
rs976242247 Chr.14: 68879218-68879218 on Build GRCh38
rs1133209 Chr.14: 68879056-68879056 on Build GRCh38
rs764887367 Chr.14: 68879185-68879196 on Build GRCh38
rs35909929 Chr.14: 68879157-68879157 on Build GRCh38
rs1133204 Chr.14: 68879063-68879063 on Build GRCh38
rs112075717 Chr.14: 68879125-68879125 on Build GRCh38
rs759510599 Chr.14: 68879094-68879094 on Build GRCh38
rs1054224 Chr.14: 68879043-68879043 on Build GRCh38
rs564590298 Chr.14: 68879151-68879151 on Build GRCh38
rs1133203 Chr.14: 68879067-68879067 on Build GRCh38
rs867081456 Chr.14: 68879195-68879195 on Build GRCh38
rs963252792 Chr.14: 68879191-68879191 on Build GRCh38
rs771815018 Chr.14: 68879079-68879079 on Build GRCh38
rs770779322 Chr.14: 68879085-68879085 on Build GRCh38
rs776686913 Chr.14: 68879091-68879091 on Build GRCh38
rs1133211 Chr.14: 68879050-68879050 on Build GRCh38
rs546461414 Chr.14: 68879135-68879135 on Build GRCh38
rs547939498 Chr.14: 68879201-68879201 on Build GRCh38
rs964840720 Chr.14: 68879185-68879185 on Build GRCh38
rs573824008 Chr.14: 68879155-68879155 on Build GRCh38

View Full Product Details