Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 21219778-21220033 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ECE1
Entrez Gene ID
1889
Gene Name
endothelin converting enzyme 1
Gene Aliases
ECE
UniGene
Hs.195080
Gene Chromosome Location
Chr.1: 21217247-21345541 on Build GRCh38
Transcripts
NM_001113347.1 NM_001113348.1 NM_001113349.1 NM_001397.2 XM_006710398.2 XM_011540872.2 XM_011540873.2 XM_017000511.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3864434 c.2302G>A p.E768K Chr.1: 21219966-21219966 on Build GRCh38
3483302 c.2271C>T p.V757V Chr.1: 21219961-21219961 on Build GRCh38
425286 c.2259C>G p.F753L Chr.1: 21220009-21220009 on Build GRCh38
4815973 c.2250C>G p.F750L Chr.1: 21220009-21220009 on Build GRCh38
3483300 c.2307C>T p.V769V Chr.1: 21219961-21219961 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs186453862 Chr.1: 21219981-21219981 on Build GRCh38
rs759455523 Chr.1: 21219956-21219956 on Build GRCh38
rs1043466955 Chr.1: 21219958-21219958 on Build GRCh38
rs971689513 Chr.1: 21219830-21219830 on Build GRCh38
rs3026907 Chr.1: 21219863-21219863 on Build GRCh38
rs201631426 Chr.1: 21219966-21219966 on Build GRCh38
rs372073084 Chr.1: 21219809-21219809 on Build GRCh38
rs778929118 Chr.1: 21219989-21219989 on Build GRCh38
rs909316602 Chr.1: 21219867-21219867 on Build GRCh38
rs780575401 Chr.1: 21219995-21219995 on Build GRCh38
rs1053393631 Chr.1: 21219902-21219902 on Build GRCh38
rs746368102 Chr.1: 21219914-21219914 on Build GRCh38
rs952704364 Chr.1: 21219885-21219885 on Build GRCh38
rs1039203230 Chr.1: 21219883-21219883 on Build GRCh38
rs765890802 Chr.1: 21219948-21219948 on Build GRCh38
rs933579238 Chr.1: 21219892-21219892 on Build GRCh38
rs747225058 Chr.1: 21219924-21219932 on Build GRCh38
rs774276352 Chr.1: 21219952-21219952 on Build GRCh38
rs752479442 Chr.1: 21219967-21219967 on Build GRCh38
rs750929744 Chr.1: 21220006-21220006 on Build GRCh38
rs780518855 Chr.1: 21219857-21219857 on Build GRCh38
rs200941917 Chr.1: 21219945-21219945 on Build GRCh38
rs967547745 Chr.1: 21219833-21219833 on Build GRCh38
rs558275025 Chr.1: 21219881-21219881 on Build GRCh38
rs533630890 Chr.1: 21219869-21219869 on Build GRCh38
rs200355455 Chr.1: 21219946-21219946 on Build GRCh38
rs754134273 Chr.1: 21219980-21219980 on Build GRCh38
rs1003005269 Chr.1: 21219838-21219838 on Build GRCh38
rs867348639 Chr.1: 21219822-21219822 on Build GRCh38
rs374518508 Chr.1: 21220007-21220007 on Build GRCh38
rs772481878 Chr.1: 21219922-21219922 on Build GRCh38
rs919953826 Chr.1: 21219876-21219876 on Build GRCh38
rs570101227 Chr.1: 21219931-21219931 on Build GRCh38
rs943497408 Chr.1: 21219874-21219874 on Build GRCh38
rs775777220 Chr.1: 21219930-21219930 on Build GRCh38
rs28368053 Chr.1: 21219854-21219854 on Build GRCh38
rs3026906 Chr.1: 21220008-21220008 on Build GRCh38
rs3026908 Chr.1: 21219821-21219821 on Build GRCh38
rs138594584 Chr.1: 21219979-21219979 on Build GRCh38
rs779511640 Chr.1: 21219907-21219907 on Build GRCh38
rs950744197 Chr.1: 21219858-21219858 on Build GRCh38

View Full Product Details