Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 36003488-36003715 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SYNE4
Entrez Gene ID
163183
Gene Name
spectrin repeat containing nuclear envelope family member 4
Gene Aliases
C19orf46, DFNB76, Nesp4
UniGene
Hs.436743
Gene Chromosome Location
Chr.19: 36003299-36008793 on Build GRCh38
Transcripts
NM_001039876.2 NM_001297735.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7277107 c.1032-3C>A p.? Chr.19: 36003523-36003523 on Build GRCh38
474607 c.1013G>C p.R338T Chr.19: 36003631-36003631 on Build GRCh38
1198328 c.1033G>A p.A345T Chr.19: 36003519-36003519 on Build GRCh38
3892378 c.1019A>G p.E340G Chr.19: 36003625-36003625 on Build GRCh38
6246283 c.1027C>A p.P343T Chr.19: 36003617-36003617 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771636023 Chr.19: 36003548-36003548 on Build GRCh38
rs796629487 Chr.19: 36003521-36003521 on Build GRCh38
rs768526921 Chr.19: 36003534-36003534 on Build GRCh38
rs748131078 Chr.19: 36003666-36003666 on Build GRCh38
rs749079483 Chr.19: 36003650-36003650 on Build GRCh38
rs749239360 Chr.19: 36003527-36003527 on Build GRCh38
rs769650086 Chr.19: 36003513-36003513 on Build GRCh38
rs980803257 Chr.19: 36003678-36003678 on Build GRCh38
rs1012249341 Chr.19: 36003553-36003553 on Build GRCh38
rs753117827 Chr.19: 36003593-36003593 on Build GRCh38
rs772794903 Chr.19: 36003550-36003550 on Build GRCh38
rs772774867 Chr.19: 36003680-36003680 on Build GRCh38
rs984302116 Chr.19: 36003662-36003662 on Build GRCh38
rs997976520 Chr.19: 36003613-36003613 on Build GRCh38
rs752047448 Chr.19: 36003675-36003675 on Build GRCh38
rs765581677 Chr.19: 36003590-36003590 on Build GRCh38
rs903039691 Chr.19: 36003530-36003530 on Build GRCh38
rs202107534 Chr.19: 36003522-36003522 on Build GRCh38
rs565733225 Chr.19: 36003631-36003631 on Build GRCh38
rs138824140 Chr.19: 36003578-36003578 on Build GRCh38
rs182578516 Chr.19: 36003544-36003544 on Build GRCh38
rs752136367 Chr.19: 36003609-36003609 on Build GRCh38
rs756886989 Chr.19: 36003544-36003546 on Build GRCh38
rs375197667 Chr.19: 36003654-36003654 on Build GRCh38
rs946956684 Chr.19: 36003514-36003514 on Build GRCh38
rs200948076 Chr.19: 36003619-36003619 on Build GRCh38
rs773883361 Chr.19: 36003536-36003536 on Build GRCh38
rs745354036 Chr.19: 36003662-36003664 on Build GRCh38
rs780869312 Chr.19: 36003548-36003549 on Build GRCh38
rs763492507 Chr.19: 36003594-36003594 on Build GRCh38
rs369956041 Chr.19: 36003589-36003589 on Build GRCh38
rs186898202 Chr.19: 36003624-36003624 on Build GRCh38
rs778667624 Chr.19: 36003660-36003660 on Build GRCh38
rs963795614 Chr.19: 36003689-36003689 on Build GRCh38
rs779976181 Chr.19: 36003649-36003649 on Build GRCh38
rs756466891 Chr.19: 36003647-36003647 on Build GRCh38
rs764547435 Chr.19: 36003607-36003607 on Build GRCh38
rs369635644 Chr.19: 36003656-36003656 on Build GRCh38

View Full Product Details