Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 206962375-206962648 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPO
Entrez Gene ID
130749
Gene Name
carboxypeptidase O
Gene Aliases
-
UniGene
Hs.684103
Gene Chromosome Location
Chr.2: 206932853-206971064 on Build GRCh38
Transcripts
NM_173077.2 XM_017003372.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4674418 c.599A>G p.Q200R Chr.2: 206962436-206962436 on Build GRCh38
5671235 c.750A>T p.K250N Chr.2: 206962587-206962587 on Build GRCh38
1565132 c.591A>G p.R197R Chr.2: 206962428-206962428 on Build GRCh38
4674419 c.625C>T p.P209S Chr.2: 206962462-206962462 on Build GRCh38
3991003 c.738C>T p.Y246Y Chr.2: 206962575-206962575 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376177466 Chr.2: 206962527-206962527 on Build GRCh38
rs764554400 Chr.2: 206962480-206962480 on Build GRCh38
rs1026086088 Chr.2: 206962403-206962403 on Build GRCh38
rs372729656 Chr.2: 206962551-206962551 on Build GRCh38
rs758305924 Chr.2: 206962506-206962506 on Build GRCh38
rs954361585 Chr.2: 206962481-206962481 on Build GRCh38
rs150008911 Chr.2: 206962609-206962609 on Build GRCh38
rs769421050 Chr.2: 206962541-206962541 on Build GRCh38
rs756313840 Chr.2: 206962598-206962598 on Build GRCh38
rs773069132 Chr.2: 206962420-206962420 on Build GRCh38
rs770374701 Chr.2: 206962437-206962437 on Build GRCh38
rs902899522 Chr.2: 206962494-206962494 on Build GRCh38
rs775825532 Chr.2: 206962479-206962479 on Build GRCh38
rs376068190 Chr.2: 206962504-206962504 on Build GRCh38
rs763265458 Chr.2: 206962547-206962547 on Build GRCh38
rs766674534 Chr.2: 206962602-206962602 on Build GRCh38
rs1057191742 Chr.2: 206962503-206962503 on Build GRCh38
rs150814545 Chr.2: 206962533-206962533 on Build GRCh38
rs921080286 Chr.2: 206962460-206962460 on Build GRCh38
rs370258208 Chr.2: 206962546-206962546 on Build GRCh38
rs1013155662 Chr.2: 206962543-206962543 on Build GRCh38
rs373047161 Chr.2: 206962398-206962398 on Build GRCh38
rs768140084 Chr.2: 206962575-206962575 on Build GRCh38
rs750459789 Chr.2: 206962576-206962576 on Build GRCh38
rs772125020 Chr.2: 206962417-206962417 on Build GRCh38
rs746493917 Chr.2: 206962421-206962421 on Build GRCh38
rs754142292 Chr.2: 206962613-206962613 on Build GRCh38
rs1008986901 Chr.2: 206962493-206962493 on Build GRCh38
rs142933512 Chr.2: 206962492-206962492 on Build GRCh38
rs62617140 Chr.2: 206962496-206962496 on Build GRCh38
rs752037970 Chr.2: 206962487-206962487 on Build GRCh38
rs758216012 Chr.2: 206962619-206962619 on Build GRCh38
rs143132020 Chr.2: 206962443-206962443 on Build GRCh38
rs780483658 Chr.2: 206962502-206962502 on Build GRCh38
rs564064517 Chr.2: 206962521-206962521 on Build GRCh38
rs777722610 Chr.2: 206962513-206962513 on Build GRCh38
rs756403695 Chr.2: 206962501-206962501 on Build GRCh38
rs765518342 Chr.2: 206962456-206962456 on Build GRCh38
rs750595243 Chr.2: 206962498-206962498 on Build GRCh38
rs947200279 Chr.2: 206962464-206962464 on Build GRCh38
rs777438246 Chr.2: 206962621-206962621 on Build GRCh38
rs373203943 Chr.2: 206962516-206962516 on Build GRCh38
rs148245128 Chr.2: 206962454-206962454 on Build GRCh38

View Full Product Details