Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 133763947-133764214 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VAV2
Entrez Gene ID
7410
Gene Name
vav guanine nucleotide exchange factor 2
Gene Aliases
VAV-2
UniGene
Hs.369921
Gene Chromosome Location
Chr.9: 133761894-133992374 on Build GRCh38
Transcripts
NM_001134398.1 NM_003371.3 XM_005272213.1 XM_017015108.1 XM_017015109.1 XM_017015110.1 XM_017015111.1 XM_017015112.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6819573 c.2614G>A p.V872I Chr.9: 133764085-133764085 on Build GRCh38
6837571 c.2594G>C p.G865A Chr.9: 133764105-133764105 on Build GRCh38
3213972 c.2493G>A p.T831T Chr.9: 133764089-133764089 on Build GRCh38
3213971 c.2610G>A p.T870T Chr.9: 133764089-133764089 on Build GRCh38
7409090 c.2589+1_2590-1del4332 p.? Chr.9: 133764110-133768441 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762978617 Chr.9: 133764097-133764097 on Build GRCh38
rs995661291 Chr.9: 133764162-133764162 on Build GRCh38
rs546006682 Chr.9: 133764086-133764086 on Build GRCh38
rs145903019 Chr.9: 133764061-133764061 on Build GRCh38
rs764403128 Chr.9: 133764141-133764142 on Build GRCh38
rs373904140 Chr.9: 133764039-133764039 on Build GRCh38
rs140064059 Chr.9: 133764089-133764089 on Build GRCh38
rs763313987 Chr.9: 133764140-133764140 on Build GRCh38
rs753390123 Chr.9: 133764138-133764138 on Build GRCh38
rs761626097 Chr.9: 133764135-133764138 on Build GRCh38
rs751377141 Chr.9: 133764100-133764100 on Build GRCh38
rs770345170 Chr.9: 133764135-133764140 on Build GRCh38
rs867276571 Chr.9: 133764079-133764079 on Build GRCh38
rs961760785 Chr.9: 133764066-133764066 on Build GRCh38
rs375990263 Chr.9: 133764152-133764155 on Build GRCh38
rs761535533 Chr.9: 133764124-133764124 on Build GRCh38
rs775919154 Chr.9: 133764135-133764142 on Build GRCh38
rs748565136 Chr.9: 133764069-133764069 on Build GRCh38
rs144182765 Chr.9: 133764060-133764060 on Build GRCh38
rs774018223 Chr.9: 133764142-133764142 on Build GRCh38
rs370254455 Chr.9: 133764146-133764146 on Build GRCh38
rs761423431 Chr.9: 133764156-133764161 on Build GRCh38
rs398069041 Chr.9: 133764154-133764155 on Build GRCh38
rs752528032 Chr.9: 133764085-133764085 on Build GRCh38
rs66574029 Chr.9: 133764135-133764136 on Build GRCh38
rs758690866 Chr.9: 133764143-133764143 on Build GRCh38
rs560455532 Chr.9: 133764051-133764051 on Build GRCh38
rs544993353 Chr.9: 133764098-133764098 on Build GRCh38
rs750353903 Chr.9: 133763972-133763972 on Build GRCh38
rs749899069 Chr.9: 133764052-133764052 on Build GRCh38
rs771247930 Chr.9: 133764151-133764151 on Build GRCh38
rs192573366 Chr.9: 133764029-133764029 on Build GRCh38
rs745955798 Chr.9: 133764134-133764135 on Build GRCh38
rs747147137 Chr.9: 133764133-133764134 on Build GRCh38
rs750641917 Chr.9: 133764137-133764138 on Build GRCh38
rs143833447 Chr.9: 133764090-133764090 on Build GRCh38
rs34181788 Chr.9: 133764146-133764147 on Build GRCh38
rs376916130 Chr.9: 133764057-133764057 on Build GRCh38
rs777808414 Chr.9: 133764092-133764092 on Build GRCh38
rs565399399 Chr.9: 133764055-133764055 on Build GRCh38
rs36090317 Chr.9: 133764180-133764181 on Build GRCh38
rs754663111 Chr.9: 133763974-133763974 on Build GRCh38
rs748509140 Chr.9: 133764142-133764142 on Build GRCh38
rs777989394 Chr.9: 133764145-133764145 on Build GRCh38
rs755458659 Chr.9: 133764139-133764139 on Build GRCh38
rs776819403 Chr.9: 133764160-133764160 on Build GRCh38
rs952284970 Chr.9: 133764021-133764021 on Build GRCh38
rs755856236 Chr.9: 133764134-133764134 on Build GRCh38
rs779423151 Chr.9: 133764140-133764140 on Build GRCh38
rs767131738 Chr.9: 133764125-133764125 on Build GRCh38
rs2797825 Chr.9: 133764176-133764176 on Build GRCh38
rs745991667 Chr.9: 133764161-133764161 on Build GRCh38
rs1040843928 Chr.9: 133764178-133764178 on Build GRCh38
rs750057582 Chr.9: 133764130-133764130 on Build GRCh38
rs765870545 Chr.9: 133764136-133764136 on Build GRCh38
rs1029315526 Chr.9: 133763976-133763977 on Build GRCh38
rs1028198039 Chr.9: 133764019-133764019 on Build GRCh38
rs745919841 Chr.9: 133764191-133764191 on Build GRCh38
rs574379828 Chr.9: 133763988-133763988 on Build GRCh38

View Full Product Details