Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 49792159-49792432 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACAA2
Entrez Gene ID
10449
Gene Name
acetyl-CoA acyltransferase 2
Gene Aliases
DSAEC
UniGene
Hs.200136
Gene Chromosome Location
Chr.18: 49783504-49813881 on Build GRCh38
Transcripts
NM_006111.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5350453 c.604G>T p.E202* Chr.18: 49792301-49792301 on Build GRCh38
1680356 c.713T>C p.V238A Chr.18: 49792192-49792192 on Build GRCh38
318463 c.674C>T p.P225L Chr.18: 49792231-49792231 on Build GRCh38
6591447 c.651G>A p.M217I Chr.18: 49792254-49792254 on Build GRCh38
7318189 c.601G>A p.D201N Chr.18: 49792304-49792304 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748795760 Chr.18: 49792302-49792302 on Build GRCh38
rs982750026 Chr.18: 49792393-49792393 on Build GRCh38
rs745789283 Chr.18: 49792354-49792354 on Build GRCh38
rs1009831250 Chr.18: 49792399-49792399 on Build GRCh38
rs757044920 Chr.18: 49792345-49792345 on Build GRCh38
rs149436920 Chr.18: 49792313-49792313 on Build GRCh38
rs146126484 Chr.18: 49792263-49792263 on Build GRCh38
rs753311217 Chr.18: 49792244-49792244 on Build GRCh38
rs975559202 Chr.18: 49792362-49792362 on Build GRCh38
rs781715231 Chr.18: 49792340-49792343 on Build GRCh38
rs756586837 Chr.18: 49792233-49792233 on Build GRCh38
rs368383594 Chr.18: 49792332-49792332 on Build GRCh38
rs750745750 Chr.18: 49792230-49792230 on Build GRCh38
rs770791532 Chr.18: 49792268-49792268 on Build GRCh38
rs777908633 Chr.18: 49792246-49792247 on Build GRCh38
rs754463910 Chr.18: 49792245-49792245 on Build GRCh38
rs372860665 Chr.18: 49792185-49792185 on Build GRCh38
rs889650590 Chr.18: 49792319-49792319 on Build GRCh38
rs761055038 Chr.18: 49792311-49792311 on Build GRCh38
rs922499773 Chr.18: 49792386-49792386 on Build GRCh38
rs907358687 Chr.18: 49792249-49792249 on Build GRCh38
rs757630188 Chr.18: 49792336-49792338 on Build GRCh38
rs1050880347 Chr.18: 49792315-49792315 on Build GRCh38
rs938244328 Chr.18: 49792246-49792248 on Build GRCh38
rs746932369 Chr.18: 49792183-49792183 on Build GRCh38
rs1033363657 Chr.18: 49792382-49792382 on Build GRCh38
rs768089753 Chr.18: 49792363-49792364 on Build GRCh38
rs778504639 Chr.18: 49792338-49792338 on Build GRCh38
rs751926164 Chr.18: 49792260-49792261 on Build GRCh38
rs777434734 Chr.18: 49792259-49792259 on Build GRCh38
rs779140450 Chr.18: 49792365-49792365 on Build GRCh38
rs781056432 Chr.18: 49792352-49792352 on Build GRCh38
rs867174773 Chr.18: 49792199-49792199 on Build GRCh38
rs375657323 Chr.18: 49792310-49792310 on Build GRCh38
rs778578981 Chr.18: 49792247-49792247 on Build GRCh38
rs11549285 Chr.18: 49792256-49792256 on Build GRCh38
rs199574771 Chr.18: 49792234-49792234 on Build GRCh38
rs751360029 Chr.18: 49792343-49792343 on Build GRCh38
rs775462658 Chr.18: 49792208-49792208 on Build GRCh38
rs202206177 Chr.18: 49792182-49792182 on Build GRCh38
rs146690959 Chr.18: 49792218-49792218 on Build GRCh38
rs774775444 Chr.18: 49792294-49792294 on Build GRCh38
rs754341856 Chr.18: 49792317-49792317 on Build GRCh38
rs933802478 Chr.18: 49792387-49792387 on Build GRCh38
rs138932410 Chr.18: 49792289-49792289 on Build GRCh38
rs185485675 Chr.18: 49792235-49792235 on Build GRCh38
rs764456603 Chr.18: 49792215-49792215 on Build GRCh38
rs563405468 Chr.18: 49792336-49792336 on Build GRCh38
rs770834763 Chr.18: 49792193-49792193 on Build GRCh38
rs759116415 Chr.18: 49792325-49792325 on Build GRCh38
rs773190370 Chr.18: 49792181-49792181 on Build GRCh38
rs762141148 Chr.18: 49792295-49792295 on Build GRCh38
rs765418629 Chr.18: 49792186-49792187 on Build GRCh38
rs765105322 Chr.18: 49792231-49792231 on Build GRCh38
rs957587326 Chr.18: 49792283-49792283 on Build GRCh38
rs764737644 Chr.18: 49792330-49792330 on Build GRCh38
rs767983865 Chr.18: 49792224-49792224 on Build GRCh38
rs767782958 Chr.18: 49792298-49792298 on Build GRCh38
rs747783652 Chr.18: 49792255-49792255 on Build GRCh38
rs539821165 Chr.18: 49792352-49792357 on Build GRCh38
rs762337379 Chr.18: 49792221-49792221 on Build GRCh38
rs780026982 Chr.18: 49792288-49792288 on Build GRCh38
rs769798115 Chr.18: 49792359-49792359 on Build GRCh38
rs150438353 Chr.18: 49792209-49792209 on Build GRCh38
rs1019928690 Chr.18: 49792403-49792404 on Build GRCh38
rs752897678 Chr.18: 49792228-49792229 on Build GRCh38
rs769397385 Chr.18: 49792245-49792246 on Build GRCh38
rs762725005 Chr.18: 49792340-49792341 on Build GRCh38
rs748199893 Chr.18: 49792376-49792376 on Build GRCh38

View Full Product Details