Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 77000578-77000839 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLN5
Entrez Gene ID
1203
Gene Name
ceroid-lipofuscinosis, neuronal 5
Gene Aliases
NCL
UniGene
Hs.30213
Gene Chromosome Location
Chr.13: 76991924-77002517 on Build GRCh38
Transcripts
NM_006493.2 XM_011534917.2 XM_017020538.1

Gene Symbol
FBXL3
Entrez Gene ID
26224
Gene Name
F-box and leucine rich repeat protein 3
Gene Aliases
FBL3, FBL3A, FBXL3A
UniGene
Hs.508284
Gene Chromosome Location
Chr.13: 76992597-77027196 on Build GRCh38
Transcripts
NM_006493.2 XM_011534917.2 XM_017020538.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6991096 c.914A>G p.Y305C Chr.13: 77000659-77000659 on Build GRCh38
948482 c.1080T>G p.I360M Chr.13: 77000825-77000825 on Build GRCh38
948481 c.1016G>T p.R339I Chr.13: 77000761-77000761 on Build GRCh38
6222764 c.900C>G p.N300K Chr.13: 77000645-77000645 on Build GRCh38
948480 c.918A>G p.T306T Chr.13: 77000663-77000663 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762726568 Chr.13: 77000606-77000606 on Build GRCh38
rs146885902 Chr.13: 77000692-77000692 on Build GRCh38
rs760212878 Chr.13: 77000668-77000668 on Build GRCh38
rs376675270 Chr.13: 77000665-77000665 on Build GRCh38
rs1057518562 Chr.13: 77000623-77000623 on Build GRCh38
rs929479143 Chr.13: 77000656-77000656 on Build GRCh38
rs386833983 Chr.13: 77000700-77000715 on Build GRCh38
rs780204258 Chr.13: 77000722-77000722 on Build GRCh38
rs752135206 Chr.13: 77000691-77000691 on Build GRCh38
rs730882146 Chr.13: 77000680-77000680 on Build GRCh38
rs764495616 Chr.13: 77000685-77000685 on Build GRCh38
rs794727641 Chr.13: 77000667-77000667 on Build GRCh38
rs761675809 Chr.13: 77000648-77000648 on Build GRCh38
rs749457168 Chr.13: 77000723-77000723 on Build GRCh38
rs772050001 Chr.13: 77000777-77000777 on Build GRCh38
rs140580715 Chr.13: 77000752-77000752 on Build GRCh38
rs121908292 Chr.13: 77000799-77000799 on Build GRCh38
rs386833982 Chr.13: 77000664-77000664 on Build GRCh38
rs768487375 Chr.13: 77000607-77000607 on Build GRCh38
rs557876713 Chr.13: 77000784-77000784 on Build GRCh38
rs775752162 Chr.13: 77000603-77000603 on Build GRCh38
rs786204644 Chr.13: 77000669-77000670 on Build GRCh38
rs770688728 Chr.13: 77000761-77000761 on Build GRCh38
rs368551639 Chr.13: 77000677-77000677 on Build GRCh38
rs575331884 Chr.13: 77000696-77000696 on Build GRCh38
rs199609750 Chr.13: 77000704-77000704 on Build GRCh38
rs372403801 Chr.13: 77000643-77000643 on Build GRCh38
rs149668314 Chr.13: 77000813-77000813 on Build GRCh38
rs760172666 Chr.13: 77000807-77000807 on Build GRCh38
rs777477178 Chr.13: 77000644-77000644 on Build GRCh38
rs768449493 Chr.13: 77000730-77000730 on Build GRCh38
rs386833963 Chr.13: 77000771-77000771 on Build GRCh38
rs398124228 Chr.13: 77000653-77000653 on Build GRCh38
rs766007943 Chr.13: 77000673-77000673 on Build GRCh38
rs767032477 Chr.13: 77000652-77000652 on Build GRCh38

View Full Product Details