Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 46873520-46873767 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MDGA2
Entrez Gene ID
161357
Gene Name
MAM domain containing glycosylphosphatidylinositol anchor 2
Gene Aliases
MAMDC1, c14_5286
UniGene
Hs.436380
Gene Chromosome Location
Chr.14: 46836518-47675319 on Build GRCh38
Transcripts
NM_001113498.2 NM_182830.4 XM_011536519.2 XM_011536525.2 XM_017021060.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5418079 c.1711T>A p.Y571N Chr.14: 46873580-46873580 on Build GRCh38
4050951 c.1727G>A p.R576Q Chr.14: 46873564-46873564 on Build GRCh38
5036994 c.1757T>C p.L586P Chr.14: 46873534-46873534 on Build GRCh38
3690076 c.1757T>A p.L586H Chr.14: 46873534-46873534 on Build GRCh38
4766939 c.1712A>G p.Y571C Chr.14: 46873579-46873579 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs143219487 Chr.14: 46873734-46873734 on Build GRCh38
rs576098856 Chr.14: 46873727-46873728 on Build GRCh38
rs199870483 Chr.14: 46873556-46873556 on Build GRCh38
rs755698149 Chr.14: 46873641-46873641 on Build GRCh38
rs988622667 Chr.14: 46873733-46873733 on Build GRCh38
rs146917844 Chr.14: 46873620-46873620 on Build GRCh38
rs763589166 Chr.14: 46873630-46873630 on Build GRCh38
rs747148813 Chr.14: 46873571-46873571 on Build GRCh38
rs369108118 Chr.14: 46873723-46873723 on Build GRCh38
rs760135089 Chr.14: 46873675-46873675 on Build GRCh38
rs767268370 Chr.14: 46873640-46873640 on Build GRCh38
rs925879076 Chr.14: 46873724-46873724 on Build GRCh38
rs953401208 Chr.14: 46873717-46873717 on Build GRCh38
rs372542940 Chr.14: 46873635-46873635 on Build GRCh38
rs768932936 Chr.14: 46873583-46873583 on Build GRCh38
rs766687252 Chr.14: 46873617-46873617 on Build GRCh38
rs534321443 Chr.14: 46873678-46873678 on Build GRCh38
rs551716495 Chr.14: 46873686-46873686 on Build GRCh38
rs1049317974 Chr.14: 46873564-46873564 on Build GRCh38
rs759044294 Chr.14: 46873636-46873636 on Build GRCh38
rs749059593 Chr.14: 46873548-46873548 on Build GRCh38
rs1029441040 Chr.14: 46873677-46873677 on Build GRCh38
rs749226941 Chr.14: 46873643-46873645 on Build GRCh38
rs139302285 Chr.14: 46873669-46873669 on Build GRCh38
rs776503553 Chr.14: 46873594-46873594 on Build GRCh38
rs773898894 Chr.14: 46873628-46873628 on Build GRCh38

View Full Product Details