Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 220884375-220884640 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HLX
Entrez Gene ID
3142
Gene Name
H2.0 like homeobox
Gene Aliases
HB24, HLX1
UniGene
Hs.74870
Gene Chromosome Location
Chr.1: 220879401-220885059 on Build GRCh38
Transcripts
NM_021958.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6124974 c.1245C>T p.L415L Chr.1: 220884482-220884482 on Build GRCh38
6680881 c.1167C>A p.S389R Chr.1: 220884404-220884404 on Build GRCh38
4232226 c.1174G>A p.E392K Chr.1: 220884411-220884411 on Build GRCh38
3376978 c.1179G>C p.R393R Chr.1: 220884416-220884416 on Build GRCh38
1254272 c.1281C>T p.S427S Chr.1: 220884518-220884518 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756876388 Chr.1: 220884586-220884586 on Build GRCh38
rs904344115 Chr.1: 220884556-220884556 on Build GRCh38
rs753891975 Chr.1: 220884478-220884478 on Build GRCh38
rs779447590 Chr.1: 220884605-220884605 on Build GRCh38
rs1056733839 Chr.1: 220884616-220884616 on Build GRCh38
rs970357816 Chr.1: 220884608-220884608 on Build GRCh38
rs760801509 Chr.1: 220884523-220884523 on Build GRCh38
rs754236879 Chr.1: 220884590-220884590 on Build GRCh38
rs773260536 Chr.1: 220884468-220884468 on Build GRCh38
rs770002080 Chr.1: 220884442-220884442 on Build GRCh38
rs758136561 Chr.1: 220884438-220884438 on Build GRCh38
rs762570970 Chr.1: 220884554-220884554 on Build GRCh38
rs769686717 Chr.1: 220884621-220884621 on Build GRCh38
rs142918916 Chr.1: 220884515-220884515 on Build GRCh38
rs746271150 Chr.1: 220884441-220884441 on Build GRCh38
rs980746405 Chr.1: 220884619-220884619 on Build GRCh38
rs751147381 Chr.1: 220884492-220884492 on Build GRCh38
rs576515869 Chr.1: 220884589-220884589 on Build GRCh38
rs370407346 Chr.1: 220884477-220884477 on Build GRCh38
rs762859339 Chr.1: 220884603-220884605 on Build GRCh38
rs376040668 Chr.1: 220884466-220884466 on Build GRCh38
rs554006386 Chr.1: 220884459-220884459 on Build GRCh38
rs752255937 Chr.1: 220884480-220884480 on Build GRCh38
rs112404524 Chr.1: 220884448-220884448 on Build GRCh38
rs577385808 Chr.1: 220884467-220884467 on Build GRCh38
rs752628182 Chr.1: 220884552-220884552 on Build GRCh38
rs765292765 Chr.1: 220884551-220884551 on Build GRCh38
rs533709615 Chr.1: 220884490-220884490 on Build GRCh38
rs750199366 Chr.1: 220884516-220884517 on Build GRCh38
rs199521070 Chr.1: 220884409-220884409 on Build GRCh38
rs780386801 Chr.1: 220884497-220884497 on Build GRCh38
rs375578569 Chr.1: 220884449-220884449 on Build GRCh38
rs1000296405 Chr.1: 220884582-220884582 on Build GRCh38
rs773525622 Chr.1: 220884521-220884521 on Build GRCh38
rs920741102 Chr.1: 220884489-220884489 on Build GRCh38
rs756571777 Chr.1: 220884495-220884495 on Build GRCh38
rs748250119 Chr.1: 220884507-220884507 on Build GRCh38
rs373865414 Chr.1: 220884488-220884488 on Build GRCh38
rs762014745 Chr.1: 220884394-220884394 on Build GRCh38
rs760141381 Chr.1: 220884478-220884478 on Build GRCh38
rs180838062 Chr.1: 220884444-220884444 on Build GRCh38
rs775709151 Chr.1: 220884513-220884515 on Build GRCh38
rs769159332 Chr.1: 220884503-220884503 on Build GRCh38
rs562919553 Chr.1: 220884525-220884525 on Build GRCh38
rs772098064 Chr.1: 220884439-220884439 on Build GRCh38
rs771063150 Chr.1: 220884612-220884612 on Build GRCh38
rs760568441 Chr.1: 220884470-220884470 on Build GRCh38
rs745344396 Chr.1: 220884410-220884410 on Build GRCh38
rs751287844 Chr.1: 220884565-220884565 on Build GRCh38
rs763478463 Chr.1: 220884562-220884562 on Build GRCh38
rs931332553 Chr.1: 220884533-220884533 on Build GRCh38
rs760920344 Chr.1: 220884512-220884513 on Build GRCh38
rs776580614 Chr.1: 220884528-220884528 on Build GRCh38
rs764413572 Chr.1: 220884517-220884519 on Build GRCh38
rs759423699 Chr.1: 220884620-220884620 on Build GRCh38
rs779354027 Chr.1: 220884505-220884505 on Build GRCh38
rs766246350 Chr.1: 220884602-220884603 on Build GRCh38
rs144497660 Chr.1: 220884428-220884428 on Build GRCh38
rs749601568 Chr.1: 220884498-220884498 on Build GRCh38
rs754952890 Chr.1: 220884479-220884479 on Build GRCh38
rs755287073 Chr.1: 220884598-220884598 on Build GRCh38
rs371016648 Chr.1: 220884433-220884433 on Build GRCh38
rs901124459 Chr.1: 220884542-220884542 on Build GRCh38
rs368112101 Chr.1: 220884613-220884613 on Build GRCh38
rs759337756 Chr.1: 220884549-220884549 on Build GRCh38
rs77213368 Chr.1: 220884415-220884415 on Build GRCh38
rs772017707 Chr.1: 220884497-220884498 on Build GRCh38
rs774746609 Chr.1: 220884427-220884428 on Build GRCh38
rs757128468 Chr.1: 220884393-220884393 on Build GRCh38
rs11578466 Chr.1: 220884397-220884397 on Build GRCh38
rs772259478 Chr.1: 220884606-220884606 on Build GRCh38
rs768785090 Chr.1: 220884458-220884458 on Build GRCh38
rs868627186 Chr.1: 220884617-220884617 on Build GRCh38

View Full Product Details