Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 1839222-1839453 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MYT1L
Entrez Gene ID
23040
Gene Name
myelin transcription factor 1 like
Gene Aliases
MRD39, NZF1, ZC2H2C2, ZC2HC4B
UniGene
Hs.434418
Gene Chromosome Location
Chr.2: 1789113-2331388 on Build GRCh38
Transcripts
NM_001303052.1 NM_015025.3 XM_011510318.1 XM_011510319.1 XM_011510320.2 XM_011510321.2 XM_011510322.2 XM_011510323.1 XM_011510324.1 XM_011510325.1 XM_011510326.1 XM_011510327.1 XM_011510328.2 XM_017003604.1 XM_017003605.1 XM_017003606.1 XM_017003607.1 XM_017003608.1 XM_017003609.1 XM_017003610.1 XM_017003611.1 XM_017003612.1 XM_017003613.1 XM_017003614.1 XM_017003615.1 XM_017003616.1 XM_017003617.1 XM_017003618.1 XM_017003619.1 XM_017003620.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4774539 c.2958A>C p.K986N Chr.2: 1839271-1839271 on Build GRCh38
3575027 c.2981C>T p.S994F Chr.2: 1839242-1839242 on Build GRCh38
2711003 c.2942C>A p.A981E Chr.2: 1839287-1839287 on Build GRCh38
4411815 c.2998G>T p.V1000F Chr.2: 1839231-1839231 on Build GRCh38
4540685 c.2886G>A p.Q962Q Chr.2: 1839343-1839343 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745509665 Chr.2: 1839319-1839319 on Build GRCh38
rs373901652 Chr.2: 1839321-1839321 on Build GRCh38
rs767499945 Chr.2: 1839412-1839412 on Build GRCh38
rs768069026 Chr.2: 1839343-1839343 on Build GRCh38
rs372435818 Chr.2: 1839413-1839413 on Build GRCh38
rs755045694 Chr.2: 1839407-1839407 on Build GRCh38
rs766663726 Chr.2: 1839387-1839387 on Build GRCh38
rs200125635 Chr.2: 1839307-1839307 on Build GRCh38
rs779983229 Chr.2: 1839267-1839267 on Build GRCh38
rs755973131 Chr.2: 1839286-1839286 on Build GRCh38
rs773769801 Chr.2: 1839364-1839364 on Build GRCh38
rs888872136 Chr.2: 1839244-1839244 on Build GRCh38
rs777718481 Chr.2: 1839301-1839301 on Build GRCh38
rs778693756 Chr.2: 1839316-1839316 on Build GRCh38
rs541692429 Chr.2: 1839392-1839392 on Build GRCh38
rs762215015 Chr.2: 1839259-1839259 on Build GRCh38
rs571062392 Chr.2: 1839322-1839322 on Build GRCh38
rs942823146 Chr.2: 1839268-1839268 on Build GRCh38
rs573219610 Chr.2: 1839422-1839422 on Build GRCh38
rs750861804 Chr.2: 1839344-1839344 on Build GRCh38
rs749215430 Chr.2: 1839306-1839306 on Build GRCh38
rs763261790 Chr.2: 1839378-1839378 on Build GRCh38
rs757246743 Chr.2: 1839415-1839415 on Build GRCh38
rs1022482889 Chr.2: 1839363-1839363 on Build GRCh38
rs751569491 Chr.2: 1839400-1839400 on Build GRCh38
rs1052642314 Chr.2: 1839334-1839334 on Build GRCh38
rs762411264 Chr.2: 1839351-1839351 on Build GRCh38
rs62114684 Chr.2: 1839304-1839304 on Build GRCh38
rs59128763 Chr.2: 1839358-1839358 on Build GRCh38
rs758706264 Chr.2: 1839263-1839263 on Build GRCh38
rs756501070 Chr.2: 1839349-1839349 on Build GRCh38
rs754127474 Chr.2: 1839255-1839255 on Build GRCh38
rs760284404 Chr.2: 1839325-1839325 on Build GRCh38
rs778677295 Chr.2: 1839417-1839417 on Build GRCh38
rs909954796 Chr.2: 1839257-1839257 on Build GRCh38
rs751728106 Chr.2: 1839277-1839277 on Build GRCh38

View Full Product Details