Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 88508500-88508725 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GRM5
Entrez Gene ID
2915
Gene Name
glutamate metabotropic receptor 5
Gene Aliases
GPRC1E, MGLUR5, PPP1R86, mGlu5
UniGene
Hs.147361
Gene Chromosome Location
Chr.11: 88504576-89065971 on Build GRCh38
Transcripts
NM_000842.4 NM_001143831.2 XM_006718828.3 XM_011542792.1 XM_017017627.1 NR_049724.1

Gene Symbol
GRM5-AS1
Entrez Gene ID
100873989
Gene Name
GRM5 antisense RNA 1
Gene Aliases
-
UniGene
Hs.616625
Gene Chromosome Location
Chr.11: 88504576-88524054 on Build GRCh38
Transcripts
NM_000842.4 NM_001143831.2 XM_006718828.3 XM_011542792.1 XM_017017627.1 NR_049724.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6676096 c.3617C>A p.T1206N Chr.11: 88508614-88508614 on Build GRCh38
4840137 c.3419C>T p.S1140L Chr.11: 88508716-88508716 on Build GRCh38
197775 c.3536C>T p.S1179L Chr.11: 88508599-88508599 on Build GRCh38
6134787 c.3568T>A p.C1190S Chr.11: 88508663-88508663 on Build GRCh38
4037289 c.3632C>T p.S1211L Chr.11: 88508599-88508599 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs199876972 Chr.11: 88508552-88508552 on Build GRCh38
rs113729474 Chr.11: 88508584-88508584 on Build GRCh38
rs748365397 Chr.11: 88508543-88508543 on Build GRCh38
rs377166167 Chr.11: 88508692-88508692 on Build GRCh38
rs772242260 Chr.11: 88508615-88508615 on Build GRCh38
rs773235460 Chr.11: 88508556-88508556 on Build GRCh38
rs202102837 Chr.11: 88508578-88508578 on Build GRCh38
rs556337599 Chr.11: 88508560-88508560 on Build GRCh38
rs763820812 Chr.11: 88508676-88508676 on Build GRCh38
rs202193167 Chr.11: 88508601-88508601 on Build GRCh38
rs748471366 Chr.11: 88508543-88508544 on Build GRCh38
rs868559917 Chr.11: 88508604-88508604 on Build GRCh38
rs773121087 Chr.11: 88508641-88508643 on Build GRCh38
rs749964733 Chr.11: 88508695-88508695 on Build GRCh38
rs183106472 Chr.11: 88508598-88508598 on Build GRCh38
rs756881891 Chr.11: 88508586-88508586 on Build GRCh38
rs749291725 Chr.11: 88508690-88508691 on Build GRCh38
rs757942734 Chr.11: 88508566-88508566 on Build GRCh38
rs114055826 Chr.11: 88508558-88508558 on Build GRCh38
rs751301223 Chr.11: 88508680-88508680 on Build GRCh38
rs776624934 Chr.11: 88508653-88508653 on Build GRCh38
rs771038192 Chr.11: 88508652-88508652 on Build GRCh38
rs951989577 Chr.11: 88508671-88508671 on Build GRCh38
rs200023089 Chr.11: 88508540-88508540 on Build GRCh38
rs147529131 Chr.11: 88508527-88508528 on Build GRCh38
rs200782117 Chr.11: 88508546-88508546 on Build GRCh38
rs747948964 Chr.11: 88508579-88508579 on Build GRCh38
rs748552029 Chr.11: 88508612-88508612 on Build GRCh38
rs755375483 Chr.11: 88508703-88508703 on Build GRCh38
rs201195373 Chr.11: 88508626-88508626 on Build GRCh38
rs776609495 Chr.11: 88508562-88508562 on Build GRCh38
rs775077618 Chr.11: 88508670-88508670 on Build GRCh38
rs769526263 Chr.11: 88508616-88508617 on Build GRCh38
rs142032384 Chr.11: 88508632-88508632 on Build GRCh38
rs770127576 Chr.11: 88508565-88508565 on Build GRCh38
rs200293788 Chr.11: 88508614-88508614 on Build GRCh38
rs772359822 Chr.11: 88508544-88508544 on Build GRCh38
rs769780408 Chr.11: 88508669-88508669 on Build GRCh38
rs759373017 Chr.11: 88508563-88508563 on Build GRCh38
rs764983018 Chr.11: 88508564-88508564 on Build GRCh38
rs761407732 Chr.11: 88508685-88508685 on Build GRCh38
rs981744794 Chr.11: 88508523-88508523 on Build GRCh38
rs777993798 Chr.11: 88508619-88508619 on Build GRCh38
rs201909095 Chr.11: 88508549-88508549 on Build GRCh38
rs200248896 Chr.11: 88508662-88508662 on Build GRCh38
rs148163807 Chr.11: 88508602-88508602 on Build GRCh38

View Full Product Details