Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 63594329-63594602 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MDH1
Entrez Gene ID
4190
Gene Name
malate dehydrogenase 1
Gene Aliases
HEL-S-32, MDH-s, MDHA, MGC:1375, MOR2
UniGene
Hs.526521
Gene Chromosome Location
Chr.2: 63588609-63607197 on Build GRCh38
Transcripts
NM_001199111.1 NM_001199112.1 NM_001316374.1 NM_005917.3 XM_017004251.1

Gene Symbol
WDPCP
Entrez Gene ID
51057
Gene Name
WD repeat containing planar cell polarity effector
Gene Aliases
BBS15, C2orf86, CHDTHP, FRITZ, FRTZ
UniGene
Hs.414952
Gene Chromosome Location
Chr.2: 63120755-63840825 on Build GRCh38
Transcripts
NM_001199111.1 NM_001199112.1 NM_001316374.1 NM_005917.3 XM_017004251.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5993853 c.4-2A>C p.? Chr.2: 63594486-63594486 on Build GRCh38
6445163 c.68G>A p.S23N Chr.2: 63594552-63594552 on Build GRCh38
1022205 c.12A>G p.P4P Chr.2: 63594496-63594496 on Build GRCh38
4401010 c.111A>G p.S37S Chr.2: 63594541-63594541 on Build GRCh38
5993852 c.58-2A>C p.? Chr.2: 63594486-63594486 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778744809 Chr.2: 63594538-63594538 on Build GRCh38
rs192689149 Chr.2: 63594435-63594435 on Build GRCh38
rs772835220 Chr.2: 63594437-63594437 on Build GRCh38
rs764060722 Chr.2: 63594467-63594467 on Build GRCh38
rs938163666 Chr.2: 63594361-63594361 on Build GRCh38
rs761105284 Chr.2: 63594538-63594538 on Build GRCh38
rs748062233 Chr.2: 63594546-63594546 on Build GRCh38
rs781426519 Chr.2: 63594523-63594523 on Build GRCh38
rs915471037 Chr.2: 63594427-63594427 on Build GRCh38
rs1054008361 Chr.2: 63594493-63594493 on Build GRCh38
rs961159456 Chr.2: 63594365-63594365 on Build GRCh38
rs199668515 Chr.2: 63594551-63594551 on Build GRCh38
rs201353649 Chr.2: 63594449-63594449 on Build GRCh38
rs900615857 Chr.2: 63594466-63594466 on Build GRCh38
rs137923103 Chr.2: 63594533-63594533 on Build GRCh38
rs763617752 Chr.2: 63594462-63594462 on Build GRCh38
rs945636285 Chr.2: 63594430-63594430 on Build GRCh38
rs757312912 Chr.2: 63594518-63594518 on Build GRCh38
rs986132697 Chr.2: 63594436-63594436 on Build GRCh38
rs111388920 Chr.2: 63594536-63594536 on Build GRCh38
rs199777376 Chr.2: 63594438-63594438 on Build GRCh38
rs17028056 Chr.2: 63594529-63594529 on Build GRCh38
rs3209597 Chr.2: 63594545-63594545 on Build GRCh38
rs933411755 Chr.2: 63594401-63594401 on Build GRCh38
rs760221711 Chr.2: 63594448-63594448 on Build GRCh38
rs766565871 Chr.2: 63594395-63594395 on Build GRCh38
rs759926989 Chr.2: 63594450-63594450 on Build GRCh38
rs751703733 Chr.2: 63594472-63594472 on Build GRCh38
rs976992015 Chr.2: 63594384-63594384 on Build GRCh38
rs371816428 Chr.2: 63594455-63594455 on Build GRCh38

View Full Product Details