Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 71164096-71164369 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NLGN3
Entrez Gene ID
54413
Gene Name
neuroligin 3
Gene Aliases
HNL3
UniGene
Hs.438877
Gene Chromosome Location
Chr.X: 71144389-71175307 on Build GRCh38
Transcripts
NM_001166660.1 NM_001321276.1 NM_018977.3 NM_181303.1 XM_006724662.3 XM_006724663.3 XM_011530974.2 XM_017029597.1

SNP Detail

SNP ID Chromosome Location
rs756593582 Chr.X: 71164174-71164174 on Build GRCh38
rs555467811 Chr.X: 71164128-71164128 on Build GRCh38
rs745489501 Chr.X: 71164294-71164294 on Build GRCh38
rs768571101 Chr.X: 71164340-71164340 on Build GRCh38
rs750837111 Chr.X: 71164156-71164156 on Build GRCh38
rs1050434123 Chr.X: 71164225-71164225 on Build GRCh38
rs756882503 Chr.X: 71164254-71164254 on Build GRCh38
rs755458577 Chr.X: 71164235-71164235 on Build GRCh38
rs761899130 Chr.X: 71164345-71164345 on Build GRCh38
rs772207741 Chr.X: 71164138-71164139 on Build GRCh38
rs767734683 Chr.X: 71164125-71164125 on Build GRCh38
rs774322363 Chr.X: 71164342-71164342 on Build GRCh38
rs182899170 Chr.X: 71164212-71164212 on Build GRCh38
rs139333574 Chr.X: 71164250-71164250 on Build GRCh38
rs777156416 Chr.X: 71164240-71164240 on Build GRCh38
rs144424373 Chr.X: 71164276-71164276 on Build GRCh38
rs202038845 Chr.X: 71164339-71164339 on Build GRCh38
rs151276700 Chr.X: 71164222-71164222 on Build GRCh38
rs17854700 Chr.X: 71164306-71164306 on Build GRCh38
rs769642654 Chr.X: 71164309-71164309 on Build GRCh38

View Full Product Details