Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 24181718-24181965 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IPO4
Entrez Gene ID
79711
Gene Name
importin 4
Gene Aliases
Imp4
UniGene
Hs.411865
Gene Chromosome Location
Chr.14: 24180219-24188918 on Build GRCh38
Transcripts
NM_024658.3 NR_051979.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4692826 c.2861A>T p.H954L Chr.14: 24181790-24181790 on Build GRCh38
6686678 c.2850G>A p.A950A Chr.14: 24181801-24181801 on Build GRCh38
7098347 c.2844C>T p.L948L Chr.14: 24181807-24181807 on Build GRCh38
1159576 c.2873G>C p.R958P Chr.14: 24181778-24181778 on Build GRCh38
1210818 c.2857C>T p.R953* Chr.14: 24181794-24181794 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs7149758 Chr.14: 24181846-24181846 on Build GRCh38
rs769441387 Chr.14: 24181744-24181744 on Build GRCh38
rs770526078 Chr.14: 24181759-24181759 on Build GRCh38
rs535037141 Chr.14: 24181794-24181794 on Build GRCh38
rs535534622 Chr.14: 24181831-24181831 on Build GRCh38
rs756951096 Chr.14: 24181910-24181910 on Build GRCh38
rs780933654 Chr.14: 24181795-24181795 on Build GRCh38
rs769189381 Chr.14: 24181845-24181845 on Build GRCh38
rs779685958 Chr.14: 24181924-24181924 on Build GRCh38
rs753584891 Chr.14: 24181884-24181884 on Build GRCh38
rs1024379332 Chr.14: 24181915-24181915 on Build GRCh38
rs373492200 Chr.14: 24181801-24181801 on Build GRCh38
rs763009983 Chr.14: 24181872-24181872 on Build GRCh38
rs571236753 Chr.14: 24181779-24181779 on Build GRCh38
rs777156907 Chr.14: 24181942-24181942 on Build GRCh38
rs777578536 Chr.14: 24181883-24181883 on Build GRCh38
rs565997832 Chr.14: 24181793-24181793 on Build GRCh38
rs765326373 Chr.14: 24181871-24181871 on Build GRCh38
rs149511997 Chr.14: 24181917-24181918 on Build GRCh38
rs886960517 Chr.14: 24181783-24181783 on Build GRCh38
rs762102390 Chr.14: 24181785-24181785 on Build GRCh38
rs778367085 Chr.14: 24181916-24181916 on Build GRCh38
rs987664555 Chr.14: 24181877-24181877 on Build GRCh38
rs766469540 Chr.14: 24181876-24181876 on Build GRCh38
rs761247753 Chr.14: 24181840-24181840 on Build GRCh38
rs778510649 Chr.14: 24181806-24181806 on Build GRCh38
rs376781517 Chr.14: 24181799-24181799 on Build GRCh38
rs59484787 Chr.14: 24181761-24181761 on Build GRCh38
rs759872782 Chr.14: 24181752-24181752 on Build GRCh38
rs201683526 Chr.14: 24181811-24181811 on Build GRCh38
rs772413854 Chr.14: 24181764-24181764 on Build GRCh38
rs745525274 Chr.14: 24181918-24181918 on Build GRCh38
rs757900081 Chr.14: 24181920-24181920 on Build GRCh38
rs764546296 Chr.14: 24181780-24181780 on Build GRCh38
rs777523326 Chr.14: 24181800-24181800 on Build GRCh38
rs536271889 Chr.14: 24181941-24181941 on Build GRCh38
rs760122991 Chr.14: 24181762-24181762 on Build GRCh38
rs752682420 Chr.14: 24181878-24181878 on Build GRCh38
rs748926988 Chr.14: 24181753-24181753 on Build GRCh38
rs776201496 Chr.14: 24181837-24181837 on Build GRCh38
rs750428999 Chr.14: 24181789-24181789 on Build GRCh38
rs770452240 Chr.14: 24181802-24181802 on Build GRCh38
rs756016788 Chr.14: 24181879-24181879 on Build GRCh38
rs777470214 Chr.14: 24181751-24181751 on Build GRCh38
rs1038144935 Chr.14: 24181752-24181752 on Build GRCh38
rs762125166 Chr.14: 24181866-24181866 on Build GRCh38
rs555910130 Chr.14: 24181935-24181935 on Build GRCh38
rs765578935 Chr.14: 24181778-24181778 on Build GRCh38
rs202194751 Chr.14: 24181784-24181784 on Build GRCh38

View Full Product Details