Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 31940070-31940321 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HUNK
Entrez Gene ID
30811
Gene Name
hormonally up-regulated Neu-associated kinase
Gene Aliases
-
UniGene
Hs.109437
Gene Chromosome Location
Chr.21: 31873315-32004064 on Build GRCh38
Transcripts
NM_014586.1 XM_011529537.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3841757 c.570G>T p.E190D Chr.21: 31940180-31940180 on Build GRCh38
1615992 c.610+8delT p.? Chr.21: 31940228-31940228 on Build GRCh38
6588486 c.591C>A p.D197E Chr.21: 31940201-31940201 on Build GRCh38
5895248 c.610+7_610+8insT p.? Chr.21: 31940227-31940228 on Build GRCh38
6215014 c.571A>C p.N191H Chr.21: 31940181-31940181 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs905361796 Chr.21: 31940278-31940278 on Build GRCh38
rs543735908 Chr.21: 31940237-31940237 on Build GRCh38
rs746826739 Chr.21: 31940286-31940286 on Build GRCh38
rs750846417 Chr.21: 31940250-31940250 on Build GRCh38
rs754089328 Chr.21: 31940228-31940230 on Build GRCh38
rs112620195 Chr.21: 31940143-31940143 on Build GRCh38
rs766745856 Chr.21: 31940228-31940229 on Build GRCh38
rs898047451 Chr.21: 31940195-31940195 on Build GRCh38
rs76694801 Chr.21: 31940107-31940107 on Build GRCh38
rs370303186 Chr.21: 31940207-31940207 on Build GRCh38
rs773187261 Chr.21: 31940227-31940228 on Build GRCh38
rs1041523148 Chr.21: 31940280-31940280 on Build GRCh38
rs945891286 Chr.21: 31940266-31940266 on Build GRCh38
rs74265539 Chr.21: 31940240-31940241 on Build GRCh38
rs773875558 Chr.21: 31940154-31940154 on Build GRCh38
rs114747218 Chr.21: 31940261-31940261 on Build GRCh38
rs138391065 Chr.21: 31940144-31940144 on Build GRCh38
rs1046588212 Chr.21: 31940275-31940275 on Build GRCh38
rs185815073 Chr.21: 31940124-31940124 on Build GRCh38
rs758336767 Chr.21: 31940245-31940246 on Build GRCh38
rs773098476 Chr.21: 31940132-31940132 on Build GRCh38
rs565313614 Chr.21: 31940257-31940257 on Build GRCh38
rs777146835 Chr.21: 31940214-31940214 on Build GRCh38
rs759175027 Chr.21: 31940186-31940186 on Build GRCh38
rs758907844 Chr.21: 31940252-31940252 on Build GRCh38
rs199674931 Chr.21: 31940149-31940149 on Build GRCh38
rs762228666 Chr.21: 31940151-31940154 on Build GRCh38
rs747883843 Chr.21: 31940123-31940123 on Build GRCh38
rs781328654 Chr.21: 31940270-31940270 on Build GRCh38
rs113053808 Chr.21: 31940274-31940274 on Build GRCh38
rs763986674 Chr.21: 31940254-31940254 on Build GRCh38
rs768547280 Chr.21: 31940228-31940228 on Build GRCh38
rs767262010 Chr.21: 31940213-31940213 on Build GRCh38

View Full Product Details