Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 119471491-119471746 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HSD17B4
Entrez Gene ID
3295
Gene Name
hydroxysteroid 17-beta dehydrogenase 4
Gene Aliases
DBP, MFE-2, MPF-2, PRLTS1, SDR8C1
UniGene
Hs.406861
Gene Chromosome Location
Chr.5: 119452443-119542335 on Build GRCh38
Transcripts
NM_000414.3 NM_001199291.2 NM_001199292.1 NM_001292027.1 NM_001292028.1

SNP Detail

SNP ID Chromosome Location
rs532183192 Chr.5: 119471667-119471667 on Build GRCh38
rs998169535 Chr.5: 119471683-119471683 on Build GRCh38
rs577121550 Chr.5: 119471531-119471531 on Build GRCh38
rs563182715 Chr.5: 119471661-119471661 on Build GRCh38
rs876657476 Chr.5: 119471660-119471660 on Build GRCh38
rs545645799 Chr.5: 119471681-119471681 on Build GRCh38
rs1037542179 Chr.5: 119471648-119471648 on Build GRCh38
rs917206429 Chr.5: 119471550-119471550 on Build GRCh38
rs948534484 Chr.5: 119471569-119471569 on Build GRCh38
rs1044213859 Chr.5: 119471573-119471573 on Build GRCh38
rs941426644 Chr.5: 119471629-119471629 on Build GRCh38
rs781379700 Chr.5: 119471700-119471700 on Build GRCh38
rs778601926 Chr.5: 119471587-119471587 on Build GRCh38
rs574865676 Chr.5: 119471580-119471580 on Build GRCh38
rs997608360 Chr.5: 119471550-119471550 on Build GRCh38
rs897135971 Chr.5: 119471653-119471653 on Build GRCh38
rs1057061684 Chr.5: 119471543-119471543 on Build GRCh38
rs541081959 Chr.5: 119471702-119471702 on Build GRCh38
rs543211815 Chr.5: 119471620-119471620 on Build GRCh38
rs747939172 Chr.5: 119471634-119471634 on Build GRCh38
rs775728663 Chr.5: 119471626-119471626 on Build GRCh38
rs1029861879 Chr.5: 119471698-119471698 on Build GRCh38
rs953135740 Chr.5: 119471562-119471562 on Build GRCh38

View Full Product Details