Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 228424746-228424995 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HIST3H3
Entrez Gene ID
8290
Gene Name
histone cluster 3, H3
Gene Aliases
H3.4, H3/g, H3FT, H3t
UniGene
Hs.248171
Gene Chromosome Location
Chr.1: 228424845-228425325 on Build GRCh38
Transcripts
NM_003493.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6006700 c.392T>G p.I131S Chr.1: 228424934-228424934 on Build GRCh38
6680392 c.335T>C p.V112A Chr.1: 228424991-228424991 on Build GRCh38
6982617 c.379C>A p.L127M Chr.1: 228424947-228424947 on Build GRCh38
4881719 c.406G>T p.A136S Chr.1: 228424920-228424920 on Build GRCh38
6528124 c.376C>G p.Q126E Chr.1: 228424950-228424950 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375937549 Chr.1: 228424949-228424949 on Build GRCh38
rs371210381 Chr.1: 228424914-228424914 on Build GRCh38
rs199657441 Chr.1: 228424938-228424938 on Build GRCh38
rs778350730 Chr.1: 228424865-228424865 on Build GRCh38
rs753240133 Chr.1: 228424891-228424891 on Build GRCh38
rs554749991 Chr.1: 228424881-228424881 on Build GRCh38
rs767959555 Chr.1: 228424930-228424930 on Build GRCh38
rs746743703 Chr.1: 228424870-228424870 on Build GRCh38
rs745500445 Chr.1: 228424867-228424867 on Build GRCh38
rs749863422 Chr.1: 228424895-228424895 on Build GRCh38
rs755941555 Chr.1: 228424964-228424964 on Build GRCh38
rs543240992 Chr.1: 228424899-228424899 on Build GRCh38
rs779716010 Chr.1: 228424869-228424869 on Build GRCh38
rs773212063 Chr.1: 228424879-228424879 on Build GRCh38
rs372821871 Chr.1: 228424790-228424790 on Build GRCh38
rs759918684 Chr.1: 228424885-228424885 on Build GRCh38
rs780917924 Chr.1: 228424959-228424959 on Build GRCh38
rs979063507 Chr.1: 228424933-228424933 on Build GRCh38
rs969644587 Chr.1: 228424884-228424884 on Build GRCh38
rs1041234882 Chr.1: 228424824-228424824 on Build GRCh38
rs571588126 Chr.1: 228424868-228424868 on Build GRCh38
rs992665139 Chr.1: 228424770-228424770 on Build GRCh38
rs372508619 Chr.1: 228424886-228424886 on Build GRCh38
rs777513592 Chr.1: 228424966-228424966 on Build GRCh38
rs761134743 Chr.1: 228424935-228424935 on Build GRCh38
rs771129689 Chr.1: 228424972-228424972 on Build GRCh38
rs79427757 Chr.1: 228424778-228424778 on Build GRCh38
rs764686882 Chr.1: 228424893-228424893 on Build GRCh38
rs769567577 Chr.1: 228424907-228424907 on Build GRCh38
rs568800459 Chr.1: 228424792-228424792 on Build GRCh38
rs755729980 Chr.1: 228424809-228424809 on Build GRCh38
rs774697896 Chr.1: 228424916-228424916 on Build GRCh38
rs185324895 Chr.1: 228424880-228424880 on Build GRCh38
rs557871170 Chr.1: 228424975-228424975 on Build GRCh38
rs375752720 Chr.1: 228424896-228424896 on Build GRCh38
rs369956929 Chr.1: 228424871-228424871 on Build GRCh38
rs749296189 Chr.1: 228424968-228424968 on Build GRCh38
rs146830002 Chr.1: 228424937-228424937 on Build GRCh38
rs376689247 Chr.1: 228424902-228424902 on Build GRCh38
rs537705653 Chr.1: 228424804-228424804 on Build GRCh38
rs944169675 Chr.1: 228424821-228424821 on Build GRCh38
rs773231016 Chr.1: 228424911-228424911 on Build GRCh38
rs148262634 Chr.1: 228424960-228424960 on Build GRCh38
rs375878056 Chr.1: 228424812-228424812 on Build GRCh38
rs776537953 Chr.1: 228424872-228424872 on Build GRCh38
rs748101693 Chr.1: 228424875-228424875 on Build GRCh38
rs117182407 Chr.1: 228424945-228424945 on Build GRCh38
rs1048038897 Chr.1: 228424876-228424876 on Build GRCh38
rs936704892 Chr.1: 228424805-228424805 on Build GRCh38
rs749845388 Chr.1: 228424891-228424892 on Build GRCh38
rs754676797 Chr.1: 228424957-228424957 on Build GRCh38
rs759900631 Chr.1: 228424929-228424929 on Build GRCh38
rs771148126 Chr.1: 228424915-228424915 on Build GRCh38
rs897404180 Chr.1: 228424853-228424853 on Build GRCh38
rs545672641 Chr.1: 228424976-228424976 on Build GRCh38
rs780890873 Chr.1: 228424900-228424900 on Build GRCh38
rs376448367 Chr.1: 228424864-228424864 on Build GRCh38
rs911316704 Chr.1: 228424816-228424816 on Build GRCh38
rs751183313 Chr.1: 228424950-228424950 on Build GRCh38
rs959828877 Chr.1: 228424768-228424768 on Build GRCh38
rs370467281 Chr.1: 228424898-228424898 on Build GRCh38
rs368646350 Chr.1: 228424866-228424866 on Build GRCh38
rs762440854 Chr.1: 228424942-228424942 on Build GRCh38
rs775764888 Chr.1: 228424932-228424932 on Build GRCh38

View Full Product Details