Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 74776553-74776780 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NSA2
Entrez Gene ID
10412
Gene Name
NSA2, ribosome biogenesis homolog
Gene Aliases
CDK105, HCL-G1, HCLG1, HUSSY-29, HUSSY29, TINP1
UniGene
Hs.482526
Gene Chromosome Location
Chr.5: 74765429-74776912 on Build GRCh38
Transcripts
NM_001271665.1 NM_014886.4 NR_073403.1 XM_011543098.1 XM_017008952.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
163002 c.767C>T p.A256V Chr.5: 74776655-74776655 on Build GRCh38
3617860 c.734C>T p.T245I Chr.5: 74776622-74776622 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs767192679 Chr.5: 74776667-74776667 on Build GRCh38
rs754982004 Chr.5: 74776655-74776655 on Build GRCh38
rs777817735 Chr.5: 74776601-74776601 on Build GRCh38
rs772188965 Chr.5: 74776599-74776599 on Build GRCh38
rs971005502 Chr.5: 74776724-74776724 on Build GRCh38
rs1002176320 Chr.5: 74776688-74776688 on Build GRCh38
rs776158945 Chr.5: 74776750-74776757 on Build GRCh38
rs563089290 Chr.5: 74776622-74776622 on Build GRCh38
rs755276767 Chr.5: 74776587-74776587 on Build GRCh38
rs960456150 Chr.5: 74776739-74776739 on Build GRCh38
rs749944235 Chr.5: 74776670-74776670 on Build GRCh38
rs767643348 Chr.5: 74776627-74776635 on Build GRCh38
rs753394751 Chr.5: 74776694-74776694 on Build GRCh38
rs779227515 Chr.5: 74776705-74776705 on Build GRCh38
rs766068134 Chr.5: 74776687-74776687 on Build GRCh38
rs754231676 Chr.5: 74776580-74776580 on Build GRCh38
rs1054287343 Chr.5: 74776663-74776663 on Build GRCh38
rs749273258 Chr.5: 74776704-74776718 on Build GRCh38
rs1000466285 Chr.5: 74776662-74776662 on Build GRCh38
rs777799061 Chr.5: 74776687-74776688 on Build GRCh38
rs768529192 Chr.5: 74776639-74776639 on Build GRCh38
rs758906071 Chr.5: 74776711-74776711 on Build GRCh38
rs756253755 Chr.5: 74776686-74776687 on Build GRCh38
rs200381888 Chr.5: 74776685-74776685 on Build GRCh38
rs771192359 Chr.5: 74776609-74776609 on Build GRCh38
rs755377355 Chr.5: 74776695-74776695 on Build GRCh38
rs371734076 Chr.5: 74776673-74776673 on Build GRCh38
rs368472594 Chr.5: 74776645-74776645 on Build GRCh38
rs762826781 Chr.5: 74776628-74776628 on Build GRCh38
rs761816280 Chr.5: 74776665-74776665 on Build GRCh38
rs778078997 Chr.5: 74776719-74776719 on Build GRCh38
rs970115546 Chr.5: 74776686-74776688 on Build GRCh38
rs117064539 Chr.5: 74776686-74776686 on Build GRCh38
rs752949774 Chr.5: 74776685-74776686 on Build GRCh38
rs755300985 Chr.5: 74776624-74776624 on Build GRCh38
rs753262891 Chr.5: 74776709-74776709 on Build GRCh38
rs747265400 Chr.5: 74776603-74776603 on Build GRCh38
rs779429219 Chr.5: 74776598-74776598 on Build GRCh38
rs751896669 Chr.5: 74776602-74776606 on Build GRCh38

View Full Product Details