Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 5199543-5199810 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OR51V1
Entrez Gene ID
283111
Gene Name
olfactory receptor family 51 subfamily V member 1
Gene Aliases
OR11-36, OR51A12
UniGene
Hs.553670
Gene Chromosome Location
Chr.11: 5199735-5200700 on Build GRCh38
Transcripts
NM_001004760.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4512801 c.916C>T p.Q306* Chr.11: 5199785-5199785 on Build GRCh38
348435 c.892C>A p.P298T Chr.11: 5199809-5199809 on Build GRCh38
3448532 c.925C>T p.H309Y Chr.11: 5199776-5199776 on Build GRCh38
6734704 c.915C>A p.T305T Chr.11: 5199786-5199786 on Build GRCh38
4033509 c.944T>C p.L315P Chr.11: 5199757-5199757 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1029625819 Chr.11: 5199736-5199736 on Build GRCh38
rs971779863 Chr.11: 5199582-5199582 on Build GRCh38
rs7933098 Chr.11: 5199607-5199607 on Build GRCh38
rs745528193 Chr.11: 5199728-5199728 on Build GRCh38
rs571224107 Chr.11: 5199621-5199625 on Build GRCh38
rs917575559 Chr.11: 5199673-5199673 on Build GRCh38
rs188175880 Chr.11: 5199734-5199734 on Build GRCh38
rs753117139 Chr.11: 5199705-5199705 on Build GRCh38
rs1045016249 Chr.11: 5199683-5199683 on Build GRCh38
rs759044535 Chr.11: 5199704-5199705 on Build GRCh38
rs889028112 Chr.11: 5199693-5199693 on Build GRCh38
rs911368698 Chr.11: 5199619-5199619 on Build GRCh38
rs770024750 Chr.11: 5199759-5199759 on Build GRCh38
rs1056126424 Chr.11: 5199667-5199667 on Build GRCh38
rs903146524 Chr.11: 5199664-5199664 on Build GRCh38
rs759856009 Chr.11: 5199776-5199776 on Build GRCh38
rs749113262 Chr.11: 5199563-5199563 on Build GRCh38
rs751951880 Chr.11: 5199692-5199692 on Build GRCh38
rs879274564 Chr.11: 5199713-5199713 on Build GRCh38
rs113527981 Chr.11: 5199724-5199724 on Build GRCh38
rs771192383 Chr.11: 5199738-5199738 on Build GRCh38
rs772176879 Chr.11: 5199616-5199616 on Build GRCh38
rs745308047 Chr.11: 5199697-5199700 on Build GRCh38
rs537278117 Chr.11: 5199695-5199695 on Build GRCh38
rs183625479 Chr.11: 5199649-5199649 on Build GRCh38
rs966174079 Chr.11: 5199580-5199580 on Build GRCh38
rs528999038 Chr.11: 5199613-5199613 on Build GRCh38
rs781613409 Chr.11: 5199703-5199703 on Build GRCh38
rs766769688 Chr.11: 5199688-5199688 on Build GRCh38
rs113679819 Chr.11: 5199701-5199703 on Build GRCh38
rs763370444 Chr.11: 5199685-5199685 on Build GRCh38
rs918894229 Chr.11: 5199585-5199585 on Build GRCh38
rs746148681 Chr.11: 5199776-5199777 on Build GRCh38
rs1010743366 Chr.11: 5199732-5199732 on Build GRCh38
rs965835299 Chr.11: 5199780-5199780 on Build GRCh38
rs548725688 Chr.11: 5199638-5199638 on Build GRCh38
rs1055882821 Chr.11: 5199645-5199645 on Build GRCh38
rs770941679 Chr.11: 5199654-5199656 on Build GRCh38
rs776481096 Chr.11: 5199747-5199747 on Build GRCh38
rs918324619 Chr.11: 5199659-5199659 on Build GRCh38
rs895931826 Chr.11: 5199646-5199646 on Build GRCh38
rs7951587 Chr.11: 5199612-5199612 on Build GRCh38
rs756805528 Chr.11: 5199722-5199722 on Build GRCh38
rs779888708 Chr.11: 5199742-5199742 on Build GRCh38
rs756693441 Chr.11: 5199781-5199781 on Build GRCh38
rs768041823 Chr.11: 5199777-5199777 on Build GRCh38
rs201009501 Chr.11: 5199771-5199771 on Build GRCh38
rs1041729733 Chr.11: 5199656-5199656 on Build GRCh38
rs753310174 Chr.11: 5199718-5199718 on Build GRCh38
rs781302604 Chr.11: 5199740-5199741 on Build GRCh38
rs534660866 Chr.11: 5199763-5199763 on Build GRCh38
rs1033082460 Chr.11: 5199588-5199588 on Build GRCh38
rs751986111 Chr.11: 5199736-5199736 on Build GRCh38
rs772735230 Chr.11: 5199695-5199699 on Build GRCh38
rs765965502 Chr.11: 5199703-5199704 on Build GRCh38
rs761936696 Chr.11: 5199751-5199751 on Build GRCh38
rs760966136 Chr.11: 5199687-5199687 on Build GRCh38
rs757489937 Chr.11: 5199737-5199738 on Build GRCh38
rs1000114977 Chr.11: 5199679-5199679 on Build GRCh38
rs772304606 Chr.11: 5199574-5199575 on Build GRCh38
rs754727998 Chr.11: 5199569-5199569 on Build GRCh38
rs773500431 Chr.11: 5199760-5199760 on Build GRCh38
rs764540524 Chr.11: 5199712-5199718 on Build GRCh38
rs1032966129 Chr.11: 5199682-5199682 on Build GRCh38
rs143609610 Chr.11: 5199778-5199778 on Build GRCh38

View Full Product Details