Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 128945920-128946147 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IRF5
Entrez Gene ID
3663
Gene Name
interferon regulatory factor 5
Gene Aliases
SLEB10
UniGene
Hs.521181
Gene Chromosome Location
Chr.7: 128937737-128950042 on Build GRCh38
Transcripts
NM_001098627.3 NM_001098629.2 NM_001098630.2 NM_001242452.2 NM_032643.4 XM_005250317.3 XM_006715974.2 XM_011516158.2 XM_011516159.2 XM_011516160.1 XM_011516161.1 XM_011516162.1 XM_011516163.2 XM_011516164.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5062175 c.331C>T p.P111S Chr.7: 128945980-128945980 on Build GRCh38
5871784 c.362G>A p.C121Y Chr.7: 128946011-128946011 on Build GRCh38
5500624 c.315C>T p.D105D Chr.7: 128945964-128945964 on Build GRCh38
6687272 c.355G>A p.E119K Chr.7: 128946004-128946004 on Build GRCh38
6225256 c.302G>A p.R101H Chr.7: 128945951-128945951 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777272768 Chr.7: 128945961-128945961 on Build GRCh38
rs773718383 Chr.7: 128946017-128946017 on Build GRCh38
rs74583517 Chr.7: 128946049-128946049 on Build GRCh38
rs573717331 Chr.7: 128946123-128946123 on Build GRCh38
rs748258245 Chr.7: 128946014-128946014 on Build GRCh38
rs761299912 Chr.7: 128946040-128946040 on Build GRCh38
rs761029569 Chr.7: 128945944-128945944 on Build GRCh38
rs750886286 Chr.7: 128945942-128945942 on Build GRCh38
rs187702198 Chr.7: 128946091-128946091 on Build GRCh38
rs772068444 Chr.7: 128946015-128946015 on Build GRCh38
rs769606450 Chr.7: 128945972-128945972 on Build GRCh38
rs745742459 Chr.7: 128945971-128945971 on Build GRCh38
rs766706257 Chr.7: 128945951-128945951 on Build GRCh38
rs751140082 Chr.7: 128945962-128945962 on Build GRCh38
rs759773700 Chr.7: 128946054-128946054 on Build GRCh38
rs139688977 Chr.7: 128946003-128946003 on Build GRCh38
rs766524164 Chr.7: 128946110-128946110 on Build GRCh38
rs779968332 Chr.7: 128945978-128945978 on Build GRCh38
rs200230921 Chr.7: 128945964-128945964 on Build GRCh38
rs199671607 Chr.7: 128945967-128945967 on Build GRCh38
rs768969857 Chr.7: 128945994-128945994 on Build GRCh38
rs938205424 Chr.7: 128946101-128946101 on Build GRCh38
rs777104512 Chr.7: 128946050-128946050 on Build GRCh38
rs1054048690 Chr.7: 128946102-128946102 on Build GRCh38
rs138098174 Chr.7: 128945952-128945952 on Build GRCh38
rs1017135613 Chr.7: 128946105-128946129 on Build GRCh38
rs749003004 Chr.7: 128945990-128945990 on Build GRCh38
rs928578383 Chr.7: 128946007-128946007 on Build GRCh38
rs758112859 Chr.7: 128945956-128945956 on Build GRCh38

View Full Product Details