Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 42155426-42155695 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GUCA2B
Entrez Gene ID
2981
Gene Name
guanylate cyclase activator 2B
Gene Aliases
GCAP-II, UGN
UniGene
Hs.32966
Gene Chromosome Location
Chr.1: 42153421-42155824 on Build GRCh38
Transcripts
NM_007102.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5550844 c.332G>A p.C111Y Chr.1: 42155589-42155589 on Build GRCh38
7370551 c.296A>T p.D99V Chr.1: 42155553-42155553 on Build GRCh38
3419232 c.292G>A p.D98N Chr.1: 42155549-42155549 on Build GRCh38
5691735 c.317T>A p.V106D Chr.1: 42155574-42155574 on Build GRCh38
909608 c.316G>A p.V106I Chr.1: 42155573-42155573 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs572716588 Chr.1: 42155615-42155615 on Build GRCh38
rs1032914101 Chr.1: 42155517-42155517 on Build GRCh38
rs752644829 Chr.1: 42155623-42155623 on Build GRCh38
rs747305345 Chr.1: 42155575-42155575 on Build GRCh38
rs145598813 Chr.1: 42155563-42155563 on Build GRCh38
rs1028771460 Chr.1: 42155468-42155468 on Build GRCh38
rs781213638 Chr.1: 42155555-42155555 on Build GRCh38
rs773317305 Chr.1: 42155580-42155580 on Build GRCh38
rs139544420 Chr.1: 42155542-42155542 on Build GRCh38
rs146925564 Chr.1: 42155572-42155572 on Build GRCh38
rs752268966 Chr.1: 42155560-42155560 on Build GRCh38
rs954694412 Chr.1: 42155639-42155639 on Build GRCh38
rs374423665 Chr.1: 42155664-42155664 on Build GRCh38
rs772385547 Chr.1: 42155521-42155521 on Build GRCh38
rs370778592 Chr.1: 42155532-42155532 on Build GRCh38
rs968542227 Chr.1: 42155450-42155450 on Build GRCh38
rs973448772 Chr.1: 42155667-42155667 on Build GRCh38
rs761221783 Chr.1: 42155531-42155531 on Build GRCh38
rs769495220 Chr.1: 42155500-42155500 on Build GRCh38
rs148141677 Chr.1: 42155585-42155585 on Build GRCh38
rs776695313 Chr.1: 42155539-42155539 on Build GRCh38
rs531643305 Chr.1: 42155586-42155586 on Build GRCh38
rs974520777 Chr.1: 42155455-42155455 on Build GRCh38
rs80206942 Chr.1: 42155499-42155499 on Build GRCh38
rs954651790 Chr.1: 42155481-42155481 on Build GRCh38
rs753342145 Chr.1: 42155646-42155646 on Build GRCh38
rs889566976 Chr.1: 42155645-42155645 on Build GRCh38
rs1008822713 Chr.1: 42155648-42155648 on Build GRCh38
rs770549769 Chr.1: 42155573-42155573 on Build GRCh38
rs987791547 Chr.1: 42155489-42155489 on Build GRCh38
rs375448042 Chr.1: 42155632-42155632 on Build GRCh38
rs370297949 Chr.1: 42155584-42155584 on Build GRCh38
rs780425991 Chr.1: 42155574-42155574 on Build GRCh38
rs920242807 Chr.1: 42155675-42155675 on Build GRCh38
rs777236388 Chr.1: 42155507-42155507 on Build GRCh38
rs893018169 Chr.1: 42155671-42155671 on Build GRCh38
rs373994919 Chr.1: 42155613-42155613 on Build GRCh38
rs2274919 Chr.1: 42155654-42155654 on Build GRCh38
rs148865510 Chr.1: 42155578-42155578 on Build GRCh38
rs775802875 Chr.1: 42155526-42155526 on Build GRCh38
rs373014813 Chr.1: 42155548-42155548 on Build GRCh38
rs1043138139 Chr.1: 42155620-42155620 on Build GRCh38
rs765343038 Chr.1: 42155543-42155543 on Build GRCh38
rs764724945 Chr.1: 42155514-42155515 on Build GRCh38
rs896829778 Chr.1: 42155628-42155628 on Build GRCh38
rs748957151 Chr.1: 42155513-42155513 on Build GRCh38
rs780874167 Chr.1: 42155492-42155492 on Build GRCh38
rs370998972 Chr.1: 42155551-42155551 on Build GRCh38
rs754999837 Chr.1: 42155552-42155552 on Build GRCh38
rs1044611837 Chr.1: 42155556-42155556 on Build GRCh38
rs781374573 Chr.1: 42155587-42155587 on Build GRCh38
rs777178660 Chr.1: 42155577-42155577 on Build GRCh38
rs182890777 Chr.1: 42155672-42155672 on Build GRCh38
rs201747322 Chr.1: 42155634-42155634 on Build GRCh38
rs377375076 Chr.1: 42155549-42155549 on Build GRCh38
rs982617205 Chr.1: 42155561-42155561 on Build GRCh38
rs867669604 Chr.1: 42155569-42155569 on Build GRCh38

View Full Product Details