Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 40291912-40292139 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLCB2
Entrez Gene ID
5330
Gene Name
phospholipase C beta 2
Gene Aliases
PLC-beta-2
UniGene
Hs.355888
Gene Chromosome Location
Chr.15: 40285496-40307973 on Build GRCh38
Transcripts
NM_001284297.1 NM_001284298.1 NM_004573.2 XM_011521674.2 XM_017022314.1 XM_017022315.1 XM_017022316.1 XM_017022317.1 XM_017022318.1 XM_017022319.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1220909 c.2466C>A p.F822L Chr.15: 40292124-40292124 on Build GRCh38
6554879 c.2484G>A p.T828T Chr.15: 40292106-40292106 on Build GRCh38
6874673 c.2518C>T p.L840L Chr.15: 40292072-40292072 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs547233327 Chr.15: 40292106-40292106 on Build GRCh38
rs1008950183 Chr.15: 40292021-40292021 on Build GRCh38
rs374154929 Chr.15: 40292049-40292049 on Build GRCh38
rs754278172 Chr.15: 40291971-40291971 on Build GRCh38
rs758858279 Chr.15: 40292107-40292107 on Build GRCh38
rs779200914 Chr.15: 40291974-40291974 on Build GRCh38
rs764170129 Chr.15: 40291965-40291965 on Build GRCh38
rs530320621 Chr.15: 40292047-40292047 on Build GRCh38
rs767620462 Chr.15: 40292051-40292051 on Build GRCh38
rs978608798 Chr.15: 40292069-40292069 on Build GRCh38
rs567710337 Chr.15: 40291994-40291994 on Build GRCh38
rs765773836 Chr.15: 40292102-40292102 on Build GRCh38
rs757794566 Chr.15: 40291972-40291972 on Build GRCh38
rs1034934368 Chr.15: 40292012-40292012 on Build GRCh38
rs747600887 Chr.15: 40292030-40292030 on Build GRCh38
rs767816452 Chr.15: 40291957-40291957 on Build GRCh38
rs267604174 Chr.15: 40292077-40292077 on Build GRCh38
rs2290550 Chr.15: 40291963-40291963 on Build GRCh38
rs879061624 Chr.15: 40292002-40292002 on Build GRCh38
rs771037738 Chr.15: 40292044-40292044 on Build GRCh38
rs775721703 Chr.15: 40292060-40292060 on Build GRCh38
rs760809615 Chr.15: 40292064-40292064 on Build GRCh38
rs769292690 Chr.15: 40292036-40292036 on Build GRCh38
rs757739588 Chr.15: 40292098-40292098 on Build GRCh38
rs752714343 Chr.15: 40291959-40291959 on Build GRCh38
rs753937697 Chr.15: 40292094-40292094 on Build GRCh38
rs988707522 Chr.15: 40292099-40292099 on Build GRCh38
rs754560852 Chr.15: 40292016-40292016 on Build GRCh38
rs780298262 Chr.15: 40292112-40292112 on Build GRCh38
rs772877823 Chr.15: 40292037-40292037 on Build GRCh38
rs780968903 Chr.15: 40292023-40292023 on Build GRCh38
rs367824252 Chr.15: 40292042-40292042 on Build GRCh38
rs1003411711 Chr.15: 40291987-40291987 on Build GRCh38
rs773846625 Chr.15: 40292074-40292074 on Build GRCh38
rs764120829 Chr.15: 40292078-40292078 on Build GRCh38
rs903776244 Chr.15: 40291970-40291970 on Build GRCh38

View Full Product Details