Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 137101471-137101732 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC101930307
Entrez Gene ID
101930307
Gene Name
uncharacterized LOC101930307
Gene Aliases
-
UniGene
Gene Chromosome Location
Chr.9: 137101679-137104976 on Build GRCh38
Transcripts
XM_017015447.1 XM_017015451.1 XM_017015452.1 XM_017015453.1 XM_017015454.1 XM_017015455.1 XM_017015456.1 XM_017015457.1 NM_016219.4 NR_045720.1 NR_045721.1 XM_006716945.3 XM_017014239.1

Gene Symbol
MAN1B1
Entrez Gene ID
11253
Gene Name
mannosidase alpha class 1B member 1
Gene Aliases
ERMAN1, ERManI, MANA-ER, MRT15
UniGene
Hs.279881
Gene Chromosome Location
Chr.9: 137086862-137109187 on Build GRCh38
Transcripts
XM_017015447.1 XM_017015451.1 XM_017015452.1 XM_017015453.1 XM_017015454.1 XM_017015455.1 XM_017015456.1 XM_017015457.1 NM_016219.4 NR_045720.1 NR_045721.1 XM_006716945.3 XM_017014239.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5581608 c.1254+6G>A p.? Chr.9: 137101678-137101678 on Build GRCh38
5998807 c.1100G>T p.R367I Chr.9: 137101518-137101518 on Build GRCh38
3655942 c.1110C>T p.S370S Chr.9: 137101528-137101528 on Build GRCh38
7222390 c.1245G>C p.K415N Chr.9: 137101663-137101663 on Build GRCh38
7077307 c.1204C>G p.Q402E Chr.9: 137101622-137101622 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375969077 Chr.9: 137101706-137101706 on Build GRCh38
rs201997182 Chr.9: 137101572-137101572 on Build GRCh38
rs776601946 Chr.9: 137101528-137101528 on Build GRCh38
rs768871118 Chr.9: 137101685-137101685 on Build GRCh38
rs141425542 Chr.9: 137101555-137101555 on Build GRCh38
rs147718581 Chr.9: 137101646-137101646 on Build GRCh38
rs761265286 Chr.9: 137101704-137101704 on Build GRCh38
rs748938366 Chr.9: 137101575-137101575 on Build GRCh38
rs113389928 Chr.9: 137101564-137101564 on Build GRCh38
rs114871830 Chr.9: 137101699-137101699 on Build GRCh38
rs765389465 Chr.9: 137101530-137101530 on Build GRCh38
rs767973131 Chr.9: 137101556-137101556 on Build GRCh38
rs774498402 Chr.9: 137101693-137101693 on Build GRCh38
rs775666191 Chr.9: 137101680-137101680 on Build GRCh38
rs768559795 Chr.9: 137101508-137101508 on Build GRCh38
rs387906885 Chr.9: 137101607-137101607 on Build GRCh38
rs759589788 Chr.9: 137101529-137101529 on Build GRCh38
rs538674051 Chr.9: 137101681-137101681 on Build GRCh38
rs1026723062 Chr.9: 137101653-137101653 on Build GRCh38
rs267602177 Chr.9: 137101707-137101707 on Build GRCh38
rs962328275 Chr.9: 137101700-137101700 on Build GRCh38
rs746937336 Chr.9: 137101666-137101666 on Build GRCh38
rs954780220 Chr.9: 137101687-137101687 on Build GRCh38
rs376314527 Chr.9: 137101701-137101701 on Build GRCh38
rs148707659 Chr.9: 137101534-137101534 on Build GRCh38
rs988741602 Chr.9: 137101703-137101703 on Build GRCh38
rs747965269 Chr.9: 137101659-137101659 on Build GRCh38
rs775798237 Chr.9: 137101586-137101586 on Build GRCh38
rs145704211 Chr.9: 137101591-137101591 on Build GRCh38
rs758251168 Chr.9: 137101660-137101660 on Build GRCh38
rs896060673 Chr.9: 137101601-137101601 on Build GRCh38
rs150891378 Chr.9: 137101557-137101557 on Build GRCh38
rs1028817659 Chr.9: 137101677-137101677 on Build GRCh38
rs571240912 Chr.9: 137101588-137101588 on Build GRCh38
rs749052064 Chr.9: 137101502-137101502 on Build GRCh38
rs747911434 Chr.9: 137101585-137101585 on Build GRCh38
rs376873444 Chr.9: 137101499-137101499 on Build GRCh38
rs773140788 Chr.9: 137101590-137101590 on Build GRCh38
rs762099474 Chr.9: 137101616-137101616 on Build GRCh38
rs777634427 Chr.9: 137101664-137101664 on Build GRCh38
rs761820151 Chr.9: 137101522-137101522 on Build GRCh38
rs764413257 Chr.9: 137101553-137101553 on Build GRCh38
rs773639735 Chr.9: 137101702-137101702 on Build GRCh38
rs201682005 Chr.9: 137101543-137101543 on Build GRCh38
rs778637907 Chr.9: 137101656-137101656 on Build GRCh38
rs1037141680 Chr.9: 137101634-137101634 on Build GRCh38
rs763141969 Chr.9: 137101606-137101606 on Build GRCh38
rs369551783 Chr.9: 137101598-137101598 on Build GRCh38
rs769910765 Chr.9: 137101596-137101596 on Build GRCh38
rs757945040 Chr.9: 137101615-137101617 on Build GRCh38
rs769815555 Chr.9: 137101673-137101673 on Build GRCh38
rs865997258 Chr.9: 137101622-137101622 on Build GRCh38
rs753936942 Chr.9: 137101576-137101576 on Build GRCh38
rs772086046 Chr.9: 137101523-137101523 on Build GRCh38
rs768531325 Chr.9: 137101635-137101635 on Build GRCh38
rs995964458 Chr.9: 137101661-137101661 on Build GRCh38
rs774733147 Chr.9: 137101610-137101610 on Build GRCh38
rs145477274 Chr.9: 137101647-137101647 on Build GRCh38
rs778873022 Chr.9: 137101581-137101581 on Build GRCh38
rs755647661 Chr.9: 137101642-137101642 on Build GRCh38
rs754708614 Chr.9: 137101654-137101654 on Build GRCh38
rs144304981 Chr.9: 137101513-137101513 on Build GRCh38

View Full Product Details