Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 399066-399293 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NME4
Entrez Gene ID
4833
Gene Name
NME/NM23 nucleoside diphosphate kinase 4
Gene Aliases
NDPK-D, NM23H4, nm23-H4
UniGene
Hs.9235
Gene Chromosome Location
Chr.16: 396755-400754 on Build GRCh38
Transcripts
NM_001286433.1 NM_001286435.1 NM_001286436.1 NM_001286438.1 NM_001286439.1 NM_001286440.1 NM_005009.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
970693 c.187A>G p.R63G Chr.16: 399085-399085 on Build GRCh38
3787163 c.202C>A p.L68M Chr.16: 399100-399100 on Build GRCh38
3771946 c.168C>T p.D56D Chr.16: 399066-399066 on Build GRCh38
5747690 c.191G>A p.R64Q Chr.16: 399089-399089 on Build GRCh38
6727351 c.200C>T p.T67M Chr.16: 399098-399098 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763746923 Chr.16: 399258-399258 on Build GRCh38
rs761829233 Chr.16: 399132-399132 on Build GRCh38
rs777814984 Chr.16: 399107-399107 on Build GRCh38
rs749465917 Chr.16: 399124-399124 on Build GRCh38
rs979888820 Chr.16: 399182-399182 on Build GRCh38
rs7185254 Chr.16: 399162-399162 on Build GRCh38
rs757952515 Chr.16: 399247-399247 on Build GRCh38
rs117078323 Chr.16: 399089-399089 on Build GRCh38
rs753423592 Chr.16: 399090-399090 on Build GRCh38
rs546432992 Chr.16: 399239-399239 on Build GRCh38
rs567911292 Chr.16: 399245-399245 on Build GRCh38
rs752431612 Chr.16: 399246-399246 on Build GRCh38
rs754729793 Chr.16: 399097-399097 on Build GRCh38
rs759835422 Chr.16: 399163-399163 on Build GRCh38
rs770609638 Chr.16: 399119-399119 on Build GRCh38
rs904080826 Chr.16: 399271-399271 on Build GRCh38
rs761289832 Chr.16: 399153-399153 on Build GRCh38
rs766652858 Chr.16: 399159-399159 on Build GRCh38
rs764936853 Chr.16: 399167-399167 on Build GRCh38
rs986983587 Chr.16: 399088-399088 on Build GRCh38
rs768855168 Chr.16: 399126-399126 on Build GRCh38
rs1056991320 Chr.16: 399207-399207 on Build GRCh38
rs113157188 Chr.16: 399125-399125 on Build GRCh38
rs945336156 Chr.16: 399234-399234 on Build GRCh38
rs374234687 Chr.16: 399143-399143 on Build GRCh38
rs746983963 Chr.16: 399108-399108 on Build GRCh38
rs186371139 Chr.16: 399205-399205 on Build GRCh38
rs752198810 Chr.16: 399098-399098 on Build GRCh38
rs535226064 Chr.16: 399264-399264 on Build GRCh38
rs781097989 Chr.16: 399123-399123 on Build GRCh38
rs780995233 Chr.16: 399267-399267 on Build GRCh38
rs528572147 Chr.16: 399208-399208 on Build GRCh38
rs904991928 Chr.16: 399135-399135 on Build GRCh38
rs758005852 Chr.16: 399106-399106 on Build GRCh38
rs901077333 Chr.16: 399197-399197 on Build GRCh38
rs961371850 Chr.16: 399093-399093 on Build GRCh38
rs927160422 Chr.16: 399183-399183 on Build GRCh38
rs772166749 Chr.16: 399138-399138 on Build GRCh38
rs552518262 Chr.16: 399204-399204 on Build GRCh38
rs1017536358 Chr.16: 399214-399214 on Build GRCh38
rs2363759 Chr.16: 399178-399178 on Build GRCh38
rs774567745 Chr.16: 399127-399127 on Build GRCh38
rs751530179 Chr.16: 399261-399261 on Build GRCh38
rs777170808 Chr.16: 399142-399153 on Build GRCh38

View Full Product Details